Fetal Bovine Serum Basics
A lab commodity whose quality may often be taken for granted, serum can make—or break—the outcome of all of your cell-based assays. These 5 key subjects will provide you with a strong educational foundation in the use of sera for cell culture.
Biologists who culture cells or tissue know that serum products including fetal bovine serum, or FBS, (also sometimes called FCS, or fetal calf serum) constitute an essential component of culture media for most applications. Serum is often one of the most abundant constituents of cell culture media (typically 2-15% by volume), so its quality can have significant impact on culture success. FBS in particular is the mostly widely-used media supplement for the culture of mammalian cells due to its ready availability, along with its rich supply of growth factors that promote the development and expansion of cells in vitro. Serum also acts as a buffer to the cell culture system against a variety of toxic effects that can disrupt cell growth such as pH change, proteolytic activity, or the presence of endotoxin.
General Biology: how to handle serum in the lab
Anyone who has ever ordered serum for the lab knows that delivery timing is important, as bottles will typically arrive frozen from the supplier. This is because the activity of critical macromolecular factors in serum are best protected by storing serum at -5 to -20°C, and maintaining serum in the frozen state until use will also reduce the incidence of events like the breakdown of lipids. Serum is thawed overnight at 2-8°C to prevent degradation; alternatively, it may be thawed at room temperature with periodic agitation to resuspend the contents. Thawed serum should be mixed thoroughly before being added to cell culture medium. Temperatures above 37°C can compromise serum performance by accelerating the degradation of critical serum factors, and should be avoided in the lab.
Serum can be sensitive to environmental conditions beyond temperature. For example, high intensity/high energy wavelengths of light may degrade the activity of growth factors, which is important to keep in mind when using serum near UV lighting intended to promote sterility in some biosafety cabinets. Typical laboratory lighting conditions will have little to no effect on serum integrity or performance.
Physicians may interpret a cloudy patient sample to be indicative of lipemia, or excess fats in the blood. The same may be true of that cloudy serum in the lab—if bacterial contamination can be ruled out by culturing a sample, cloudy serum is most likely explained by the precipitation of lipids and lipoproteins. This can be minimized by aliquoting bulk stock into single-use tubes to avoid repeated freeze-thaw cycles. Although the presence of the clotting protein fibrin is sometimes blamed for serum cloudiness, serum is prepared from the post-coagulation fraction of blood, and therefore would not typically contain significant amounts of fibrin or other clotting factors.
serum aliquoting
Good practices such as aliquoting serum into single use tubes for frozen storage reduces the potential for repeated exposure to contaminants. Aliquoting also helps to prevent cloudiness that can arise from the breakdown of lipids and lipid-associated macromolecules as a consequence of repeated freeze-thaw cycles.
It’s also useful to realize that lipid content contributing to cloudiness in serum can vary significantly among species.  For example (and perhaps not surprisingly), porcine serum is higher in lipid content than its bovine counterpart.  So, serum from diverse biological sources requires unique handling and additional processing steps at the manufacturing facility to prepare the final product for use in the lab. Suppliers that offer sera from multiple species will therefore have differential protocols in place for processing raw materials in order to provide a consistent, appropriate serum product to the end user, regardless of lipid biology.
World Geography: understanding sourcing and countries of origin
Because serum is a biological product, it is subject to variations that arise from the health of the source animals, which is in turn influenced by dietary, environmental, and global disease factors. One of the most important attributes knowledgeable serum product users therefore seek is the country of origin, or source.
Although the incidence and impact of various species-specific pathogens such as bovine spongiform encephalopathy (BSE) on serum are still being studied, the USDA maintains lists of countries approved for importation of serum that are based in part on up-to-date pathogen incidence data. Serum consumers should therefore always expect suppliers to provide geographical source information for their sera and other products of animal origin.
serum approved countries
The USDA maintains a list of countries approved for importation of serum into the U.S. (guidelines revised annually). This differs from U.S.-origin serum, which is collected from facilities in the U.S. that undergo continuous inspection by the USDA.
International Regulatory Practices
Reputable suppliers will typically employ consensus testing methods specified by the serum industry to ensure that serum supplied to laboratories meets standards for levels of common contaminants that may impact its performance in cell culture. Some common tests are:
Contaminant/
Parameter
Test Standard
(if specified)
Endotoxin Limulus amebocyte lysate (LAL) Test and record
Bacteria/fungi Broth or solid culture of a serum sample Not detected
Viruses (various) PCR, anti-viral antibody screen, cytopathology, hemadsorption
Not detected
Mycoplasma LookOut® kit, mycoplasma detection kits/reagents Not detected
pH meter or standard laboratory pH measurement 6.5 – 8.5 (rec: 7.0-8.0)
Standards for the testing of general serum properties and components have also been established to enable consistent evaluation for osmolality, total protein, hemoglobin, and general performance in culture.
Atlantic horseshoe crabs
Limulus polyphemus, the Atlantic horseshoe crab, is particularly sensitive to the presence of endotoxins produced by the cell wall of Gram-negative bacteria that may also contaminate serum. Suppliers that follow industry guidelines will test for endotoxin in serum using an assay (the LAL, or Limulus Ameobocyte Assay) developed to mimic this sensitivity.
History: the importance of traceability and documentation in the serum industry
While “origin” indicates the country in which serum was collected, “traceability” refers to how well the journey of a serum product---from collection at site of origin through manufacturing and distribution---has been documented.
Serum delivered frozen to a processing facility is thawed and tested for the presence of bacteria, virus, endotoxin, and for hemoglobin content. The best quality serum is then filtered through a sequence of membrane filters, terminating with a 0.1 µm pore filtration under aseptic conditions. After filtration, aseptic procedure is maintained as serum is dispensed into sterile bottles, quickly frozen to -20°C and quarantined until all lots have been shown to meet or exceed quality control standards. A certificate of analysis for each lot should always be available upon request from the supplier.
Traceability for biologics is critical to ensure that label descriptions are an accurate representation of product history and contents.  Complete documentation ensures that both the customer and regulatory agencies can audit the processing of serum products, and can uncover whether suppliers are engaging in serum pooling to offset local supplier cost fluctuations---a practice which can compromise performance.
Economics 101: factors that may affect serum pricing
Serum is a commodity product for which price varies among countries of origin. For example, the cost of serum originating from Australia and New Zealand may be higher than that from other countries.  The OIE’s current Geographical Risk Assessment rating for these countries is high because they continue to be free of BSE and FMD (foot and mouth disease), in part due to geographical isolation. This, coupled with strict agricultural regulatory practices, may increase demand for serum products from the ANZ region, which in turn can drive up price.
The availability of FBS and other animal sera is inherently subject to fluctuations in global animal disease states, climate, and food and water supply. These factors make industry-wide serum pricing sensitive in ways that pricing for non-biological reagents is not. And last—but certainly not least—as cell culture becomes more prevalent, particularly in the pharmaceutical sector, serum pricing is sure to reflect the changes in supply this demand may precipitate.