Freezing and Thawing FBS and Other Sera
Best practices for ensuring stability and performance of FBS and sera
Animal serum is an ideal source of nutrients for cells in culture because it contains proteins, lipids, salts, vitamins, minerals, amino acids and other components necessary for growth. When stored and handled correctly, the performance characteristics of serum can be maintained for many years. Improperly storing and/or thawing serum products can decrease not only their immediate and long-term stability, but also their effectiveness.
Growth-promotion data demonstrate that serum maintains its growth-supporting characteristics throughout its shelf life when stored correctly. The table below contains stability data for Fetal Bovine Serum (FBS) for a five-year period. To assure stability, sera must be appropriately stored and handled.
Long-term Storage of SERA: Test Results
Product Test Year 0 Year 1 Year 2 Year 3 Year 4 Year 5
Irradiated FBS Multiple Passage 110 104 116 126 93 90
Plating Efficiency 61 36 51 35 40 50
Cloning Efficiency 35 43 31 21 25 23
FBS Multiple Passage 113 103 99 95 103 102
Plating Efficiency 59 58 57 74 32 42
Cloning Efficiency 49 56 42 47 28 30
Multiple Passage is % of control growth, while Plating Efficiency (PE) and Cloning Efficiency (CE) are % PE and CE.
How to freeze and thaw FBS and other serum to safeguard its stability
To efficaciously preserve the integrity of animal serum, it should be stored frozen and protected from light. The recommended storage temperature is -10 to -40 °C. At temperatures below -40 °C, storage bottles may become brittle, increasing the risk of breakage.
Multiple thaw/freeze cycles should be avoided as they will hasten the degradation of serum nutrients and can induce the formation of insoluble precipitates. For this reason, serum should never be stored in "frost-free" freezers. These appliances occasionally warm themselves to prevent internal ice deposits,and are therefore detrimental to the clarity and stability of frozen serum products.
Recommended Thawing Procedure for FBS and other sera
  • Remove the serum bottles from the freezer and allow them to acclimate to room temperature for approximately 10 minutes.
  • Place each container in a 30 to 37 °C water bath or incubator. Higher temperatures will degrade heat-labile nutrients. If using a water bath, do not submerge the bottles beyond the level of the serum content, to prevent the bottle caps from being submerged. Water baths are common sources of contamination for media and serum.
  • Gently swirl or shake the bottles every 10 - 15 minutes until the serum is completely thawed.
Avoid cryoprecipitates when thawing serum (FBS)
The method used to thaw serum is vital to its optimum performance. The key to proper thawing is periodic agitation. If a bottle of serum is not regularly shaken or swirled as it thaws, salt and protein gradients will form throughout the liquid fraction. Within these gradients are high concentrations of salts, proteins, and lipids which can lead to the formation of crystalline or flocculent precipitates. These "cryoprecipitates" are not toxic to cell cultures, but they affect the appearance and consistent composition of the FBS or other animal sera.
Occasional, sparse cryoprecipitates are not uncommon, even in serum that is thawed using the recommended procedure. This is normal and will not affect product performance.
If serum is thawed incorrectly, increased cryoprecipitate may form, and is often insoluble. Filtering serum to remove cryoprecipitates is not recommended as it could result in the unintentional loss of nutrients, such as growth factors, mitogens, and other proteins.
Protect thawed FBS and other sera with safe refrigeration and storage
Use thawed serum promptly. Liquid FBS and other serum may be stored refrigerated (2 to 8 °C) up to four weeks. To avoid thaw/freeze cycles or discarding unused serum after more than four weeks of refrigeration, it is recommended that any unneeded serum be immediately dispensed into small, application-appropriate aliquots under sterile culture conditions, and refrozen to protect stability for future use.
For more information about serum freezing, thawing, and storage or our wider products and services, please contact our Technical Services department here.