Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Centriprep® Centrifugal Filters
Concentration of samples with high solute content
Centriprep® Centrifugal FiltersCentriprep® centrifugal filters are disposable ultrafiltration devices for purification, concentration and desalting of biological samples (2-15 mL volume range). The filters have a high flow rate and are easy to use, fitting standard swinging bucket rotors for 50 mL tubes.
Features and advantages of Centriprep® Filters
  • Twist-lock, air-seal cap for sample isolation and security
  • Low-absorptive Ultracel® regenerated cellulose membrane for high macromolecule recovery
  • Unique, inverse flow mode of operation with large deadstop
NOTE:  Centriprep® centrifugal filter devices with Ultracel® 10K and 30K membranes are approved for in vitro diagnostic use.
Centriprep® Centrifugal Filters
Product No. Description Pack Size Add to Cart
4302 Centriprep Centrifugal Filter Unit with Ultracel-3 membrane 24
4303 Centriprep Centrifugal Filter Unit with Ultracel-3 membrane 96
4304 Centriprep Centrifugal Filter Unit with Ultracel-10 membrane 24
4305 Centriprep Centrifugal Filter Unit with Ultracel-10 membrane 96
4306 Centriprep Centrifugal Filter Unit with Ultracel-30 membrane 24
4307 Centriprep Centrifugal Filter Unit with Ultracel-30 membrane 96
4310 Centriprep Centrifugal Filter Unit with Ultracel-50 membrane 24
4311 Centriprep Centrifugal Filter Unit with Ultracel-50 membrane 96