Critical Micelle Concentrations (CMCs) - Avanti® Polar Lipids
Definition
The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn
CMC Values
Products not available in the USA
Lipid CMC (mM)
5:0 PC 90
6:0 PC 15
7:0 PC 1.4
8:0 PC 0.27
9:0 PC 0.029
10:0 PC 0.005
12:0 PC 90 nM
14:0 PC 6 nM
16:0 PC 0.46 nM
8:0 PG 1.21
10:0 PG 0.42
12:0 PG 0.13
14:0 PG 0.011
4:0 PS (5mM Ca2+) 4
5:0 PS (5mM Ca2+) 2
6:0 PS (5mM Ca2+) 1.2
7:0 PS (5mM Ca2+) 0.8
8:0 PS (5mM Ca2+) 0.4
8:0 PS 2.282
10:0 PS 0.096
8:0 Lyso PC 60
10:0 Lyso PC 6-8
12:0 Lyso PC 0.4-0.9
14:0 Lyso PC 0.043-0.090
16:0 Lyso PC 4-8.3 µM
18:0 Lyso PC 0.4 µM
† Data provided by Dr. Rinku Majumder, Department of Biochemistry, The University of North Carolina – Chapel Hill.
Lipid Acronyms Key
PC = phosphatidylcholine
PG = phosphatidylglycerol
PS = phosphatidylserine
Lipid Acronyms Key
CMC’s were determined in house by using a Pyrene Fluorescence technique or were obtained from the literature. Many of the literature values were confirmed by our in-house method.
The method is generally outlined by: Fluorescent Probes For Critical Micelle Concentration
References
  • Handbook of Lipid Bilayers by Derek Marsh. ISBN 0849332559. CRC Press, Inc.
Materials