Pure and Simple Primers
Order Pure and Simple Primers, and you will be assured a quality start to all of your research applications with our ultra-high base coupling efficiency and optimized cartridge purification process.
Guaranteed Yield 3 OD
Format Shipped dry in 2 mL screw cap tubes
Length 12 - 35 bases
Purification Reverse-phase cartridge
Bases DNA (A, C, G or T)
Modifications None
Backbone Phosphodiester bond
Documentation All oligos are delivered with Quality Assurance documentation including yields, Tm (melting temperature), MW, and µg/OD
Quality Control Every oligo is quality checked by mass spectrometry
Shipping Schedule Ships within 24 - 48 hours
Price & Ordering Contact your local office for pricing and availability
Materials