Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
D-Tube™ Dialyzers
Gently and easily dialyze protein or nucleic acid samples using D-Tube™ dialyzers. These devices feature dialysis membrane windows in a capped centrifuge tube format, enabling efficient buffer exchange to remove urea, detergents, salt, and other microsolutes.
Designed with a double membrane, D-Tube™ dialyzers spread the sample over a large area, so that dialysis is complete in 2-5 hours.
D-Tube™ Mini Dialyzer
D-Tube™ Midi Dialyzer
D-Tube™ Maxi Dialyzer
Advantages of D-Tube™ Dialyzers
  • Low-binding regenerated cellulose membrane and housing enhance sample recovery
  • Leak-resistant design — no clamps or knots required
  • Screw cap makes it easy to open and close
  • Easily withdraw even submilliliter volumes, from the rigid frame of the device
  • No syringe required to load or withdraw samples. Simply load your sample with standard pipette tip.
  • Float racks of D-Tube™ dialyzers in most standard-sized beakers containing exchange buffer
  • Use D-Tube™ dialyzers for electroelution of samples from agarose or acrylamide
Select the D-Tube™ dialyzer that’s right for your sample.
Select devices are now available on our website. Check back periodically for updates.
      D-Tube™ Mini D-Tube™ Midi D-Tube™ Maxi D-Tube™ Mega D-Tube™ Mega
      Maximum initial sample volume
MW (Proteins or Nucleic Acids) Nominal MW Cutoff Qty/pk 10 to 250 μL 50 to 800 μL 100 μL to 3 mL 3 mL to 10 mL 10 mL to 15 mL
MW < 7 k 3500
3500
10
50
  71506-M 71508-M Coming soon Coming soon
7 k < MW < 24 k 7000
7000
7000
10
50
1 plate of 96
71504-M
71712-M
71507-M 71509-M Coming soon
Coming soon
Coming soon
Coming soon
Coming soon
Coming soon
24 k < MW 13000
13000
13000
10
50
1 plate of 96
71505-M
71713
  71510-3    
Floating Rack     Mini (10) Midi (10) Maxi (10) Mega (10) Mega (10)
      71512-M 71513-M 71514-M 71748-M 71748-M
Related Links