Diffinity RapidTip® Technology
Description
The Diffinity RapidTip pipette tips contain everything needed for PCR purification. The functional tips contain a proprietary adsorption technology that has a differential affinity for PCR reaction components.
  • Fast, Simple – Recovers clean DNA in a one minute, one step process
  • Effective Purification – Removes impurities for downstream applications
  • Cost Effective – Requires no capital equipments, extra plastic or liquids
  • Environmentally Friendly – Limits waste to the functional pipette tip
Available in Two Formats
Diffinity RapidTip® Pipette Tip
Diffinity RapidTip®2 Pipette Tip
  • Removes 90% of primers, ssDNA, and primer-dimers
  • Application: standard (Sanger) sequencing
  • Removes 90% of primers, ssDNA, primer-dimers and DNA polymerase
  • Application: Restriction digest, T-A cloning and standard (Sanger) sequencing
Ordering Information
Diffinity RapidTip® Pipette Tip
Diffinity RapidTip®2 Pipette Tip
D1947
D1947L
for PCR Purification
for PCR Purification, Compatible with Rainin LTS™ pipettes
D2947L for PCR Purification with Polymerase Removal, Compatible with Rainin LTS™ pipettes
(Each product is available in 8 reaction, 48 reaction, and 96 reaction quantities.)
RapidTip is a registered trademark of Diffinity Genomics, Inc.
LTS is a registered trademark of Rainin Instrument, LLC.
Related Links