Duolink® Tips and Tricks
1. Experiment
Sample and Antibodies
Sample fixed on glass slides
Optimized:
  • Pretreatment
  • Blocking
  • Primary antibodies, diluent, titer
Protocol: Your own or take a working one and optimize
E.g. DAKO Education Guide, IHC world, Nordiq, Abcam etc.
Primary Antibodies:
  • Affinity purified polyclonal
  • Monoclonal (avoid ascites)
Duolink In Situ Assay: Equipment Needed
Biological Controls
Best and recommended controls include:
Positive control:
  • Known protein interaction or modification
  • Overexpression, stimulation
Negative controls:
  • Irrelevant protein (e.g. non-interacting, present in different compartment)
  • Sample not expressing target protein or down-regulated/repressed, silencing, mutation etc.
Lower HER2 in MDA Cells
Duolink In Situ Assay: Technical Controls
Technical negative controls:
  • Leave one primary antibody out
  • Information about unspecific binding of primary antibodies
    • Helpful to find an optimal titer
    • Leave both primary antibodies out
  • Information about non specificity of PLA® probes
    • Do not use unspecific sera!
Duolink In Situ Control Kit:
  • Useful to check assay conditions in your lab
  • Positive control of Detection Reagent
Above: positive control; Below: negative control.
Left: Brightfield; Right: Fluorescence.
Common Parameters to Consider
Check antibody titer to obtain specific and distinct staining
  • Use appropriate blocking agent. Dilute PLA probes in a buffer containing the blocking agent
  • Never let your sample dry out during the Duolink In Situ assay
  • Gently tap-off excess solutions
  • Wash in at least 70 mL of wash buffers
  • Use Wash Buffers A and B where specified
Incubation temperature:
Wash temperature:
  • Bring wash buffers to room temperature
  • Perform wash at room temperature
Amplification:
Amplify for 100 min to avoid coalescent signals
Proper thawing technique:
Thaw reagents but use freeze block for Ligase and Polymerase
Image Interpretation: Common Autofluorescence Interference
  • Use Wash B and Duolink In Situ Mounting Medium with DAPI
  • Do not overexpose
Autofluorescence
  • Inherent to sample
  • Can be observed in green channel
  • Alternative: Use either FarRed or Brightfield Detection Reagents
2. Sample Analysis and Storage
Mounting and storage
  • Duolink In Situ Brightfield Mounting Medium: non-aqueous, xylene based
  • Duolink In Situ Mounting Medium with DAPI: aqueous, contains anti-fade and DAPI for Fluorescence application
Notes:
We have tested many other media and PLA signals fade away with many of them.
Add a few drops and press out any excess media.
Image Acquisition: Filters
Image Acquisition: Parameters
Brightfield Application
  • Good contrast
  • White background / unstained tissue
Fluorescence Application
  • 20x or 40x objective
  • Do not overexpose
    • Signals can coalesce
    • Can give rise to autofluorescence
  • Z-stacks if possible
3. Image Analysis and Presenting Data
Quantification
Compare only samples that have been run in parallel
  • Images taken with same acquisition parameters under same session
Use 20x or 40x
  • 63x or 100x can give nice images for publication but not worth quantifying
Quantification is relative, e.g.
  • Positive vs. negative controls
  • Signals in nuclei vs. signals in cytoplasm
Counterstaining
Add counterstaining after Wash buffer B step in the protocol, wash and mount
Presenting Data
  • Zoom in and scale up
  • Increase brightness and contrast for presentations and publications
Zoom in on a region of interest.
Scale up
Example on how increasing brightness/contrast (numerical values above) can be useful when presenting data.
4. Probemaker
Probemaker: antibody requirements
Good quality antibodies
  • Affinity purified polyclonals
  • Non-ascites monoclonals
  • Non-modified (e.g. no biotinylation)
  • Stock 1 mg/mL
  • Stock buffer additive free, ideally PBS. Pretreatment
Buffer exchange
G25, G50
Small molecules: e.g. azide, Tris
Affinity purification
Protein A, G
Macromolecules: e.g. gelatin, BSA
Probemaker: assay
Concentration of conjugated antibody
Dilution of conjugated antibody
  • Custom solutions: blocking agents + Assay Reagent
  • Duolink In Situ solutions: PLA probe Diluent
Contact for Support
You are welcome to contact us for further support requests.
When contacting us, please, provide the following information:
  • Description of your assay
  • Images of your results
  • Controls (positive/negative, biological/technical) that have been performed (if any)
  • Previous IF/IHC results (if any)