Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Evaluation of Recombinant, Chemically Treated Trypsin in Proteomics and Protein Characterization Assays
By: Judy Boland, Nicolas Caffarelli, Gordon Nicol, Amber Henry, Jeffrey L. Turner, and Kevin Ray, MilliporeSigma, A business of Merck KGaA, Darmstadt, Germany, St. Louis MO 63103
Overview
  • We compared recombinant trypsin to native-source in various MS workflows.
  • Native source trypsin is dimethylated to prevent autolysis and TPCK treated to inhibit chymotrypsin.
  • Native and recombinant trypsins gave similar amounts of proteins and peptides identified in a complex mixture.
  • TIC of a single-protein digest revealed more complete peptide processing by recombinant trypsins relative to native trypsin.
  • Trypsin dimethylation leads to slower processing but with fewer autolytic peptides present.
Introduction
Porcine Pancreatic Trypsin
Porcine Pancreatic Trypsin
Proteomics grade trypsin is:
  1. Dimethylated (prevent autolysis)
  2. TPCK treated (inhibit chymotrypsin)
Dimethylated
Are there better options?
Materials and Methods
Materials and Methods
Results
SOLu-Trypsin Stability
SOLu-Trypsin Stability
Human Protein Extract Digest
Trypsin Average number of proteins identified
37°C for 18 hr 45°C for 2 hr
Proteomics
Grade Trypsin
2,021 2,167
SOLu-Trypsin 2,055 2,128
SOLu-Trypsin
Dimethylated
2,025 2,162
Human protein extract was digested using native and recombinant trypsins in a traditional overnight 37°C digest and a rapid 2 hr 45°C protocol. In all cases, similar numbers of proteins were identified.
Apolipoprotein A-1 Digest
Apolipoprotein A-1 Digest
To Dimethylate or Not Dimethylate
To Dimethylate or Not Dimethylate
Conclusions
  • Recombinant trypsin digests yielded comparable amounts of peptides and proteins identified across all conditions tested versus standard sequencing-grade trypsin.
  • Dimethylation was shown to reduce the presence of autolytic fragments, whereas unmodified enzyme yielded faster processing of certain cut sites.
  • SOLu-Trypsin is solution-stable for >49 days at 37°C and >800 days at 4°C based on accelerated stability studies.
  • Recombinant solution-stable trypsin can be used in lieu of native trypsin with no changes to work flow.