EX-CELL® Advanced™ CHO Fed-batch Medium
Chemically Defined, Animal-Component Free Medium for CHO Cells
Without L-glutamine, without hypoxanthine, and without thymidine
Liquid Catalog No. 14366C
Powder Catalog No. 24366C
EX-CELL® Advanced™ CHO Fed-Batch Medium is a chemically defined, next generation media platform. The formulation was developed using multivariate analysis of 10,000+ data points that included performance, physical, regulatory and safety design specifications. This medium is designed to be used in conjunction with Advanced™ CHO Feed 1 for superior platform performance in fed-batch cultures on all industrial CHO cell lineages (CHO-S, DuxB11, DG44, CHO-M, and CHOZN GS).
Intended Use
This product is intended For Further Manufacturing in the bio-manufacturing industry. It is not intended or approved for in-vitro diagnostic use in the human or veterinary industries.
Product Handling & Storage
Do not use if liquid medium is cloudy or contains precipitates. Powder product should be stored dry at 2-8°C and protected from light for up to 2 years. Liquid product should be stored at 2-8°C and protected from light for up to 1 year. Use aseptic technique when handling or supplementing this medium. Read the Safety Data Sheets (SDSs) and follow the handling instructions. Wear appropriate protective eyewear, clothing, and gloves.
Methods for Use
Preparation Instructions
EX-CELL® Advanced™ CHO Fed-Batch Medium is formulated without L-glutamine and without hypoxanthine/thymidine. Aseptic supplementation instructions:
  1. Add L-glutamine (Catalog No. 59202C), 2–8 mM final concentration, before use in applications not requiring GS selection.
  2. Add HT Media Supplement (Catalog No. H0137) before use in applications not requiring DHFR selection.
Note: Shelf life of the product may be affected by the nature of the supplements.
Initiating Cultures
  1. Rapidly thaw (<1 minute) a vial of frozen cells in a 37°C water bath.
  2. Transfer the entire contents aseptically into a 125-mL shake flask containing 30 mL prewarmed complete EX-CELL® AdvancedTM CHO Fed-Batch Medium.
  3. Incubate at 37°C in a humidified atmosphere of 5% CO2 in air on an orbital shaker platform (19mm diameter orbit) rotating at 120–140 rpm.
  4. Maintain cell density between 0.5–1 ×106 viable cells/mL for the first two passages following recovery; thereafter, return to your normal maintenance schedule.
  1. Verify that the incubator is set to 37C, 5% CO2, and has water for humidity control (~80%).
  2. Pre-warm complete EX-CELL® Advanced™ CHO Fed-Batch Medium to room temperature.
  3. Aseptically remove a small volume of cell culture sample from the flask and count by trypan blue exclusion using a hemocytometer or an automated cell counter. Do not proceed if cell viability is less than 90%. Note: If cell viability is below 90%, troubleshoot conditions prior to continuing.
  4. Determine the correct volume of cell culture to inoculate a new flask at a starting cell density of 2-3 x105 cells/ml in a total volume of 30 mL fresh EX-CELL® Advanced™ CHO Fed-Batch Medium per 125-mL shake flask.
  5. Aseptically transfer the appropriate amount of cells to the new flask, and add pre-warmed medium up to the desired volume.
  6. Incubate flasks in a humidified 37°C incubator with 5% CO2 on an orbital shaker at 120-140 rpm.
  7. Passage cells by repeating the above steps at least twice weekly, and expand culture volume as necessary. Note: When passing the cells, medium carryover should not exceed 25% of the final volume.  If carryover exceeds 25%, centrifugation is recommended.
  1. Prepare the desired quantity of cells, harvesting in mid-logarithmic phase of growth with viabilities >90%.
  2. Prepare a freezing medium consisting of 46.5% cold EX-CELL® Advanced™ CHO Fed-Batch Medium, 46.5% conditioned medium and 7% dimethyl sulfoxide (DMSO).
  3. Harvest cells by centrifugation at 200 g for 5 minutes. Remove the supernatant.
  4. Resuspend cell pellet in the freezing medium at 5 x106 to 1 x107 cells/mL.
  5. Rapidly transfer 1 – 2 mL of this suspension to sterile cryovials.
  6. Place the vials in a controlled rate freezing apparatus following standard procedures (1°C decrease per minute).
  7. For long-term storage, transfer the vials to liquid nitrogen (vapor phase).
How to Order
For additional information, please contact your Regional representative or call Customer Service.
Related Links