pSF-FLAG® Vectors from Oxford Genetics
FLAG and 3xFLAG Expression Vectors
The classic FLAG and 3xFLAG expression vectors have been converted to SnapFast™ Expression Vectors – combining two highly functional systems into one product. Additional information about the SnapFast™ system includes the Background & Popular Vectors, Selection Charts, and the SnapFast™ FAQs.
The FLAG and 3xFLAG vectors are available for bacterial and mammalian expression, using T7, Tac or CMV promoters. Vectors have an enterokinase cleavage site for removal of the tag from purified protein. Together with c-myc, some vectors offer dual-labeling. See charts below for quick-reference vector specifications, and click the links to the product pages for more information, including vector maps.
Bacterial vector options with FLAG and 3xFLAG tags
Product No. FLAG tag OmpA secretion tag Ampicillin Selection Marker EKT cleavage tag Promoter
N-terminal C-terminal Inducible TAC T7 Lac
OGS632      
OGS633    
OGS634      
OGS635    
OGS636      
Mammalian vector options with FLAG and 3xFLAG tags
Product No. FLAG tag 3X FLAG tag C-myc tag EKT cleavage tag PPT secretion tag Ampicillin Selection Marker (Bacterial) Neomycin Selection Marker (Mammalian) Promoter
N-terminal C-terminal N-terminal C-terminal C-terminal Constitutive CMV
OGS618            
OGS619            
OGS620            
OGS621          
OGS622        
OGS623          
OGS624        
OGS625          
OGS626        
OGS627          
OGS628        
OGS629          
OGS630      
OGS631        
FLAG: a proven system for detection and purification of proteins
The FLAG Expression System is an established way to express, purify and detect recombinant fusion proteins. FLAG and 3xFLAG have proven utility in numerous applications such as Western blotting, immunocytochemistry, immunoprecipitation, flow cytometry, protein purification, and in the study of protein-protein interactions, cell ultrastructure and protein localization. These small hydrophilic tags facilitate superior detection and purification of recombinant fusion proteins when using our highly specific and sensitive ANTI-FLAG® antibodies. Ideal epitope tags are small, hydrophilic and cleavable.
The FLAG expression system utilizes a short, hydrophilic 8-amino acid peptide that is fused to the recombinant protein of interest. Because of its hydrophilic nature, the FLAG peptide is likely to be located on the surface of the fusion protein. As a result, the FLAG peptide is easily accessible for cleavage by enterokinase (Ek) and for detection with antibodies. In addition, because of the small size of the FLAG peptide tag, it is not likely to obscure other epitopes, domains, or alter function, secretion, or transport of the fusion protein.
3xFLAG: the most sensitive epitope tag expression system
The 3xFLAG system is an improvement upon the original system by fusing 3 tandem FLAG epitopes (22 amino acids). Detection of fusion proteins containing 3xFLAG is enhanced up to 200 times more than any other system. Like the original FLAG tag, 3xFLAG is hydrophilic, contains an Ek cleavage site, and is relatively small. Therefore, the risk of altering protein function, blocking other epitopes or decreasing solubility is minimized.
Low-level expression of recombinant proteins in mammalian cells is a common problem that often makes detection of epitope tags difficult or impossible. Detection of 3xFLAG using the ANTI-FLAG® M2 Ab and anti-mouse-HRP is 20-200 times more sensitive than any commonly used tag. An extensive selection of p3xFLAG vectors utilizes the strong CMV promoter to drive expression.
3xFLAG
Western blot of purified 3xFLAG bacterial alkaline phosphatase and FLAG bacterial alkaline phosphatase transferred on to nitrocellulose membrane. Detection was performed with ANTI-FLAG M2 monoclonal antibody as primary, anti-mouse-HRP seconday antibody, and ECL™ chemiluminescent substrate.
3xFLAG
C-terminal fusions of GST and the tags listed above were prepared and analyzed on Western Blots. All proteins were detected using the appropriate tag antibodies, anti-mouse HRP and ECL.
Highly Specific Antibodies to FLAG and 3xFLAG epitopes
The FLAG and 3xFLAG sequences include the binding sites for several highly specific ANTI-FLAG monoclonal (M1, M2, M5) and polyclonal antibodies and conjugates, each with different recognition and binding characteristics. ANTI-FLAG antibodies exhibit little or no cross-reactivity in most mammalian and bacterial cell lysates. One-step purification formats include affinity gels and 96-well plates.
Note: if you have used a FLAG vector in the past, this chart will help you pick the closest SnapFast™ FLAG and 3xFLAG vector.