Guava® easyCyte Flow Cytometers
Guava easyCyte Flow CytometersGuava benchtop microcapillary flow cytometers now provide up to 14 parameters of detection, with more powerful violet, blue, green, and red lasers for exceptional sensitivity to enable detection of small particles and rare signals.
Discover the Versatility of Microcapillary Flow Cytometry
Download our Guava Brochure
Advantages of Guava Flow Cytometers
Compared to traditional sheath fluid-based instruments, Guava systems consume less sample, produce less liquid waste (<50mL in 8 hours of continuous use), and are easier to use.
Other advantages shared by all Guava instruments:
  • Automated sampling option handles 96-well plates or tubes
  • Cost-effective single sample-loading option
  • Microcapillary enables direct, absolute cell counts – no need for reference beads
  • Laser and detector configurations are compatible with many fluorochromes
  • Intuitive InCyte™ software makes it easy to import settings, set gates, construct dose response curves, and use heat maps to compare plate-based experiments
What instrument type, laser configuration, and options are the best suited to your lab, your budget, and your research needs? Ask to be contacted by one of our cellular specialists, who can guide you to the best solution, and refer to the instrument comparison table below.
Guava Instrument Comparison Table (Laser Configurations)
  Wavelength of Excitation Laser
Model of Guava easyCyte instrument 405 nm 488 nm
(50 mW)
488 nm
(150 mW)
532 nm 642 nm
5, 5HT        
5HPL, 5HPL HT        
6-2L, 6HT-2L      
8, 8HT      
BG HT      
11HT      
BGR HT    
12HT    
BGV HT    
Since 1998, Guava® benchtop flow cytometers have been helping scientists obtain insightful cellular analysis
Free Poster
Track cell health with our new poster, illustrating pathways of mitochondrial damage, cell cycle, DNA damage, autophagy, apoptosis, and more.
   REQUEST POSTER  
Free Poster
The immunology of Cancer.
   REQUEST POSTER