Glycine Hydrochloride
PharmaGrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. Suitable for cell culture.
Product Regulatory Datasheet
1. General Product Information
Product Name Glycine hydrochloride
Product Number PHG0016
Qualifier PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. Suitable for cell culture.
SAFC Quality Level ELITE
Grade Non-Compendial
1. Manufacturing, Packaging, Release Site and Supplier Information
  • Site of manufacturing, packaging and product release:
    3300 S. Second St., St. Louis, MO 63118 USA, internally referred to as the Cherokee site and
    3500 Dekalb St., St. Louis, MO 63118 USA.
  • This product is manufactured under appropriate controls to be used in pharma or biopharmaceutical production.
  • This product is manufactured using multi-purpose equipment.
2. Physico-chemical Information
2.1 Synonyms
  • Aminoacetic acid
  • Aminoethanoic acid
  • Glycocoll
2.2 Structure Formula
2.3 Molecular Formula
  • C2H5NO2 • HCl
2.4 Molecular Mass
  • 111.53
2.5 CAS Number
  • 6000-43-7
2.6 Origin
  • The product PHG0016 is manufactured by organic synthesis. Only raw materials of synthetic origin are used
2.7 Manufacturing process
  • Formation, filtration, distillation, and subsequent crystallization of Glycine Hydrochloride from Glycine free base;
    centrifugation of crystallized material, drying, and homogenization.
  • Equipment cleaning is verified.
  • While this product does not officially fall under ICH Q7A Good Manufacturing Practice for Active Pharmaceutical
    Ingredients, manufacturing process design is based on this standard and uses Failure Modes and Effects Analysis
    (FMEA) process where Critical Quality Attributes and Critical Control Points controlling them are identified. Any
    changes to these parameters, or deviations around them, are appropriately evaluated.
2.8 Specification (Note: the section should match Spec sheet on Product Description web page, if available. Also an active web link to the specification sheet file on the Product Description web page can be used rather than using the enclosed table).
Analytical items Specifications
Appearance (Color) White
Appearance (Form) Powder
Solubility (Color) Colorless
Solubility (Turbidity)
    25g plus 50mL water
Clear
Infrared Spectrum Conforms to Structure
Loss on Drying ≤ 1.0%
Purity (HPLC) ≥ 99%
Purity (Titration by NaOH) ≥ 98%
Iron (Fe) ≤ 10 ppm
Arsenic (As) ≤ 3.0 ppm
Heavy Metals (as Lead) ≤ 10 ppm
Ethanol ≤ 5000 ppm
Endotoxin ≤ 50 EU/g
Total Aerobic Microbial Count ≤ 100 CFU/g
Total Yeast and Mold ≤ 100 CFU/g
Cytotoxicity Pass
Recommended Retest Period 1 Year
2.9 Batch results
  • Certificates of Analysis are available for each lot produced.
3. Regulatory Information
3.1 Compendial compliance
  • Not applicable
3.2 Master file
  • Drug Master File (DMF) is not available for this product.
  • EDQM Certificate of Suitability is not currently available for this product.
  • Please contact us for further regulatory support.
3.3 BSE/TSE and viral risk statement {select most applicable description}
  • This product is not produced using components from animal species (bovine, ovine or caprine) known to harbor
    BSE/TSE, and is therefore considered of negligible risk. The buffers facility and its equipment are animal-component
    free.
3.4 Allergen information {select most applicable description}
  • This product is not produced with any known major food allergens (as defined by the FDA and WHO).
3.5 GMO information {select most applicable description}
  • This product is not derived from viable Genetically Modified Organism (GMO).
3.6 Residual solvents information {select most applicable description}
  • Glycine Hydrochloride is produced using Ethanol, as class 3 solvent.
3.7 Metal catalyst and metal reagent residues {select most applicable description}
  • Glycine Hydrochloride is produced without use of metal catalysts or metal reagents.
  • Contact surfaces during production include glass, stainless steel, Teflon-lined hoses, and titanium.
3.8 Kosher/Halal status
  • No status.
3.9 Melamine statement {select most applicable description}
This material is not assayed by a test that detects nitrogen. This product is considered negligible risk for melamine
contamination.
4. Miscellaneous Product Information
4.1 Explanation of the lot/batch numbering system
  • The product is manufactured in a batch process.
  • Batch size is approximately 400-900 kg.
  • Each produced batch is identified by a specific, unique batch number.
  • The batch number consists of 4 characters followed by four digits and a ‘V’, e.g., SLBB1234V.
4.2 Expiration date and/or recommended re-evaluation interval
  • The recommended retest period of 1 year.
4.3 Handling, Shipping Conditions and Long-term Storage Recommendation
  • If stored in the original unopened containers under the specified conditions, no significant change of the material can be observed over the retest period
  • Storage and shipping conditions:
    o Storage at ambient temperature in a dry storage place.
    o Shipping at ambient temperature
4.4 Packaging
  • SAFC Cherokee can accommodate various package sizes.
  • Typical materials of construction consist of: 1. Retail sizes HDPE plastic containers. 2. Bulk quantities HDPE Plastic
    Drums with double lined LDPE liners.
5. Revision History
  • Managed in the Electronic Document Management System (EDMS).
Biological Buffers Facility Quality Overview
Our facilities that are able to adapt their supply chain and quality management systems to support your raw materials supply requirements for manufacturing use.
Our buffers are fully scalable regardless of need or requirement and are manufactured under stringent quality systems that meet your regulatory needs. Packaging, testing and labeling can be customized to your specific requirements.
1. Facility overview
 Scope
  • Biological buffers facility, 3300 S. Second St., St. Louis, MO 63118 USA
Customer audit policy
  • Customers are welcome to audit our sites depending upon the business value.
  • All inspections are planned, coordinated and accompanied by Quality Assurance; a thirty-day notice and confidentiality agreement may be required.
Site details
  • General site information:
    - 390,000 ft2 (36,000 m2) total for site; 35,000 ft2 (3,300 m2) for buffers
    - Buffers manufactured since 1970; manufactured at this site since 1986
    - Manufacturing operates continuously, seven days/week (24/7), with biannual preventive maintenance shutdowns
  • Site activities conducted:
    Chemical manufacture, special testing (such as cell culture and microbial testing), packaging, raw material and bulk storage. Packaging at the facility includes some products not manufactured at the site.
    Routine specification testing is conducted at the site (e.g, aqueous, non-aqueous, redox, complexometic and Karl Fisher titrations, melting point, UV-Vis analysis, infrared, elemental analysis, optical rotation, ICP, GC, NMR, HPLC and endotoxin).
  • Primary applications of products manufactured:
    Buffers for laboratory use, pharmaceutical raw materials or other industry applications.
  • No production or packaging of antibiotics, toxins, steroids or hormone products at the buffers facility.
  • Each in-process and batch lot is tested to conform to internal and customer specifications. Lot specific certificates of analysis are available for each lot.
  • Processes are monitored to ensure conformance to specifications and procedures. Quality Assurance is responsible for raw material approval, manufacturing batch and packaging record review, and product release of buffers with EQP (Enhanced Quality Program) classification of Elite. Quality Control is responsible for product release of buffers of Standard EQP. QA reporting is independent of Manufacturing. Organizational charts are available.
  • Sub-contractors are qualified as part of our internal service supplier qualification program and used in some instances of preventive maintenance, calibration and pest control, and may be contracted for special validation or testing requests if not met by our in-house capabilities.
2. Compliance Evidence
2.1. ISO registration number and certificates
  • Quality Systems governing the SAFC Biological Buffers manufacturing operations are based on selected elements of ICH Q7A as applicable to buffers for manufacturing use. Customer audits by pharma companies have concurred that this ensures adequate control for producing biological buffers.
  • Support activities (testing, packaging, warehouse, maintenance, distribution, purchasing and customer support): ISO 9001:2008, Registration No QSR-825. Can be downloaded at http://safcglobal.com/safcglobal/en-us/home/quality-management-systems/iso-certificates.html
2.2. GMP Inspections by competent authorities (Regulatory Agencies):
  • Not applicable to the Biological Buffers facility. FDA Registration Number 1937990 applies to other SAFC manufacturing facilities within the Cherokee site.
2.3. General GMP and compliance statements
  • GMP not applicable to the Biological Buffers facility portion of this site.
  • Quality Systems governing the SAFC Biological Buffers manufacturing operations are based on selected elements of ICH Q7A Good Manufacturing Practice Guidance for Active Pharmaceutical Ingredients with support activities under ISO 9001:2008.
2.4. Other certifications or external audit programs:
  • MSD - Metropolitan St. Louis Sewer District – quarterly routine compliance inspections
  • City of St. Louis - Air Pollution Control Division – annual random compliance inspections
  • MDNR (Missouri Dept of Natural Resources) – EPA compliance
3. Quality Management Systems Compliance Details
IPEC-PQG GMP compliance not applicable for the biological buffers.
3.1. Quality Management Systems
  • General
    - We have established a comprehensive Management System covering all aspects of its business and has a Corporate Quality Policy.
    - A Quality Manual for biological buffers describes the policies and systems to ensure that the quality of our products meet customer expectations.
    - Internal and external audits, Process Improvement programs and nonconformances from the CAPA system are reported to Executive Management.
    - Responsibilities of Quality Assurance functions are clearly defined.
    - Customer specifications can be managed on a custom basis.
  • Documentation
    - The system and rules regarding document control and management are detailed in the management system description.
    - Administration and control of documents is QA responsibility.
    - All activities are defined in Processes, Standard Operating Procedures (SOP), Operating Procedures (OP), Manufacturing Procedures and auxiliary documents.
    - All Documents have a unique identifier, revision number, effectiveness date, next review date and distribution date.
    - All activities connected to the manufacturing and testing of a batch (batch record), and the equipment (logbooks), are stored with protected access and archived for at least ten years after production
  • Change Control
    - A change control system covering all quality relevant changes is in place.
    - Quality and Manufacturing evaluate change requirements prior to manufacture and/or release to customers. Quality is responsible for change approval and customer notification. Change control requirements of the customer are evaluated. Customer notification on changes is possible provided that a specific agreement is set up between us and customer.
3.2. Management Responsibility
  • We are committed to fulfill all customer and regulatory requirements, and to align site
    goals with the quality policy.
  • Adequacy of the quality system is assessed by Key Performance Indicators, which include, e.g., audits and review of metrics on CAPAs, customer complaints, failed batches & recalls. Executive Management review occurs yearly.
  • Management is responsive to customer perceptions as relayed back from customer interactions and supplier scorecards.
3.3. Resource Management
  • Management assigns resources and responsibility where needed to ensure that the quality management system operates efficiently.
  • Personnel are qualified and trained on procedures and processes relevant to their function.
  • Equipment is subject to written preventive maintenance procedures and qualification programs, if required.
3.4. Product Realization
  • For EQP Elite, raw materials and their suppliers are qualified.
  • For EQP Elite, process critical control points have been established by FMEA, with in-process testing as needed for verification.
  • Process Equipment Qualification and commissioning is undertaken on all new equipment installations.
    Retrospective Installation Qualification and Operations Qualification were performed on original equipment.
  • Equipment is designed for cleanability and to provide non-reactive product contact surfaces. Any multiuse equipment is cleaned per verified procedure and cleaning results are reviewed by QA.
  • Equipment used in manufacturing and testing are under a preventive maintenance and calibration program.
  • For EQP Elite, the Master Packaging Formulation (MPF) details the labeling and packaging procedures, components and accountability.
  • No animal-sourced materials, latex or GMO materials are used in the buffers facility.
  • All measuring and control devices are controlled and calibrated on a regular basis.
  • Quality Control Testing
    - Test methods are not validated (except compendial). Depending on customer requirements, method development activities up to and including method verification may be undertaken.
    - For EQP Elite, shelf life is established as a recommended retest date on the Certificate of Analysis.
    - Retained samples are kept for 1 year after release. For EQP Elite, samples are retained for a minimum of 2 years.
  • Planning: Make to stock based upon First In, First Out (FIFO) or make-to-order.
  • Purchasing: Global Supply Chain capabilities with vendor management per quality, delivery, pricing, and compliance to regulatory requirements.
  • Service: Pre- and post-sales technical and compliance services. Global distribution capabilities.
3.5. Measurement, Analysis and Improvement
  • Key Performance Indicators are designed and implemented for all relevant processes. Regular management reviews are performed to assess continuing compliance of the system with customer and regulatory requirements.
  • Nonconforming product controls are electronic; the SAP MRP system prevents product in a quality rejected status from being shipped.
  • Corrective and Preventive processes: Data relevant for product quality are trend analyzed to detect potential drifts and corrective/ preventive actions are implemented as appropriate.
  • Continuous improvement represents an essential part of our strategy. All employees are highly motivated to participate in various local and corporate improvement programs. These programs focus on quality, safety, 5S programs and economic improvements in all areas of our business and operations.
4. Miscellaneous Site Information
  • Social Responsibility - We are committed to serving the communities where we live and work, to a safe and fair work environment where our employees can grow and enrich themselves in the diversity that this worldwide organization offers, and to sustainability programs.
    - Our foundation began in 2004 and has contributed large and small gifts to over 250 worthwhile charitable organizations. See /customer-service/about-us.html for Global Citizenship and Corporate Giving.
  • A batch is a specified quantity of material that is intended to have uniform character and quality, within specified limits, and is produced according to a single manufacture order during the same cycle of manufacture. Each batch is uniquely identified with one lot number. For products manufactured since late
    2011, the lot format is SL followed by 2 more letters, 4 numbers, and an optional “V”.
  • Environmental controls and monitoring: Cold weather protection (heat supplied) and make-up air is coarsefiltered to remove large particulates. The process equipment remains closed throughout the process so that environmental factors do not affect final product quality. Water used in manufacture is deionized and port is monitored to ensure conformance to established specifications.
  • Buffers with similar characteristics are manufactured in shared equipment. Cleaning is verified per established procedures to prevent cross-contamination.
  • Enhanced Quality Program (EQP)
    - EQP is an internal four-level classification system for our products: STANDARD, PREMIUM, ELITE and GMP. These tiers are set up to reflect the various levels of product quality and support documentation.
    The Cherokee site Biological Buffers facility participates in Standard- and Elite-tier products.
  • The site has a business continuity plan for disaster recovery in order to mitigate any potential supply interruptions to our customers.
Biological Buffers Site Supply Chain Security Overview
1. Scope
  • 3300 S. Second St., St. Louis, MO 63118 USA, internally referred to as the Cherokee site.
2. Supply Chain Security
The following controls are executed to ensure the integrity and security of the product in transit from manufacturer to end user:
  • Products are packed in tamper-evident packaging. All packing/labeling operations are performed on-site or
    Switzerland (the latter for routine Eurasian distribution).
  • Environmental controls can be implemented during transport (e.g., to control cold chain).
  • Intermediate storage locations are partnered warehouses that utilize approved procedures provided by us. They are monitored and reviewed on a routine bases. Full inventory audits occur at least once a year.
3. Security Information
3.1 Scope of Security Plan
  • The Environmental Health and Safety department is responsible for site security.
  • All relevant procedures and policies are part of our management system. The overall system is internally audited at least once every three years.
  • All employees receive regular training regarding security and safety.
  • Procedures for data and computer system protection are in place and part of the management system.
  • Site access control
    - Facility is security-fenced and has closed-circuit TV surveillance at a central security station.
    - Ingress/egress of employees is controlled through manned security checkpoints or electronic badge system.
    - Ingress/egress of suppliers/contractors is monitored via a central gate, which is manned with an external security service.
    - Sensitive areas within the facility are additionally locked with mechanical or electronic locking systems.
3.2 Personnel Security
  • Pre-employment background checks are performed on all employees.
  • Background checks are performed on temporary and contract personnel.
  • Each employee receives initial and ongoing training regarding safety, health and environment.
  • Individual employee badges are collected upon permanent leave of the company to avoid unapproved access to the site. Contractors receive temporary badges which are returned daily.
4. Environmental Health & Safety Program (EHS)
  • Develops programs and procedures to implement EHS regulatory and principle requirements.
  • Develops and delivers EHS training.
  • Monitors and reports site EHS performance.
  • Investigates significant EHS incidents.
  • Provides continuous improvement in EHS Performance through periodic and organized Management Systems Reviews.
  • Provides an annual review of the facility management systems for assuring compliance with applicable EHS laws, regulations and Corporate EHS Principles and Policies.
  • There are no registrations to ISO 14001, OHSAS 18001 or others.
  • A documented emergency response plan consists of clear responsibilities and defined processes in case of events
    related to EHS.
5. Miscellaneous site information
  • The Business Continuity Plan utilizes our vast raw material/chemical supply chain and potential transfers of projects to our other facilities in the US and Europe in order to mitigate potential supply interruptions.
  • Medical, fire and HAZMAT teams are on-site. Procedures exist for emergency shut-downs, utilities and information systems. An Emergency Operation Center coordinates internal mitigation and recovery activities, and liaisons with external emergency agencies.
  • Our initiatives promote greater environmental responsibility and support energy
    conservation, natural resources conservation, material reuse, reduction and recycling, and the application of green
    chemistry into our processing operations worldwide.