[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[3D Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-technique/3d-cell-culture)HyStem™ Hyaluronic Acid Based Hydrogels for 3D Cell Culture Applications
# HyStem™ Hyaluronic Acid Based Hydrogels for 3D Cell Culture Applications
[Introduction](https://www.sigmaaldrich.com#introduction)
[HyStem™ Features and Benefits](https://www.sigmaaldrich.com#features)
[Publications using HyStem™ with Applications](https://www.sigmaaldrich.com#publications)
[Examples of Cells That Have Been Cultivated Successfully with HyStem™ Platform](https://www.sigmaaldrich.com#examples)
[](https://www.sigmaaldrich.com)
## Introduction
HyStem™ is a semi-synthetic 3D hydrogel platform that allows cell growth in a 3D environment. Their main component is one of the major constituents of the [extracellular matrix](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/extracellular-matrix) (ECM), hyaluronic acid (HA). HA is the simplest glycosaminoglycan (a negatively charged, linear, non-sulfated polysaccharide) that provides compression strength, lubrication, and hydration within the ECM1. It also regulates cell adhesion and motility2,3 and mediates cell proliferation and differentiation4, making it not only a structural component of tissues but also an active signaling molecule. Hystem™ hydrogels have been extensively published for numerous applications including stem cell, cancer and neuroscience research.
Three different HyStem™ platform options are available:
- HyStem™ ([GS311F](https://www.sigmaaldrich.com/US/en/product/sigma/gs311f)) is composed of thiol modified HA (__Glycosil__®) and thiol reactive crosslinker (__Extralink__®); the crosslinker allows for the gelification process (without it, HA solution will stay liquid)
- HyStem™ C ([GS313F](https://www.sigmaaldrich.com/US/en/product/sigma/gs313f)) includes __Glycosil__® and __Extralink__® but also thiol modified denatured collagen fibris called
__Gelin-S__® to accommodate some cell attachment needs (e.g. Stem cells)
- HyStem™ HP ([GS315F](https://www.sigmaaldrich.com/US/en/product/sigma/gs315f)) includes all of HyStem™ C plus heparin sulfate to ensure growth factor release at the immediate proximity of cells
__Figure 1. Biochemical structure of Hystem™ hyaluronic acid based 3D hydrogels.__HyStem™ HA hydrogels closely mimic the native extracellular matrix protein environment giving customers options of HA, HA+ collagen or HA+ collagen+ heparin sulfate linked hydrogels for 3D cell culture support and growth. The hydrogels are customizable and allow attachment of other growth factors and ECM proteins.
## [](https://www.sigmaaldrich.com)HyStem™ Features and Benefits
- __Biologically accurate:__ HyStem™ kits are optimal for culturing stem cells whose natural environments are rich in hyaluronic acid. The HyStem™ hydrogel scaffold closely mimics the rich, natural extracellular matrix environment with hyaluronic acid and collagen fibrils.
- __Customizable:__ The HyStem™ platform allows growth factor incorporation, attachment factor incorporation and ECM protein incorporation so you can modify the rigidity of the hydrogel, and do cell encapsulation vs. top plating.
- __Semi-synthetic:__ Because the HyStem™ platform is a synthesized matrix and not a biological extract, the composition of their cells’ environment is more closely controlled. HyStem™ components include chemically synthesized HyStem™ (thiolated hyaluronic acid) and Extralink® (thiol-reactive crosslinker).
### Selecting the Right HyStem™ Kit for your Application
HyStem™HyStem™ CHyStem™ HP
__Recommended applications__For researchers who require an animal component free system.
For researchers who will customize with their own attachment factors and/or ECM proteins/peptides
For researchers who require a minimal number of cell attachment sitesFor researchers who require a large number of generalized cell attachment sites for their stem cell culture(s)For researchers planning to incorporate and gradually release growth factors into the stem cell environment
__Customization__
- Amount and type of ECM proteins
- Amount and type of cell attachment factors
- Amount and type of growth factors
- Hydrogel rigidity
- Choice of cell encapsulation or top plating
- Amount and type of ECM proteins
- Amount and type of growth factors
- Hydrogel rigidity
- Choice of cell encapsulation or top plating
- Amount and type of ECM proteins
- Amount and type of growth factors
- Hydrogel rigidity
- Choice of cell encapsulation or top plating
__Ordering Information__[HYS020](https://www.sigmaaldrich.com/US/en/product/sigma/gs311f) - HyStem™ Cell Culture Scaffold Kit (for 7.5 mL of hydrogel scaffold solution)
[HYS010](https://www.sigmaaldrich.com/US/en/product/sigma/HYS010) - HyStem™ Cell Culture Scaffold Trial Kit (for 2.5 mL of hydrogel scaffold solution)[HYSC020](https://www.sigmaaldrich.com/US/en/product/sigma/gs313f) - HyStem™-C Cell Culture Scaffold Kit (for 7.5 mL of hydrogel scaffold solution)
[HYSC010](https://www.sigmaaldrich.com/US/en/product/sigma/HYSC010) - HyStem™-C Cell Culture Scaffold Trial Kit (for 2.5 mL of hydrogel scaffold solution)[HYSHP020](https://www.sigmaaldrich.com/US/en/product/sigma/gs315f) - HyStem™-HP Cell Culture Scaffold Kit (for 7.5 mL of hydrogel scaffold solution)
[HYSHP010](https://www.sigmaaldrich.com/US/en/product/sigma/HYSHP010) - HyStem™-HP Cell Culture Scaffold Trial Kit (for 2.5 mL of hydrogel scaffold solution)
__Animal Component Free__YesNoNo
__Cell Types tested__
- Human embryonic stem cells (H9s)
- Umbilical cord blood CD23+ stem cells
- Hepatic stem cells
- Hepatic progenitor cells
- Human embryonic stem cells
- Human mesenchymal stem cells
- Neural progenitor cells
- Hepatic progenitor cells
__Appropriate conditions__Recommended for *in vivo* and *in vitro* experimentationRecommended for *in vitro* experimentation onlyRecommended for *in vivo* and *in vitro* experimentation
__Cell Encapsulation__Accomodates cell encapsulation (true 3D) as well as plating on top of the hydrogel for pseudo-3D growth
__Cell Attachment__Offers a minimal number of cell attachment sites; if desired, researchers may incorporate attachment factors of their choice.Enables attachment of a wide variety of cell types through Gelin-S (thiolated gelatin)Enables attachment of a wide variety of cell types through Gelin-S (thiolated gelatin)
__Growth Factor Incorporation__Researchers will customize with their own growth factors; growth factors will likely require replenishmentResearchers will customize with their own growth factors; growth factors will likely require replenishmentEnables ionic binding and slow release of growth factors through thiolated heparin; frequent replenishment of growth factors is not required.
__Gelation Time__<20 minutes20 minutes23 minutes
Each Kit is sufficient to produce 7.5 mL of Hydrogel Scaffold Each Trial Kit is sufficient to produce 2.5 mL of Hydrogel Scaffold
Table 1.HyStem™ platform selection guide.
## [](https://www.sigmaaldrich.com)Publications using HyStem™ with Applications
[A hyaluronan hydrogel scaffold-based xeno-free culture system for *ex vivo* expansion of human corneal epithelial stem cells](https://www.sigmaaldrich.com/US/en/tech-docs/paper/28211875). Chen D, Qu Y, Hua X, Zhang L, Liu Z, Pflugfelder S C and Li DQ. Eye (Lond). 2017 Jun;31(6):962-971. Demonstrates that HyStem™ C can successfully support the *ex vivo* cultivation of human corneal epithelial stem cells (CESCs). It has been shown several times that HyStem™ C is a very suitable carrier for *ex vivo* experimentation with numerous types of stem cells.
[Mesenchymal stem cells support growth and organization of host-liver colorectal-tumor organoids and possibly resistance to chemotherapy](https://www.sigmaaldrich.com/US/en/tech-docs/paper/28589925). Devarasetty M, Wang E, Soker S, Skardal A. Biofabrication. 2017 Jun 7;9(2):021002. This paper used HyStem™ HP in a study of host-liver colorectal-tumor spheroids composed of primary human hepatocytes, mesenchymal stem cells (MSC) and colon carcinoma HCT116 cells were created in simulated microgravity rotating wall vessel (RWV) bioreactors. The cells were seeded on HyStem™ microcarriers, loaded with liver-specific growth factors and ECM components.
[3D microvascular model recapitulates the diffuse large B-cell lymphoma tumor microenvironment *in vitro*](https://www.sigmaaldrich.com/US/en/tech-docs/paper/28054086)*.* Mannino RG, Santiago-Miranda AN, Pradhan P, Qiu Y, Mejias JC, Neelapu SS, Roy K, Lam WA. Lab Chip. 2017 Jan 31;17(3):407-414. UsedHyStem™ C to develop a lymphoma-on-chip model consisting of a hydrogel based tumor model traversed by a vascularized, perfusable, round microchannel that successfully recapitulates key complexities and interactions of the *in vivo* tumor microenvironment *in vitro.*
[Cell-cell interaction between vocal fold fibroblasts and bone marrow mesenchymal stromal cells in three-dimensional hyaluronan hydrogel](https://www.sigmaaldrich.com/US/en/tech-docs/paper/23653427). Chen X, Thibeault SL. J Tissue Eng Regen Med. 2016 May;10(5):437-46. This paper used HyStem™ C for coculture of vocal fold fibroblasts (VVF) and bone-marrow deriver mesenchymal stromal cells (BM-MSCs).
[Multilayered Hyaluronic Acid-Based Hydrogel Formulations Suitable for Automated 3D High Throughput Drug Screening of Cancer-Stromal Cell Cocultures](https://www.sigmaaldrich.com/US/en/tech-docs/paper/26059746). Engel BJ, Constantinou PE, Sablatura LK, Doty NJ, Carson DD, Farach-Carson MC, Harrington DA, Zarembinski TI. Adv Healthc Mater. 2015 Aug 5;4(11):1664-74. This paper used HyStem™ C to develop HTS suitable for 3D co-culture systems, mixing cancer and stromal cells.
### Examples of Cells That Have Been Cultivated Successfully with HyStem™ Platform
__Example 1.__ H9 human embryonic stem cells plated on HyStem™ hydrogels.
__Example 2__. Human mesenchymal stem cells grown (5 days) on the surface of a HyStem™ hydrogel with collagen I noncovalently incorporated.
__Example 3__. Endothelial progenitor cells cultivated in HyStem™-HP. Blue = mouse cell nuclei, brown = human cell nuclei, red = CD31 protein (courtesy of Robert Grove, Endgenitor Technologies, Inc., Indianapolis, IN)__Example 4__. Neurosphere-derived human embryonic stem cells (H9) seeded in HyStem™-C and grown for 5 days. Red = Beta III Tubulin. Blue = Draq-5.
Figure 2.Example of cells that have been successfully cultured with a HyStem™ platform.
## Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
1999\. Book review. Biochemical Education. 27(4):241. [https://doi.org/10.1016/s0307-4412(99)00089-8](https://doi.org/10.1016/s0307-4412%2899%2900089-8)
2\.
Dowthwaite GP, Edwards JCW, Pitsillides AA. 1998. An Essential Role for the Interaction Between Hyaluronan and Hyaluronan Binding Proteins During Joint Development. J Histochem Cytochem.. 46(5):641-651. [https://doi.org/10.1177/002215549804600509](https://doi.org/10.1177/002215549804600509)
3\.
Cheung W, Cruz TF, Turley EA. 1999. Receptor for hyaluronan-mediated motility (RHAMM), a hyaladherin that regulates cell responses to growth factors. 27(2):135-141. [https://doi.org/10.1042/bst0270135](https://doi.org/10.1042/bst0270135)
4\.
Entwistle J, Hall C, Turley E. 1996. Hyaluronan receptors: regulators of signalling to the cytoskeleton.. J Cell Biochem. 61569–77.
__Related Articles__
- [Attachment Factors for Three-dimensional Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3-dimensional-cell)
- [Bioink Selection for 3D Bioprinting](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-bioprinting-bioinks)
- [Advanced Cell Culture Technology for Generation of In Vivo-like Tissue Models](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-cell-culture-models)
- [3D Cell Culture Tools and Techniques](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-cell-culture-technology)
- [Hydrogels for 3D Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-hydrogels)
- [3D Imaging of Optically Cleared Spheroids in Corning® Spheroid Microplates](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-imaging-optically-cleared-spheroids-corning-microplates)
- [3D Organoid Culture: New In Vitro Models of Development and Disease](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-organoid-culture)
- [Cancer Stem Cell Proliferation in Human Prostate and Breast Cancer Cell Lines Utilizing a New Defined Serum-Free 3D Spheroid Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/3d-spheroid-cancer-stem-cell-media)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Hydrogels](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/3d-cell-culture/hydrogels)
- [3D Cell Culture Techniques](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/3d-cell-culture)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)