Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8:00 PM CDT Friday, June 21st until 9:00 AM CDT Saturday morning June 22nd.
We apologize for any inconvenience this may cause.
Immobilon® NOW Transfer Membrane rolls for Western blotting
Immobilon® NOW rolls offer the convenience of pre-cut sheets with the flexibility of rolls in the formats you depend on: Immobilon®-E, Immobilon®-P, Immobilon®-FL and Immobilon®-PSQ membranes.
Features
  • Single-cut convenience
  • Compatibility with mini or midi gels
  • No waste
  • More compact package saves space in your lab
Four Immobilon® PVDF membranes, optimized for your application
All Immobilon® PVDF membranes offer exceptional durability and high protein binding, for strong signal with low background.
Immobilon®-E Membrane
  • The only PVDF membrane that wets out in transfer buffer—no alcohol required
  • Ideal for most Western blot experiments where the target protein is >20 kDa
  • Use with chemiluminescent or colorimetric detection
Immobilon®-P Membrane
  • The most widely cited PVDF for immunoblotting
  • For Western blot and dot blot experiments where the target protein is >20 kDa
  • Recommended for chemiluminescent and colorimetric detection
Immobilon®-FL Membrane
  • Optimized for fluorescence detection including near-infrared wavelengths
  • Lowest background fluorescence improves sensitivity of all fluorescence detection protocols
Immobilon®-PSQ Membrane
  • Highest protein binding and retention
  • Best choice for low MW targets and low abundance targets
  • Low background fluorescence
  • Compatible with chemiluminescent and colorimetric detection
Immobilon® NOW Dispenser
The optional Immobilon® NOW Dispenser adds even more convenience by offering a complete solution to measure, cut and store the transfer membrane. Simply retract the unused membrane into the container, and always see when it’s time to refill.
Dispenser Schematic
Ordering Information
Description Catalog Number
Immobilon®-E transfer membrane, 8.5 cm x 10 m roll IEVH85R
Immobilon®-P transfer membrane, 8.5 cm x 10 m roll IPVH85R
Immobilon®-FL transfer membrane, 8.5 cm x 10 m roll IPFL85R
Immobilon®-PSQ transfer membrane, 8.5 cm x 10 m roll ISEQ85R
Immobilon® NOW Dispenser IMDISP
Immobilon® NOW Bundle: Immobilon®-P transfer membrane (two 8.5 cm x 10 m rolls) with Immobilon® NOW dispenser NWKITIP
Immobilon® NOW Bundle: Immobilon®-E transfer membrane (two 8.5 cm x 10 m rolls) with Immobilon® NOW dispenser NWKITIE
Immobilon® NOW Bundle: Immobilon®-FL transfer membrane (two 8.5 cm x 10 m rolls) with Immobilon® NOW dispenser NWKITFL
Immobilon® NOW Bundle: Immobilon®-PSQ transfer membrane (two 8.5 cm x 10 m rolls) with Immobilon® NOW dispenser NWKITSQ