Immobilon®-E PVDF membrane for Western blotting
Immobilon®-E provides the first PVDF blotting membrane that wets in transfer buffer or water. It removes the alcohol pre-wet step while maintaining the high performance associated with Immobilon® transfer membranes.
Click here to request a sample of Immobilon®-E PVDF membrane!
  • 0.45 µm pore size ideal for most Western blotting applications with chemiluminescence or chromogenic detection
  • Save time and reduce waste by eliminating the alcohol pre-wet step
  • Higher protein retention delivers stronger signal than most nitrocellulose membranes
Watch the following video to learn how the Immobilon®-E membrane provides the benefits of PVDF without an alcohol pre-wet step.
Materials