Immobilon®-FL PVDF membrane for Western blotting
The Immobilon®-FL PVDF transfer membrane (0.45 μm) is the first transfer membrane specifically optimized for fluorescence-based detection of Western blots. Its extremely low background autofluorescence improves sensitivity of all fluorescence detection protocols.
Key Benefits
  • The best membrane for near-infrared wavelengths (700-800 nm)
  • Strong signals due to higher protein adsorption and retention on the membrane
  • Low background to detect even faint bands
  • High tensile strength for multiple stripping and reprobing cycles