Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Accelerated Osteocyte Differentiation of Human Mesenchymal Stem Cells in a Novel Xeno-free and Serum-free Osteogenesis Differentiation Media
By: Anna Abai, Nick Asbrock and Vi Chu1
1 MilliporeSigma, Bioscience Division, Temecula, CA, USA
,  
Introduction
Methods
Results
Conclusions
Introduction
Mesenchymal stem cells (MSCs) have the capacity for multi-lineage differentiation, giving rise to a variety of mesenchymal phenotypes such as osteoblasts (bone), adipocytes (fat), and chondrocytes (cartilage). MSC-derived osteoblasts may also have applications in fracture repairs and generation of stem cell-derived bone grafts, and spinal fusion. The generation of osteoblasts from hMSCs has enormous value in developing and evaluating drugs for alleviation of orthopedic conditions. Similarly, the ability to generate MSCs from iPSCs can unwrap the possibility of generating disease-specific MSCs, which may be useful in monitoring orthopedic disease progression and examining therapeutic efficacies of drugs.
Difficulties in directing MSCs and other stem cells to generate fully functional, specific cell types prevent the full exploitation of the enormous potential of these multipotent and pluripotent cells. Generating specific cell types in large scale, in a reproducible and cost effective manner, is even more challenging. One of the principal factors to consider here is the need for specific cell culture medium where variables and animal derived components are eliminated. This is particularly significant for osteoblast production since osteogenic differentiation protocols invariably include serum that contains essential components for generation of these cells.
To overcome these difficulties, in collaboration with Plasticell Ltd., we have developed OsteoMAX-XF™ differentiation media: a novel serum-free and xeno-free differentiation medium for the generation of osteoblasts from human MSCs with accelerated differentiation rates and frequencies. By screening roughly 3,500 combinations of serum and xeno-free media, we discovered a remarkably efficient, fully defined, reproducible medium to promote the differentiation of hMSCs into osteoblasts, offering an excellent, cost effective method to produce large amounts of human bone cells for multiple applications.
Back to top
Methods
The discovery of OsteoMAX-XF™ differentiation medium was made possible by the use of Plasticell’s Combicult® high-throughput combinatorial platform, which combines miniaturization of cell culture on microcarrier beads, a pooling/splitting protocol and a unique tagging system to allow multiplexing of thousands of experiments in one screen (Figure 1). In this system, stem cells grown on microcarrier beads are shuffled randomly, stepwise through multiple differentiation media using a split-pool method. The iterative process of systematically splitting, culturing and pooling samples all possible combinations of media in a pre-determined matrix. Each medium is spiked with a unique fluorescent tag that attaches to the beads. At the end of the differentiation process, beads bearing differentiated cells are identified by a screening assay, such as immunostaining or reporter gene expression. Individual positive beads are isolated using an automated large-particle sorter. The cell culture history of each positive bead is then deduced by analysis of the fluorescent tags attached to the bead. Typically 100 or more positive differentiation protocols are discovered in each screen. These are then analyzed using bespoke Ariadne® bioinformatics software, which uses criteria such as hierarchical clustering and probability analysis to select the optimal protocols for further validation.
Combicult® screen design
Figure 1. Combicult® screen design. The iterative process of systematically splitting, culturing and pooling samples all possible combinations of media in a pre-determined matrix.
We screened 15 different serum-free media on each of the 3 stages of differentiation, thereby testing 3,375 unique differentiation protocols. At the end of each experiment, beads were analyzed for cells expressing osteocalcin, a marker of osteoblasts. We found 97 unique protocols that were analyzed using bioinformatics software, which helped us identify those predicted to be the most efficient and optimal. Of these, 12 were selected for validation studies and were tested on microcarrier beads, as used in the screening experiment, in a monolayer culture system. From these validation experiments the most effective protocol, B372, was identified. In particular, this protocol gave very extensive mineralization of cultures compared to other selected protocols.
The next step was to optimize protocol B372 and the formulation for laboratory and larger-scale use. It was found that mixing together the media components from the 3 stages of differentiation into one formulation provided more efficient differentiation than the serial protocol. This ‘one-application’ formulation was termed B372+ (now known commercially as OsteoMAX-XF™, SCM121) and was tested across several sources of MSCs and compared to other commercially available osteogenic differentiation kits.
Back to top
Results
Figure 2. Validation and refinement of serum-free differentiation protocols determined by CombiCult® technology resulted in identification of protocols that drive MSC differentiation to mineralizing osteocytes. The novel protocols are more effective than commercially available kits and give consistent results across multiple cell lines. Alizarin red staining of MSC cultures differentiated for 28 days; cell lines 1-3: bone marrow-derived MSC; cell line 4: adipose-derived MSC.
Differentiation of multiple MSC lines at day 7, 14, and 21
Mineralization kinetics - control, day 6, 10, and 14
Figure 3. Differentiation of multiple MSC cell lines using (A) (i) OsteoMAX-XF™ Differentiation Medium; (ii) Supplier LT Osteogenic Differentiation Kit. Differentiation was induced over 21 days in 48-well plate cultures of four different human MSC cell lines. Alizarin Red staining of representative wells at day 7, 14 and 21 are shown. B) Mineralization kinetics of human bone-marrow derived MSC (SCC034) differentiated in OsteoMAX-XF™ Medium.
Back to top
Large scale osteocyte differentiation of bone-marrow MSC cell lines after 10 days
Figure 4. Large scale osteocyte differentiation of bone-marrow MSC cell lines using OsteoMAX-XF™ Differentiation Medium stained with Alizarin Red mineral staining (TMS-008) after 10 days of differentiation. Approximately 107 mineralizing osteoblasts could be obtained from a 600 cm2 cell culture flask or 500 cm2 roller bottle.
Quantitative alkaline phosphatase determination of OsteoMAX-XF™ stability
Figure 5. Quantitative alkaline phosphatase determination of OsteoMAX-XF™ stability. OsteoMAX™ medium was subjected to stressed conditions ranging from 24 hours at RT to 7 days at 37 °C. Cells were exposed to varying concentrations of the stressed supplements for 7 days before the conditioned medium was collected for quantitative determination of AP activity. EMD Millipore’s Quantitative Alkaline Phosphatase ES Characterization Kit (SCR066) was used. Stressed conditions were: #1: One freeze-thaw cycle (control); #2: Two freeze-thaw cycles; #3: Two freeze-thaw cycles, 24 hours at RT; #4: Two freeze- thaw cycles, 2 days at RT; #5: Two freeze-thaw cycles, 3 days at RT; #6: Two freeze-thaw cycles, 4 days at RT; #7: Two freeze-thaw cycles, 6 days at RT; #8: Two freeze-thaw cycles, 8 days at RT; #9: Two freeze-thaw cycles, 7 days at 37°C.
Back to top
Conclusions
We describe the discovery and commercial development of a novel, serum-free and xeno-free MSC osteogenic differentiation medium. The formulation is extremely efficient, generating mineralized cultures in under 7 days, and works consistently across all sources of MSCs tested, including those derived from pluripotent stem cells. This is particularly important for generation of autologous therapies or disease and patient-specific cells for drug screening. Differentiation can be scaled to generate the number of cells required for large-scale applications. OsteoMAX-XF™ medium offers a reliable, simple, cost effective method to generate human osteoblasts for research, clinical and drug discovery applications.