[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Microbial Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-cell-type/microbial-cell-culture)Introduction to Microbial Media # Introduction to Microbial Media - [Introduction](https://www.sigmaaldrich.com#introduction) - [Types of Microbial Media](https://www.sigmaaldrich.com#microbial-media) - [FAQs](https://www.sigmaaldrich.com#faqs) - [Microbial Medial Selection Guide](https://www.sigmaaldrich.com#selection-guide) ## [](https://www.sigmaaldrich.com)Introduction to Microbial Media Microbiological studies require the availability of different species of bacteria, yeast or viruses in the laboratory. Large scale processes such as fermentation, protein and vaccine production require large numbers of bacteria in physiologically active state. A suitable nutrient medium that provides appropriate biochemical environment and maintains all the characteristics of microbes is therefore required for various applications. Any microbial medium should include sources of nutrition, energy, essential minerals, buffering agents and pH indicators. Sometimes they also include selective agents and solidifying agents. Obligate parasites such as bacteriophages may be cultivated in a nutrient medium that also contains host bacteria. The following are the essential components of a typical microbial medium: - Nutrition sources – sources of proteins, vitamins, mineral and carbohydrates; mostly mixture of peptone and meat extract - Energy sources – sources of carbohydrates; mostly glucose - Essential minerals – sources of micro and macro minerals; mostly present in peptone and meat extract - Buffering agents – agents that maintain optimum pH; mostly specific amino acids, phosphates, citrates and zwitterions - pH indicators – agents that indicate variation in pH of the medium by change in color; mostly phenol red - Selective agents – agents that allow the growth of only specific bacteria; antibiotics, tellurites, azides and bile salts are examples - Solidifying agents – agents that maintain the medium in solid or semi-solid state, if required; widely used agent is agar though gelatin is sometimes used - Specific substances such as growth factors, enzymes may be incorporated into the medium for specific bacteria ## [](https://www.sigmaaldrich.com)Types of Microbial Media - __Simple or basal media__: Include nutrient broth and peptone water; used routinely to isolate and culture a variety of bacteria in a molecular biology research laboratory - __Complex media__: Contain mixture of a variety of nutrients; the exact composition of amino acid source is not defined. Examples include chocolate agar, MacConkey agar, Lowenstein-Jensen medium - __Defined media__: Contain known and defined sources of carbon and nitrogen as required by certain bacteria - __Special media__: Contain specific substances that encourage growth of fastidious bacteria, discourage the growth of unwanted bacteria or contain indicators that distinguish different types of bacteria | | | | |-------------------------------------------------|----------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------|-----------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------| | __Name__ | __Introduction__ | __Application__ | | __Bacterial Media__ | | | | LB media | Lysogeny Broth or LB is the most popular and commonly used medium to culture E. coli. It was first created by Bertani G to culture Shigella indicator strain. LB is a rich medium containing peptone, yeast extract, NaCl and agar (for solid medium). Depending on the concentration of NaCl, the LB medium may be classified as follows: LB - Miller, LB - Lennox and LB - low salt. | LB media are suitable for non-selective cultivation of *E. coli* strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. Plates containing sterile LB agar with or without antibiotics are also available for selective culture of antibiotic-resistant strains of E. coli. | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | | Minimal salts (M9) | M9 medium contains essential salts and nitrogen. | Minimal salts (M9) medium is suitable for non-selective cultivation of E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. | | | | | | Terrific Broth - modified | Terrific broth is a nutrient rich medium that results in higher yield of bacteria in less time. | Terrific broth (modified) is suitable for non-selective cultivation of E. coli strains for cloning and high yield production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. | | | | | | Hanahan’s Broth (SOB Medium) | SOB medium has higher peptone, amino acids and peptides than LB medium. | Hanahan’s Broth (SOB medium) enables the preparation of high efficiency competent cells. | | | | | | SOC Medium | SOC medium is a modified SOB medium with added glucose. In the presence of glucose the cellular processes utilize other sugars are repressed. The expression of recombinant proteins that might affect the host cell physiology is inhibited thus resulting in high efficiency transformation reactions. | SOC medium enables the high efficiency transformation of competent cells. | | 2X YT medium | 2X YT medium contains double the concentration of yeast extract compared to typical LB medium. It is useful for the growth and maintenance of high density of cells for a longer growth period. | 2X YT medium is suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. | | | | | | | | | | NZCYM Broth | NZCYM Broth is nutrient rich broth with amino acids, peptides, vitamins and low salt concentration. | NZCYM Broth is suitable for non-selective cultivation of lambda bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. | | | | | | | | | | Yeast Media | | | | Yeast Nitrogen Base with or without amino acids | Yeast Nitrogen Base is nutrient-rich medium with sources of nitrogen, vitamins, trace elements and salts. It acts as the basic nutrient medium for most fermentation reactions involving yeast cultures. | Yeast Nitrogen Base with and without added amino acids for the cultivation of yeast and for classification based on amino acid and carbon requirements, respectively. | | | | | | Phage Media | | | | NZ Amine®Broth | NZ Amine® Broth is nutrient rich nutrient-rich medium with amino acids, peptides, vitamins and low salt concentration. | NZ Amine® Broth is suitable for the growth of lambda bacteriophage. | Table 1Types of microbial media [](https://www.sigmaaldrich.com) ## FAQs __What are the differences among the Luria, Lennox and Miller LB formulations?__ LB, (originally termed lysogeny broth) was initially composed of tryptone, yeast extract, NaCl and glucose.Soon after, the glucose was omitted (Miller's version), and later the NaCl content lowered by half (Lennox's version).For some applications, even lower salt is required (Luria's low salt version). __What is the difference among the LB - Miller products?__ LB - Miller is available in many types to suit your needs.The different product formats include powder and liquid form.The powder form is also available with agar for easy LB-agar plate preparation.L2542 (LB Miller liquid)L3522 (LB Miller powder)L3147 (LB Miller powder with agar) __Which bacterial culture medium is the best choice for my application?__ Each of the broths will likely grow E. coli very well, but there are still general guidelines for choosing a broth when you are working without a protocol.Generally:LB - Miller and LB - Lennox are used for E. coli growth and maintanence, DNA plasmid production and protein production.The Lennox formulation has a lower salt content required for some salt-sensitive selection antibiotics.LB - Luria low salt is used for special applications where the E. coli growth or other constraints require the lowest possible salt content.Terrific Broth is used for higher yield protein production and high yield DNA plasmid production, because of the faster growth of the E. coli in this medium.SOB is used for protein production, DNA plasmid production and the generation of high-efficiency competent cells.SOC is used for initial growth of competent cells and the transformation procedure. __Will adding magnesium to the culture medium increase cell density?__ In microbial broth formulations that do not already contain magnesium, the addition of 10-20 mM MgCl2 or MgSO4 may increase cell densities. You may need to also increase the shaking speed of the incubator. [](https://www.sigmaaldrich.com)[](https://www.sigmaaldrich.com) ## Microbial Media Selection Guide __Name____Components____Product No.__    __Format__  __Preparation Notes____Application__ LB – Miller - 10 g/L Tryptone - 5 g/L yeast extract - 10 g/L NaCl [L2542](https://www.sigmaaldrich.com/US/en/product/sigma/l2542)LiquidReady-to-useSuitable for regular E. coli culture. - 10g/L Tryptone - 10g/L NaCl - 5g/L Yeast Extract [L3522](https://www.sigmaaldrich.com/US/en/product/sigma/l3522)PowderMix 25 g in 1L water; autoclave and cool before use - 15g/L Agar - 10g/L Tryptone - 10g/L NaCl - 5g/L Yeast Extract [L3147](https://www.sigmaaldrich.com/US/en/product/sigma/l3147)Powder with agarMix 40 g in 1L water, heat to dissolve; autoclave and cool before use - 15g/L Agar - 10g/L Tryptone - 10g/L NaCl - 5g/L Yeast Extract - 100μg/mL ampicillin [L5667](https://www.sigmaaldrich.com/US/en/product/sigma/l5667)Agar plates with 100 μg/mL ampicillinReady-to-use - 15g/L Agar - 10g/L Tryptone - 10g/L NaCl - 5g/L Yeast Extract - 50μg/mL ampicillin [L0168](https://www.sigmaaldrich.com/US/en/product/sigma/l0168)Agar Plate with 50 μg/mL ampicillin - 15g/L Agar - 10g/L Tryptone - 10g/L NaCl - 5g/L Yeast Extract - 50μg/mL kanamycin [L0543](https://www.sigmaaldrich.com/US/en/product/sigma/l0543)Agar plates with 50 ug/mL kanamycin - 15 g/L Agar - 10 g/L Tryptone - 10 g/L NaCl - 5 g/L Yeast Extract - 100 μg/mL carbenicillin [L0418](https://www.sigmaaldrich.com/US/en/product/sigma/l0418)Agar plates with 100 μg/mL carbenicillin LB – Lennox - 10 g/L peptone - 5 g/L yeast extract - 10 g/L NaCl [L3022](https://www.sigmaaldrich.com/US/en/product/sigma/l3022)PowderMix 20 g in 1L water; autoclave and cool before useSuitable for culture of E. coli strains that are salt-sensitive. - 15 g/L Agar - 10 g/L Tryptone - 5 g/L Yeast Extract - 5 g/L NaCl [L2897](https://www.sigmaaldrich.com/US/en/product/sigma/l2897)Powder with agarMix 35 g in 1L water, heat to dissolve; autoclave and cool before use - 10 g/L Tryptone - 5 g/L Yeast Extract - 5 g/L NaCl - 2.2 g/L inert binding agents [L7275](https://www.sigmaaldrich.com/US/en/product/sigma/l7275)TabletsAdd each 1.2 g tablet to 48.3 mL water; autoclave and cool before use - 13.72g/L Agar - 9.14g/L Tryptone - 4.57 g/L Yeast Extract - 4.57 g/L NaCl - 1.6g/L inert binding agents [L7025](https://www.sigmaaldrich.com/US/en/product/sigma/l7025)Tablets with agarAdd each 1.68 g tablet to 48.3 mL water; autoclave and cool before use - 10g/L Tryptone - 5 g/L Yeast Extract - 5 g/L NaCl - 0.6 g/L inert binder (EZMix formulation only) [L7658](https://www.sigmaaldrich.com/US/en/product/sigma/l7658)EZMix™Mix 20.6 g in 1L water; autoclave and cool before use - 15g/L Agar - 10g/L Tryptone - 5 g/L Yeast Extract - 5 g/L NaCl [L7533](https://www.sigmaaldrich.com/US/en/product/SIGMA/L7533)EZMix™ with agarMix 35.6 g in 1L water, heat to dissolve; autoclave and cool before use - 15g/L Agar - 10g/L Tryptone - 5 g/L Yeast Extract - 5 g/L NaCl [L5542](https://www.sigmaaldrich.com/US/en/product/sigma/l5542)PlatesReady-to-use - 10 g/L Tryptone - 5.0 g/L Yeast Extract - 5.0 g/L NaCl [L9234](https://www.sigmaaldrich.com/US/en/product/SIGMA/L9234)LiquidReady-to-use LB – low salt - 10 g/L peptone - 5 g/L yeast extract - 0.5 g/L NaCl (and 15 g/L agar for solid medium) [L3397](https://www.sigmaaldrich.com/US/en/product/sigma/l3397)PowderMix 15.5 g in 1L water; autoclave and cool before useSuitable for culture of E. coli strains that require low salt conditions to grow. - 15g/L Agar - 10g/L Tryptone - 5g/L Yeast Extract - 0.5g/L NaCl [L3272](https://www.sigmaaldrich.com/US/en/product/SIGMA/L3272)Powder with agarMix 30.5 g in 1L water, heat to dissolve; autoclave and cool before use Overnight Express Instant LB Medium - Overnight Express Autoinduction System 1 components - Luria Bertani (LB) broth [71757-M](https://www.sigmaaldrich.com/US/en/product/mm/71757m)Granulated mediumJust add the EasyPak contents to 1 L sterile water, supplement with 10 mL glycerol, and microwave for 2 min to dissolve. Overnight Express Instant LB Medium is also available in 1 kg bottles. Granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder.Enables regulated protein expression in E. coli, without monitoring the culture or adding IPTG during cell growth. Overnight Express Instant TB Medium - Overnight Express Autoinduction System 1 components - Terrific Broth (TB) medium [71491](https://www.sigmaaldrich.com/US/en/product/mm/71491)Granulated mediumEasyPak is an aluminum foil pouch containing 60 g granulated medium, sufficient for 1 L culture. Just add the EasyPak contents to 1 L sterile water, supplement with 10 mL glycerol, and microwave for 2 min to dissolve. Overnight Express Instant TB Medium is also available in 1 kg bottles. Granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder. Table 1LB Media __Name____Components____Product No.____Format__  __Preparation Notes____Application__ Minimal salts (M9) - 33.9 g/L Na2HPO4.7H2O - 15 g/L KH2PO4 - 5 g/L NH4Cl and 2.5 g/L NaCl [M6030](https://www.sigmaaldrich.com/US/en/product/sigma/m6030)PowderMix 56.4 g in 1L water to obtain 5X stock solution; autoclave, cool and dilute to 1X before useSuitable for recombinant E. coli strains - 15 g/L KH2PO4 - 64 g/L Na2HPO4 - 2.5 g/L NaCl - 5.0 g/L NH4Cl [M9956](https://www.sigmaaldrich.com/US/en/product/sigma/m9956)LiquidReady-to-use Table 2Minimal Salts (M9) __Name____Components____Product No.____Format____Preparation Notes____Application__ [](https://www.sigmaaldrich.com)Hanahan’s Broth (SOB Medium) - 20 g/L tryptone - 5 g/L yeast extract - 2.4 g/L MgSO4 - 0.5 g/L NaCl - 0.186 g/L KCl [H8032](https://www.sigmaaldrich.com/US/en/product/sigma/h8032)PowderMix 28 g in 1L water; autoclave and cool before useEnables the preparation of high efficiency competent cells. [](https://www.sigmaaldrich.com)SOC Medium - 20 g/L tryptone - 5 g/L yeast extract - 4.8 g/L MgSO4 - 3.603 g/L dextrose - 0.5 g/L NaCl - 0.186 g/L KCl [S1797](https://www.sigmaaldrich.com/US/en/product/sigma/s1797)LiquidReady-to-useSOC medium is a modified SOB medium with added glucose. In the presence of glucose the cellular processes utilize other sugars are repressed. The expression of recombinant proteins that might affect the host cell physiology is inhibited thus resulting in high efficiency transformation reactions. It enables the high efficiency transformation of competent cells. Table 3Minimal Salts (M9) NameComponentsProduct No.FormatPreparation NotesApplication Terrific Broth - modified - 23.6 g/L yeast extract - 11.8 g/L tryptone - 9.4 g/L K2HPO4 - 2.2 g/L KH2PO4 [T5574](https://www.sigmaaldrich.com/US/en/product/sigma/t5574)LiquidReady-to-useSuitable for non-selective cultivation of E. coli strains for cloning and high yield production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. [T9179](https://www.sigmaaldrich.com/US/en/product/sigma/t9179)EZMix™Mix 48.2 g in 1 L water, add 4 mL glycerol; autoclave and cool before use Table 4Terrific Broth - Modified __Name____Components____Product No.____Format____Preparation Notes____Application__ 2X YT medium - 16 g/L tryptone - 10 g/L yeast extract - 5 g/L NaCl [Y1003](https://www.sigmaaldrich.com/US/en/product/sigma/y1003)LiquidReady-to-useSuitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. [Y2377](https://www.sigmaaldrich.com/US/en/product/sigma/y2377)PowderMix 31 g in 1 L water; autoclave and cool before use [Y2627](https://www.sigmaaldrich.com/US/en/product/sigma/y2627)EZMix™Mix 31.6 g in 1 L water; autoclave and cool before use Table 52X YT Medium __Name____Components____Product No.____Format____Preparation Notes____Application__ NZCYM Broth - 10 g/L tryptone - 5 g/L yeast extract - 5 g/L NaCl - 1 g/L Casamino acids - 1 g/L MgSO4 [N3643](https://www.sigmaaldrich.com/US/en/product/sigma/n3643)PowderMix 22 g in 1 L water; autoclave and cool before useSuitable for non-selective cultivation of lambda bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. Table 6NZCYM Broth __Name____Components____Product No.____Format____Preparation Notes____Application__ Yeast Nitrogen Base with amino acids - Nitrogen Sources: - Ammonium sulfate, 5.0 g/L Amino Acids: - L-histidine, 10 mg/L - DL-methionine, 20 mg/L - DL-tryptophan, 20 mg/L Vitamins: - Biotin, 2.0 - Calcium pantothenate, 400 - Folic acid, 2.0 - Inositol, 2.0 mg/L - Nicotinic acid, 400 - p-Aminobenzoic acid, 200 - Pyridoxine HCl, 400 - Riboflavin, 200 - Thiamine HCL, 400 Trace Elements: - Boric acid - Copper sulfate - Potassium iodide - Ferric chloride - Magnesium sulfate - Sodium molybdate - Zinc sulfate Salts: - Potassium phosphate monobasic, 1.0 g/L - Magnesium sulfate, 0.5 g/L - Sodium chloride, 0.1 g/L - Calcium chloride, 0.1 g/L [Y1250](https://www.sigmaaldrich.com/US/en/product/sigma/y1250)PowderMix 6.7 g in 100 mL water, add 5 g glucose to obtain 10X stock solution; filter sterilize and dilute to 1X with sterile water before useYeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains amino acids, nitrogen, vitamins, trace elements and salts. Yeast Nitrogen Base without amino acids - Nitrogen Sources: - Ammonium sulfate, 5.0 g/L Vitamins: - Biotin, 2.0 - Calcium pantothenate, 400 - Folic acid, 2.0 - Inositol, 2.0 mg/L - Nicotinic acid, 400 - p-Aminobenzoic acid, 200 - Pyridoxine HCl, 400 - Riboflavin, 200 - Thiamine HCL, 400 Trace Elements: - Boric acid, 500 - Copper sulfate, 40 - Potassium iodide, 100 - Ferric chloride, 200 - Manganese sulfate, 400 - Sodium molybdate, 200 - Zinc sulfate, 400 Salts: - Potassium phosphate - monobasic, 1.0 g/L - Magnesium sulfate, 0.5 g/L - Sodium chloride, 0.1 g/L - Calcium chloride, 0.1 g/L [Y0626](https://www.sigmaaldrich.com/US/en/product/SIGMA/Y0626)PowderMix 6.8 g in 100 mL water, add 5 g glucose and 5-10 mg of appropriate amino acids to obtain 10X stock solution; filter sterilize and dilute to 1X with sterile water before useYeast Nitrogen Base is considered ideal for culturing wild type yeast. The presence of amino acid is known to prevent selectable marker utilization. Hence, this media is formulated without amino acids and is useful for cloning and manipulation of yeast artificial chromosome. Table 7Yeast Media __Name____Components____Product No.____Format__  __Preparation Notes____Application__ NZ Amine®Broth - 10 g/L tryptone - 5 g/L NaCl - 1 g/L MgSO4 [N3518](https://www.sigmaaldrich.com/US/en/product/sigma/n3518)PowderMix 16 g in 1 L water; autoclave and cool before useSuitable for the growth of lambda bacteriophage. Table 8Phage Media ### References 1\. Sambrook J, Fritsch E, Maniatis T. 1989. Molecular Cloning: A Laboratory Manual. 1. New York: Cold Spring Harbor Laboratory Press. 2\. Parija SC. 2012. Textbook of Microbiology & Immunology. 2. Elsevier India. 3\. Types of Growth Media Used to Culture Bacteria. \[Internet].\[updated 07 Aug 2016; cited 17 Jul 2020]. Available from: [https://www.scienceprofonline.org/microbiology/types-culture-media-for-growing-bacteria.html](https://www.scienceprofonline.org/microbiology/types-culture-media-for-growing-bacteria.html) __Related Articles__ - [Streptococci - Overview of Detection, Identification, Differentiation and Cultivation Techniques](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/streptococci-overview-of-detection) - [Virus Cultivation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/microbial-cell-culture/virus-cultivation) - [Yeast and Molds in Food and Beverages](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/microbial-cell-culture/yeasts) - [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media) - [Media for Staphylococcus Aureus Detection](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/staphylococcus-aureus) - [Identification of Microorganisms Based on Color](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/cytology-and-microscopy/color-identification-microorganisms) - [EN ISO 11133 Microbiological Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/en-iso-11133-water-for-the-preparation-and-performance-testing) - [ISO 11133:2014 Quality Culture Media for Food and Water Testing](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/regulatory-compliance-for-food-and-beverage/iso-11133-culture-media-food-and-water-testing) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) __Related Product Categories__ - [Microbial Culture Media Raw Materials](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media-raw-materials) - [Microbial Culture Media](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media) - [Tryptic soy broth](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media/tryptic-soy-broth) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)