User Guide: MultiScreen® 96-well Transport Receiver Plate
Introduction
The MultiScreen® Transport Receiver Plate is a non-sterile  96-well disposable device designed for transport assays such as drug compound permeability assessment using an artificial membrane. The transport receiver plate is a disposable alternative to the MultiScreen® polytetrafluoroethylene (PTFE) Acceptor Plate (cat. no. MSSACCEPT0R). The transport receiver plate has the following attributes:
  • High drug recovery for micromolar (µM) to millimolar (mM) concentration samples
  • Shake and stir compatibility
  • Low evaporation at room temperature
Automation compatible; conforms to ANSI/SLAS 1-2004 (footprint dimensions), 3-2004 (bottom outside flange dimensions), and 4-2004 (well positions) microplate standards The transport receiver plate is designed to be assembled with a standard 96-well MultiScreen® filter plate, and can be used manually or with many standard automated liquid handlers. For specific recommendations please contact Technical Service.
Materials Required
In addition to the MultiScreen® Transport Receiver Plate, the user must supply the following materials and equipment:
  • MultiScreen® 96-well Filter Plate (e.g., cat. nos. MAIPN4550 or MPC4NTR10)
  • Compound samples in concentrated or pre-prepared dilutions
  • Aqueous transport buffer
  • Artificial membrane solution and solvent
  • UV/Vis spectroscopic microplate reader, HPLC/UV or LC/MS/MS
Optional Equipment
  • MultiScreen® 96-well Deep Well Collection Plate for transport sample prep (cat. no. MDCPN2M50)
  • MultiScreen®HTS 96-well UV-compatible Collection Plate  (cat. no. MSCPNUV40)
  • MultiScreen® Single-well Culture Tray (cat. no. MAMCS0110)
  • Titer plate shaker
  • Magnetic tumble stirrer
  • Tumble stir discs
Usage Guidelines
  • For research use only
  • Single use only
  • Designed for use with MultiScreen® filter plates; not compatible with MultiScreen®HTS or third party filter plates.
  • Careful assembly and disassembly are required to avoid air entrapment or cross-well contamination.
  • Plates are designed to support aqueous transport assays.
  • Solvent systems should be tested prior to running samples.
  • Minimize evaporation during extended incubation by covering filter plate with the lid provided. When stacking assemblies, place lid only on the top filter plate.
  • For certain compounds, mixing, shaking, or stirring may increase permeation, enabling faster sample processing.
  • Disassembly may be required to retrieve the permeant for further analysis by UV/Vis spectroscopy, HPLC/UV or LC/MS/MS.
  • See references 1–4 for more details on running hexadecanepermeability and lipid-PAMPA assays.
Permeability Assay Procedure
If the filter plate has an underdrain, remove it and place the filter plate into a single-well tray before beginning the permeability assay. The single-well tray will protect the bottom of the filter wells during artificial membrane formation.
  1. Prepare transport samples in 96-well deep well plate, if necessary.
  2. Apply lipid or hexadecane solution to center of each filter plate well to form an artificial membrane. For more details, see references 1 and 2.
  3. Add buffer to transport receiver plate wells. The recommended volume is 300 µL per well.
  4. Transfer transport samples to filter plate wells. The recommended volume is 150 µL per well.
  5. Align the cut corners and stack the filter plate on top of the transport receiver plate. Cover filter plate with lid to prevent evaporation during extended incubation periods.
    NOTE:
    Handle plates carefully during steps 5, 6, and 7 to prevent air entrapment or spillover into adjacent wells.
  1. Incubate transport receiver plate assembly at 25 °C for desired length of time.
    NOTE: Incubation times can range from 5 to 16 hours depending on filter, artificial membrane composition, mixing, nature of compounds, and detection method sensitivity.
  2. After incubation, remove the filter plate from the receiver plate to retrieve the permeant.
  3. Evaluate permeant using a standard detection method such as a UV/Vis plate reader, HPLC/UV or LC/MS/MS.
Specifications
Operating Temperature Ambient
Operating Volume  
Minimum 290 µL
Maximum 310 µL
Dimensions  
Length 127.8 mm
Width 85.5 mm
Height 17.4 mm
Material of Construction Polystyrene
Device Storage
Store at room temperature for up to 3 years from date of manufacture. Refer to expiration date on package label.
Notice
The information in this document is subject to change without notice and should not be construed as a commitment by MilliporeSigma or an affiliate. Neither MilliporeSigma nor any of its affiliates assumes responsibility for any errors that may appear in this document.
Materials