Nonsterile Millex® Syringe Filters
Prepare samples using low-extractable Millex® syringe filters to ensure clean baselines and maximize instrument uptime for sensitive analyses, such as dissolution testing, LC-MS, UHPLC, and ion chromatography.
Quality manufacturing, 100% integrity testing. Because we verify membrane integrity for 100% of Millex® filters shipped each day, you can rely on them to remove all particles from your sample.
For the best results, choose a filter based on the following criteria:
  • Membrane that is chemically compatible with your sample
  • Pore size recommended for your analytical method
  • Filter diameter appropriate for your sample volume
  • Choose an integrated prefilter (as in Millex®-HPF filters) for samples with high particulate load
Lots of different samples? Stop by Booths 2318/2319 at Pittcon 2019 and learn how our HPLC-certified hydrophilic PTFE Millex® syringe filters can protect your HPLC results.
Millex® Syringe Filters with Hydrophilic PTFE Membranes
Product No. Millipore
Catalog No.
Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLLGR04NL SLLGR04NL Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 4 0.2 100
SLLHR04NL SLLHR04NL Millex®-LH Syringe Filter Hydrophilic Millipore® PTFE Membrane 4 0.45 100
SLLGX13NL SLLGX13NL Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.2 100
SLLGX13NK SLLGX13NK Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.2 1000
SLCRX13NL SLCRX13NL Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.45 100
SLCRX13TL SLCRX13TL* Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.45 100
SLCRX13NK SLCRX13NK Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.45 1000
SLLG033NS SLLG033NS Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.2 50
SLLG033NB SLLG033NB Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.2 250
SLLG033NK SLLG033NK Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.2 1000
SLCR033NS SLCR033NS Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.45 50
SLCR033NB SLCR033NB Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.45 250
SLCR033NK SLCR033NK Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 33 0.45 1000
*Tube Outlet
Millex® Syringe Filters with Hydrophobic PTFE Membranes
Product No. Millipore
Catalog No.
Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLFGR04NL SLFGR04NL Millex®-FG Syringe Filter Hydrophobic Millipore® PTFE Membrane 4 0.2 100
SLFHR04NL SLFHR04NL Millex®-FH Syringe Filter Hydrophobic Millipore® PTFE Membrane 4 0.45 100
SLFGX13 SLFGX13NK Millex®-FG Syringe Filter Hydrophobic Millipore® PTFE Membrane 13 0.2 1000
SLFGX13 SLFHX13NL Millex®-FH Syringe Filter Hydrophobic Millipore® PTFE Membrane 13 0.45 100
SLFHX13TL SLFHX13TL* Millex®-FH Syringe Filter Hydrophobic Millipore® PTFE Membrane 13 0.45 100
SLFHX13 SLFHX13NK Millex®-FH Syringe Filter Hydrophobic Millipore® PTFE Membrane 13 0.45 1000
Z227528 SLFG025NS Millex®-FG Syringe Filter Hydrophobic Millipore® PTFE Membrane 25 0.2 50
SLFG025 SLFG025NB Millex®-FG Syringe Filter Hydrophobic Millipore® PTFE Membrane 25 0.2 250
SLFG025 SLFG025NK Millex®-FG Syringe Filter Hydrophobic Millipore® PTFE Membrane 25 0.2 1000
SLFG025 SLFH025NS Millex®-FH Syringe Filter Hydrophobic Millipore® PTFE Membrane 25 0.45 50
*Tube Outlet
Millex® Syringe Filters with Hydrophilic Durapore® PVDF Membranes
Product No. Millipore Catalog No. Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLGVR04 SLGVR04NL Millex®-GV Syringe Filter Durapore® PVDF Membrane 4 0.22 100
SLGVR04 SLGVR04NK Millex®-GV Syringe Filter Durapore® PVDF Membrane 4 0.22 1000
Z227552 SLHVR04NL Millex®-HV Syringe Filter Durapore® PVDF Membrane 4 0.45 100
SLHVR04 SLHVR04NK Millex®-HV Syringe Filter Durapore® PVDF Membrane 4 0.45 1000
SLGVX13 SLGVX13NL Millex®-GV Syringe Filter Durapore® PVDF Membrane 13 0.22 100
SLGVX13TL SLGVX13TL* Millex®-GV Syringe Filter Durapore® PVDF Membrane 13 0.22 100
SLGVX13 SLGVX13NK Millex®-GV Syringe Filter Durapore® PVDF Membrane 13 0.22 1000
SLHVX13 SLHVX13NL Millex®-HV Syringe Filter Durapore® PVDF Membrane 13 0.45 100
SLHVX13TL SLHVX13TL* Millex®-HV Syringe Filter Durapore® PVDF Membrane 13 0.45 100
SLSV025NB SLSV025NB Millex®-SV Syringe Filter Durapore® PVDF Membrane 25 5 250
SLGV033N SLGV033NS Millex®-GV Syringe Filter Durapore® PVDF Membrane 33 0.22 50
SLGV033N SLGV033NB Millex®-GV Syringe Filter Durapore® PVDF Membrane 33 0.22 250
SLGV033N SLGV033NK Millex®-GV Syringe Filter Durapore® PVDF Membrane 33 0.22 1000
SLHV033N SLHV033NS Millex®-HV Syringe Filter Durapore® PVDF Membrane 33 0.45 50
SLHV033N SLHV033NB Millex®-HV Syringe Filter Durapore® PVDF Membrane 33 0.45 250
SLHV033N SLHV033NK Millex®-HV Syringe Filter Durapore® PVDF Membrane 33 0.45 1000
*Tube Outlet
Millex® Syringe Filters with PES Millipore Express® Membrane
Product No. Millipore
Catalog No.
Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLGPX13NL SLGPX13NL Millex®-GP Syringe Filter Millipore Express® PES Membrane 13 0.22 100
SLGPX13NK SLGPX13NK Millex®-GP Syringe Filter Millipore Express® PES Membrane 13 0.22 1000
SLHPX13 SLHPX13NL Millex®-GP Syringe Filter Millipore Express® PES Membrane 13 0.45 100
SLHPX13 SLHPX13NK Millex®-GP Syringe Filter Millipore Express® PES Membrane 13 0.45 1000
SLGP033N SLGP033NS Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.22 50
SLGP033N SLGP033NB Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.22 250
SLGP033N SLGP033NK Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.22 1000
SLHP033N SLHP033NS Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.45 50
SLHP033N SLHP033NB Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.45 250
SLHP033N SLHP033NK Millex®-GP Syringe Filter Millipore Express® PES Membrane 33 0.45 1000
Millex® Syringe Filters with Nylon Membranes
Product No. Millipore
Catalog No.
Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLGN033 SLGN033NB Millex®-GN Syringe Filter Nylon 33 0.20 250
SLGN033 SLGN033NK Millex®-GN Syringe Filter Nylon 33 0.20 1000
SLGN033 SLGN033NS Millex®-GN Syringe Filter Nylon 33 0.20 50
SLGNX13 SLGNX13NK Millex®-GN Syringe Filter Nylon 13 0.20 1000
SLGNX13 SLGNX13NL Millex®-GN Syringe Filter Nylon 13 0.20 100
SLGNX13TL SLGNX13TL Millex®-GN Syringe Filter Nylon 13 0.20 100
SLHN033 SLHN033NB Millex®-HN Syringe Filter Nylon 33 0.45 250
SLHN033 SLHN033NK Millex®-HN Syringe Filter Nylon 33 0.45 1000
SLHN033 SLHN033NS Millex®-HN Syringe Filter Nylon 33 0.45 50
SLHNX13 SLHNX13NK Millex®-HN Syringe Filter Nylon 13 0.45 1000
SLHNX13 SLHNX13NL Millex®-HN Syringe Filter Nylon 13 0.45 100
SLHNX13TL SLHNX13TL Millex®-HN Syringe Filter Nylon 13 0.45 100
Millex® Syringe Filters Certified for HPLC
Product No. Millipore
Catalog No.
Description Membrane Diameter (mm) Pore Size (µm) Qty/Pk
SLLGX13 SLLGX13NL Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.2 100
SLLGX13 SLLGX13NK Millex®-LG Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.2 1000
SLCRX13 SLCRX13NL Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.45 100
SLCRX13 SLCRX13NK Millex®-LCR Syringe Filter Hydrophilic Millipore® PTFE Membrane 13 0.45 1000
SLGN033 SLGN033NB Millex®-GN Syringe Filter Nylon 33 0.20 250
SLHNX13 SLHNX13NK Millex®-HN Syringe Filter Nylon 13 0.45 1000
SLHN033 SLHN033NB Millex®-HN Syringe Filter Nylon 33 0.45 250
SLLG033 SLLG033NS Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.20 50
SLLG033 SLLG033NB Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.20 250
SLLG033 SLLG033NK Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.20 1000
SLCR033 SLCR033NS Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.45 50
SLCR033 SLCR033NB Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.45 250
SLCR033 SLCR033NK Millex®-LG
Syringe Filter
Hydrophilic Millipore® PTFE Membrane 33 0.45 1000