PCR Technologies Additional Resources
A Technical Guide to PCR Technologies
Use the table of contents on the right to navigate to other sections of A Technical Guide to PCR Technologies.
Books, Seminars and Tools
Recommended Textbooks
Recorded Seminars
A series of short seminars covering all aspects of MIQE compliant qPCR are available here.
These seminars include basic qPCR topics and a specific series covering aspects of the MIQE guidelines:
qPCR Topics
MIQE Guidelines
Assay Design Tools
Assay design is a worthwhile investment of time and effort. There are several complimentary design services provided by oligo manufacturers and independent websites that support the design of a custom assay and will provide all the information regarding the assay design.
Predesigned Assays
Software
General Information
We provide these links as a courtesy for our customers. Inclusion on this list does not imply endorsement or validation of applications or results. We will neither collect nor make available any data, analysis or content information.
Products and Support
CleanAmp™ dNTP
CleanAmp™ dNTPs are modified nucleoside triphosphates that block DNA polymerase nucleotide incorporation. CleanAmp™ dNTPs are activated by the initial heating step and subsequent denaturing steps in typical hot start PCR cycling conditions. This process limits the amount of activated dNTPs during each cycle of PCR, allowing for more specific and efficient amplification of the desired product and also reducing or completely avoiding mis-priming or primer dimer formation. CleanAmp™ dNTPs provide a more affordable solution than hot start enzymes for a variety of PCR-based applications.
Custom DNA and RNA Oligos
As the world’s leading supplier of custom DNA and RNA oligonucleotides, we provide an unrivalled product portfolio, including a wide selection of synthesis scales, modifications and format options to fit all of your PCR and qPCR research needs. Add Locked Nucleic Acid to your primers for increased thermal stability and hybridization specificity (available outside of the USA).
For more information, visit our Oligo information page.
Custom qPCR Probes
Whether your application is gene expression analysis, genotyping, pathogen detection, or other, our wide selection of reporter and quencher dyes will support all major qPCR detection chemistries, including Dual-Labeled Probes, Molecular Beacons, LightCycler® Probes and Scorpions® Probes. Add Locked Nucleic Acid® (Locked Nucleic Acid®) to your probe or Molecular Beacon for increased thermal stability and hybridization specificity.
For more information, visit our informational page.
Extract-N-Amp™ PCR Kits
Our Extract-N-Amp™ PCR technology provides all the reagents necessary to extract DNA from a wide variety of cells and tissues and amplify targets of interest by PCR. A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR ReadyMix™ for amplification directly from the extract. The kit contains validated protocols to extract and amplify genomic DNA from a variety of starting materials.
For more information, visit our informational page.
GenElute™ Endotoxin-free Plasmid Prep Kit
This line of kits offers a simple, rapid, cost effective method for isolating endotoxin-free plasmid DNA from recombinant E. coli cultures. Endotoxins (also known as lipopolysaccharides or LPS) are often co-purified with plasmid DNA and significantly reduce transfection efficiencies in endotoxin-sensitive cell lines. High-quality, endotoxin-free DNA is ready for immediate use for the most demanding applications, including transfection into endotoxin-sensitive cells.
For more information, visit our informational page.
GenElute™ Genomic DNA Kits
The GenElute Genomic DNA Purification Kits provide a simple and convenient method to isolate pure, high molecular weight DNA from a variety of mammalian sources. These kits use a silica-based membrane, specially selected for genomic DNA purification. Mini and 96-well formats are available.
For more information, visit our informational page.
GenElute™ HP Plasmid Prep Kits
GenElute™ HP Plasmid Prep Kits offer a simple, rapid, cost effective solution for plasmid preparation from E. coli cultures. These kits combine silica-based membrane technology and the convenience of spin or vacuum format. The recovered plasmid DNA is ready for immediate use in applications such as restriction digest, cloning, PCR, transfection and sequencing.
Mini, 96-well, Midi, Maxi and Mega formats are available.
For more information, visit our informational page.
KiCqStart® Primers
Compatible with any thermal cycler, KiCqStart Primers areready-to-order, pre-designed primer pairs for quantifying gene expression by SYBR® Green I RT-qPCR. To comply with MIQE, they have been developed using sophisticated bioinformatics tools and validated in silico to avoid off-target amplification and all sequences are provided at purchase. KiCqStart Primers are available as up to three ranked sets of forward and reverse primer pairs for all genes from common model organisms.
For more information, visit our informational page.
KiCqStart SYBR Green qPCR ReadyMix™
KiCqStart SYBR Green qPCR ReadyMix is a 2× concentrated, ready-to-use reaction cocktail that contains all components, except primers and template, for real-time quantitative PCR (qPCR). This unique combination of proprietary buffer, stabilizers and KiCqStart Taq DNA Polymerase delivers maximum PCR efficiency, sensitivity, specificity and robust fluorescent signal using fast, or conventional, cycling protocols with SYBR Green I qPCR.
Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. We offer a variety of reagents to accommodate your instrument requirements.
Cat. No. KCQS00: For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
Cat. No. KCQS01: Low ROX™, for ABI and Stratagene instruments,
Cat. No. KCQS02: with ROX™ for ABI instruments
Cat. No. KCQS03: iQ™, with fluorescein for Bio-Rad systems
LuminoCt® qPCR ReadyMix™
LuminoCt qPCR ReadyMix is designed to deliver unsurpassed assay speeds without sacrificing accuracy, precision, or sensitivity. Inclusion of JumpStart™ Taq antibody in the ReadyMix eliminates polymerase activity at ambient temperatures without causing the performance issues associated with chemically modified hot start Taq. This allows for very rapid activation of the Taq and delivers unparalleled assay sensitivity, while allowing for benchtop reaction setup.
Cat. No. L6669: For fast probe-based quantitative PCR
LuminoCt® SYBR® Green qPCR ReadyMix™
LuminoCt SYBR Green qPCR ReadyMix is designed to deliver unsurpassed assay speeds without sacrificing accuracy, precision, or sensitivity Inclusion of JumpStart™ Taq antibody in the ReadyMix eliminates polymerase activity at ambient temperatures without causing the performance issues associated with chemically modified Hot Start Taq. This allows for very rapid activation of the Taq and delivers assay sensitivity, while allowing for benchtop reaction setup.
Cat. No. L6544: For fast SYBR Green quantitative PCR
MISSION® siRNA
Our MISSION line of siRNA are designed utilizing the Rosetta siRNA Design Algorithm and provide a guarantee of ≥75% knockdown on 2-out-of-3 sequences per gene.
Custom siRNA
High quality and cost-effective custom siRNA sequence design available for your unique or custom sequence through our free in-house design service. Choose from a wide range of modifications, purifications and quantities to tailor to your specific needs.
For more information, visit our informational page.
MISSION esiRNA
MISSION esiRNAs are endoribonuclease-prepared siRNA pools comprised of a heterogeneous mixture of siRNAs that all target the same mRNA sequence. These multiple silencing triggers lead to highly specific and effective gene silencing while providing lower off-target effects. MISSION esiRNA are available individually (pre-designed or custom), as custom arrayed panels, or genome scale libraries that target either human or mouse genes.
For more information, visit our informational page.
MISSION miRNA Mimics
Human miRNA mimics are available in a ready-to-use miRBase version 17 library or choose from over 1,900 individual mimics. Custom mimics are available for all species, including the current miRBase version content. Our mimics utilize a unique design and modification to significantly reduce possible sense strand off-target effects.
For more information, visit our informational page.
MISSION Pre-designed siRNA
The most complete collection of pre-designed siRNA for human, mouse and rat. Multiple sequences are available for every gene, with rankings to indicate performance. Sequences are updated with each version of NCBI RefSeq.
For more information, visit our informational page.
MystiCq® microRNA cDNA Synthesis Mix
Starting with total RNA, or samples pre-enriched or microRNAs, the MystiCq microRNA cDNA Synthesis Mix kit contains all the components necessary to convert mature microRNAs into cDNA templates for qPCR. Since microRNAs are not naturally polyadenylated, the first step in the cDNA synthesis process is to polyadenylate the microRNAs through a poly(A) polymerase reaction. Next, the ReadyScript® reverse transcriptase, an oligodT adapter primer and other required reagents are added to convert the polyadenylated microRNAs into cDNA. The adapter primer includes a unique sequence at the 5’ end that is complementary to the sequence of the MystiCq Universal PCR Primer. The kit also contains a Positive Control Primer that is specific to SNORD44, a small nucleolar RNA that is widely expressed in most human tissues. Sufficient amounts of the Poly(A) Tailing Buffer and the MicroRNA cDNA Reaction mix are included in the kit to allow for the inclusion of no poly(A) polymerase and no reverse transcriptase control reactions. To analyze individual microRNAs, use the MystiCq Universal PCR Primer (which targets the adapter primer sequence) together with the MystiCq microRNA qPCR Assay Primer and the MystiCq microRNA SYBR Green qPCR ReadyMix.
Cat. No. MIRRT: MystiCq® microRNA cDNA Synthesis Mix
MystiCq® microRNA qPCR Assay Primer
The MystiCq microRNA qPCR Assay Primer is an integral part of the MystiCq microRNA qPCR Assay System. It has been designed to target specific microRNAs with the MystiCq Universal PCR primer and the MystiCq microRNA SYBR Green qPCR ReadyMix on cDNA templates generated by the MystiCq microRNA cDNA Synthesis Mix kit.
For more information, visit our informational page.
MystiCq® microRNA SYBR® Green qPCR ReadyMix™
The MystiCq microRNA SYBR Green qPCR ReadyMix is one of the integral components of the MystiCq microRNA qPCR Assay System. This ReadyMix is a specially-formulated 2× concentrate with an antibody-based Hot Start Taq DNA polymerase and all the optimized components needed for highly specific, reliable and reproducible microRNA qPCR assays. Simply add together the specific MystiCq microRNA qPCR Assay Primer, the MystiCq Universal PCR primer and cDNA template generated by the MystiCq microRNA cDNA Synthesis Kit. Since there are different methods used for fluorescent normalization on different realtime PCR instruments, there are platform MystiCq microRNA SYBR Green qPCR ReadyMixes available.
Cat. No. MIRRM00: Formulated for systems that do not require normalization
Cat. No. MIRRM01: Low ROX™ formulation for miRNA RT-qPCR
Cat. No. MIRRM02: with ROX™, formulation for miRNA RT-qPCR
Cat. No. MiRRM03: Formulation for miRNA RT-qPCR on iQ Bio-Rad platforms
OligoArchitect™ Online
OligoArchitect Online is a free, robust qPCR assay design tool powered by the industry standard Beacon Designer™ platform from PREMIER Biosoft. In addition to providing primer and probe homology searching and template secondary structure analysis, additional features of OligoArchitect Online include automatic retrieval of template sequences, visualization of the template with positional display of regions of interest, design quality ratings and tools for alternative and multiplex designs.
For more information, visit our informational page.
OligoEvaluator™
OligoEvaluator is a free, online calculator of oligonucleotide sequence analysis, re-suspension and dilution requirements that is powered by PREMIER Biosoft. In addition to providing information such as melting temperature and molecular weight, additional features include secondary structure analysis and BLAST searching.
For more information, visit our informational page.
ReadyScript® cDNA Synthesis Mix
ReadyScript® cDNA Synthesis Mix is a sensitive and easy-touse solution for two-step RT-PCR. This 5× concentrated mix provides all necessary components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, ReadyScript® reverse transcriptase and stabilizers. ReadyScript® is a RNase H(+) derivative of MMLV reverse transcriptase, optimized for reliable cDNA synthesis over a wide dynamic range of input RNA concentrations. The unique blend of oligo (dT) and random primers in the ReadyScript® cDNA Synthesis Mix works exceptionally well with a wide variety of targets. It is optimized for the production of targets <1 kb in length. ReadyScript® cDNA Synthesis Mix produces excellent results in both real-time and conventional RT-PCR.
Cat. No. RDRT: Complete reagent for first strand cDNA synthesis for RT-qPCR
Design Your Probe Online with OligoArchitect™
We are pleased to offer OligoArchitect for all of your primer and probe design requirements. OligoArchitect is complimentary and includes both our online design tool (complete a design by yourself) and our consultative service (have an expert complete a design for you).
Enter the Gateway
Our OligoArchitect Online v4.0 design tool is powered by the industry standard Beacon Designer™ (PREMIER Biosoft). To start here, visit our infomational page.
When You Need a Consultant
For complex and demanding applications, utilize our consultative service to ensure the success of your PCR, qPCR, or other assays.
With personal consultation from our expert, technical support scientists, your request, including all sequences and data analysis, will be MIQE compliant and usually returned to you within 24 hours.
If required, you will also receive followup assay optimization, data analysis assistance, and troubleshooting support.
Do Your Own Design
Open the design tool with the click of a button.
Reference documents found here include a Glossary of Parameters and the Exon Design Protocol. Both are available
in PDF format.
This tool has been enhanced for a third time and is now able to create designs with Locked Nucleic Acid® (Locked Nucleic Acid).
Login or Register with Us
Are you ready to start? Register as a new user and choose your area. Returning users can login and proceed.
To start, visit oligoarchitect.com
Easy to Use
Type in the name of your assay, enter your sequence, and click Search (see OligoArchitect™ Assay Design).
All reported sequences, associated properties, and assay parameters are available for export to Excel and convenient email ordering.
Trademarks
The following trademarks and registered trademarks are accurate to the best of our knowledge at the time of publicaton.
Trademarks noted as registered are registered in the United States and possibly other jurisdictions. For the latest information on a specific product, see our website or consult individual manufacturers.
Agilent Technologies, Inc. — Mx3000P®, Mx3005P®, Mx4000™
Applied Biosystems — VIC®
BioMark Technologies Inc. — BioMark™
bioMérieux Société anonyme à conseil d’administration — THxID®
Bio-Rad Laboratories, Inc. — Bio-Rad®, CFX384™, CFX96™, Chromo4™, ddPCR™, Droplet Digital™, iQ™, MiniOpticon™, MyiQ™, Opticon™, QX100™
Bio-Rad QL, Inc — Quantasoft®
Biosearch Technologies, Inc— BHQ®, Black Hole Quencher®, Cal Fluor®, Quasar®
Boehringer Ingelheim International GmbH — GILOTRIF®
Cepheid, Inc. — SmartCycler®
Eppendorf AG — Eppendorf®, Mastercycler®
Excel Scientific, Inc. — ThermalSeal RT2RR™, ThermalSeal RTS™
Exiqon A/S — LNA®
Cytiva — Cy®
GlaxoSmithKline LLC — Mekinist®, Tafinlar®
Lehn & Fink, Inc. — Amphyl®
Life Technologies — Applied Biosystems®, FAM™, HEX™, JOE™, OpenArray®, QuantStudio®, QuBit®, ROX™, StepOne™, StepOnePlus™, SYBR®, TAMRA™, TET™
Merck KGaA, Darmstadt, Germany and/or its affiliates — BlueView™, DirectLoad™, Extract-N-Amp™, GenElute™, JumpStart™, LuminoCt®, miRPremier®, MISSION®, OligoArchitect™, OligoEvaluator™, Onyx Quencher™, OQ™, ReadyMix™, ReadyScript®, REDTaq®
Microsoft Corporation — Excel®, Microsoft®
Molecular Research Center, Inc. — RNAzol®, TRI Reagent®
OSI Pharmaceuticals, Inc. — TARCEVA®
Premier Biosoft International — Beacon Designer™
Qiagen Group — Rotor-Gene®, Scorpions®, therascreen®
RainDance Technologies, Inc. — RainDrop™
Roche Diagnostics GmbH — MycoTOOL®
Roche Diagnostics Operations, Inc. — cobas®
Roche Molecular Systems, Inc. — LightCycler®, Roche®, TaqMan®
The Dow Chemical Company or an affiliated company of Dow — Triton®
Thermo Fisher Scientific, Inc. — NanoDrop®
TriLink BioTechnologies, Inc. — CleanAmp™
Used under license — KiCqStart®, MystiCq®