Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Plasma profiling of ageing and disease on antibodies
By: Jochen M. Schwenk, Biobank Profiling Group, Peter Nilsson and Mathias Uhlén, Poster - The Human Potein Atlas
Science for Life Laboratory Stockholm, KTH -­- Royal InsJtute of Technology, Box 1031, SE-­-171 21 Solna, Sweden
Download the poster (2.2 Mb PDF)
Affinity proteomic profiling of plasma
High-throughput methods for screening samples stored in biobanks are of growing interest, since they would allow to discover biomarkers of ageing and disease for diagnostics and patient care at a larger scale. With more than 15,000 well-characterised affinity reagents in the Human Protein Atlas (www.proteinatlas.org), a systematic exploration of protein profiles from serum and plasma can be achieved by single-binder affinity arrays. Assays have been developed that profile up to 384 proteins in each of the 384 labeled, non-fractionated samples in parallel on magnetic, color-coded beads.
References - Uhlén M, et al. (2010) Nat Biotech. 28(12): 1248-50; Schwenk JM, et. al. (2010) Mol Cell Prot. Nov;9(11):2497-507
Strategy for profiling of plasma using antibodies
Antibodies are being generated based on bioinformatics target selection and selected based on specificity and concentration. Single-binder bead arrays are then created with 384 antibodies each. Studies are then designed by the randomization of to apply bead arrays for profiling discovery cohorts. Interesting antibodies are then qualified for further verification in new cohorts. The overall aim is to build sandwich assays for qualification via epitope mapped binders.
Strategy for profiling of plasma using antibodies
Exploration of plasma protein profiles
The affinity--based strategy has been applied to explore profiles males and female aged 5-85 (n=384) as well as to diseases. For the latter, more than 20 different disease entities (n=570) were profiled in parallel and to propose antibodies for single disease verification.
Identification of protein profiles associated with age
Linear regression (left); Loess Clustering (mid); Signature score (right)
Identification of protein profiles associated with age
Identification of protein profiles associated with disease
The boxplots represent the differential plasma protein profiles generated by four different antibodies for matched comparisons in Alzheimer's Disease (alz) and Mild Cognitive Impairment (mci).
Identification of protein profiles associated with disease
Related Links