[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Protein Expression](https://www.sigmaaldrich.com/US/en/applications/protein-biology/protein-expression)Protein Expression Systems
# Protein Expression Systems
## Introduction
The development of genetic engineering and cloning has opened many possibilities of expression and isolation of heterologous proteins for research purposes. Considerable advances in technology have enabled expression and isolation of recombinant proteins in large scale. However, for large scale applications such as enzyme, antibody or vaccine production, the amount of protein required is considerably high. In such cases the system in which the protein is expressed must be easy to culture and maintain, grow rapidly, and produce large amounts of protein. Moreover, mammalian proteins also undergo various post-translational modifications. These requirements led to the discovery of protein expression systems. The various protein expression systems are bacteria, yeast, insect or mammalian systems.
The following factors determine the type of expression system used to produce recombinant proteins:
- time spent in expressing the protein
- ease of handling the expression system
- amount of protein needed
- mass of the protein
- type of post-translational modifications, number of disulfide bonds
- destination of the expressed protein
The process of expressing a recombinant protein in an expression system requires the following information/components.
- identification of the gene that encodes the protein of interest
- generation of cDNA from the respective mRNA
- selection of suitable expression vector to insert the gene sequence
- selection of suitable system that can express the vector
- appropriate screening and scaling up methods
The core steps involved in producing the desired recombinant protein are similar across the various expression systems (__Figure 1__).
__Figure 1.__Steps involved in optimizing protein expression system to produce desired recombinant protein
## Bacterial protein expression systems – *Escherichia coli*
Bacteria act as rapid and simple systems of expressing recombinant proteins due to the short doubling time. The media required to culture them are not expensive and the methods adapted to scale-up bioproduction are straightforward. The most widely used host system is *E. coli* since there is ample knowledge about its genetics, genome sequence and physiology. The genetic manipulation is easy and it also grows to high densities and is suitable for large-scale fermentations. However, the cell wall of *E. coli* contains toxic pyrogens and the expressed proteins may have to be extensively tested before use.
__Figure 2.__Bacterial protein expression system – Escherichia coli
Expression of protein using *E. coli* involves the following steps:
- use of competent *E. coli* (Catalog Numbers [CMC0001](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0001), [CMC0004](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0004), [CMC0014](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0014)) cells to take up DNA sequence of interest
- integration of the DNA into bacterial genome or circularization of the DNA sequence to exist as a plasmid
- selection of transformed *E. coli* using a selection marker (antibiotic) (Catalog Numbers [L5667](https://www.sigmaaldrich.com/US/en/product/sigma/L5667), [L0168](https://www.sigmaaldrich.com/US/en/product/sigma/L0168), [L0543](https://www.sigmaaldrich.com/US/en/product/sigma/L0543), [L0418](https://www.sigmaaldrich.com/US/en/product/sigma/L0418), [L8795](https://www.sigmaaldrich.com/US/en/product/sigma/L8795))
- expansion of selected *E. coli* to a higher scale in appropriate culture media, such as classic LB options (product numbers [L2542](https://www.sigmaaldrich.com/US/en/product/sigma/L2542), [L3522](https://www.sigmaaldrich.com/US/en/product/sigma/L3522), [L3147](https://www.sigmaaldrich.com/US/en/product/sigma/L3147)) or EnPresso™ B Growth Systems (product number [B11001](https://www.sigmaaldrich.com/US/en/product/sigma/B11001))
- isolation and purification of intracellular/secreted proteins
__Features__
- low cost culture methods
- flexible system – can carry plasmids with multiple promoters, tags and restriction sites
- easy to scale up and produce higher yield of protein
## Yeast protein expression systems *– Saccharomyces cerevisiae*
The highly developed genetic system, ease of use, reduced time input and costs have made *S. cerevisiae* an attractive organism for the expression and production of recombinant proteins. Yeasts are able to carry specifically designed plasmids and this ability is valuable in a recombinant protein expression system. The plasmid used consists of restriction sites that can be used to insert the gene sequence of interest. Transformation of yeasts with the plasmid produces the desired protein and can be appropriately scaled up.
__Figure 3.__Yeast protein expression system – Saccharomyces cerevisiae
Expression of protein using *S. cerevisiae* involves the following steps:
- use of competent *E. coli* cells to take up DNA sequence of interest (Catalog Numbers [CMC0001](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0001), [CMC0004](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0004), [CMC0014](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0014))
- integration of the DNA into bacterial genome or circularization of the DNA sequence to exist as a plasmid
- selection of transformed *E. coli* using a selection marker (antibiotic) (Catalog Numbers [L5667](https://www.sigmaaldrich.com/US/en/product/sigma/L5667), [L0168](https://www.sigmaaldrich.com/US/en/product/sigma/L0168), [L0543](https://www.sigmaaldrich.com/US/en/product/sigma/L0543), [L0418](https://www.sigmaaldrich.com/US/en/product/sigma/L0418), [L8795](https://www.sigmaaldrich.com/US/en/product/sigma/L8795))
- expansion of selected *E. coli* in appropriate culture media, such as classic LB options (product numbers [L2542](https://www.sigmaaldrich.com/US/en/product/sigma/L2542), [L3522](https://www.sigmaaldrich.com/US/en/product/sigma/L3522), [L3147](https://www.sigmaaldrich.com/US/en/product/sigma/L3147)) or EnPresso™ Y Defined Media (product number [Y22001](https://www.sigmaaldrich.com/US/en/product/sigma/Y22001))
- isolation of DNA or plasmid
- transformation into yeast (yeast transformation kit, Catalog Number [YEAST1](https://www.sigmaaldrich.com/US/en/product/sigma/YEAST1))
- screen the transformants for integration of DNA into yeast chromosome
- selection and scaling-up of high expressing yeast clones in appropriate culture media (Catalog Numbers [Y1375](https://www.sigmaaldrich.com/US/en/product/sigma/Y1375), [Y1501](https://www.sigmaaldrich.com/US/en/product/sigma/Y1501), [Y1751](https://www.sigmaaldrich.com/US/en/product/sigma/Y1751), [Y2001](https://www.sigmaaldrich.com/US/en/product/sigma/Y2001), [Y1376](https://www.sigmaaldrich.com/US/en/product/sigma/Y1376), [Y0750](https://www.sigmaaldrich.com/US/en/product/sigma/Y0750))
- isolation and purification of intracellular/secreted proteins
### __Features__
- low cost culture methods
- suitable for both intracellular and secreted proteins
- provides eukaryotic post-translational glycosylation of proteins although it results in high
## Insect cell expression systems – *Sf*9 and *Sf*21
The cell lines derived from *Spodoptera frugiperda*, *Sf*9 and *Sf*21, are frequently used as recombinant protein expression system. Baculovirus is a lytic, dsDNA virus, routinely amplified in cells of the insects belonging to Lepidoptera family. It is noninfectious in vertebrates and its promoters are inactive in mammalian cells.
__Figure 4.__Insect cell expression systems – Sf9 and Sf21
Expression of protein using baculovirus/insect cells involves the following steps:
- use of competent *E. coli* cells to take up DNA sequence of interest (Catalog Numbers CMC0001, CMC0004, CMC0014)
- integration of the DNA into bacterial genome or circularization of the DNA sequence to exist as a plasmid
- selection of transformed *E. coli* using a selection marker (antibiotic) (Catalog Numbers L5667, L0168, L0543, L0418, L8795)
- expansion of selected *E. coli* in appropriate culture media (Catalog Numbers L2542, L3522, L3147)
- isolation of DNA or plasmid
- preparation of a second plasmid containing viral genes required for multiplication and formation of virus particles
- co-transfection of the expression plasmid and the second plasmid into *Sf*9 or *Sf*21 insect cells
- purification of the recombinant viral stock
- amplification of the virus and additional plaque assays to increase the titer of the recombinant viral stock
- infection of the insect cells with high-titer recombinant virus stock
- isolation and purification of intracellular/secreted proteins
__Features__
- recombinant protein is highly expressed during the last phases of lytic cycle before cell lysis
- suitable to generate both cytoplasmic and secreted proteins
- disulfide bonds in proteins are efficiently generated
- provide majority of post-translational modifications found in mammalian cells
## Mammalian cell expression systems – HEK293 and CHO
The main challenge of using mammalian cells for expressing recombinant proteins is the reduced efficiency and levels of the protein expressed. However, cell lines such as HEK293 and CHO have been developed as efficient transient and stable expression systems, respectively. HEK293 cells are transiently transfected using liposomes, calcium phosphate or PEG as transfection reagents. Though transient expression is relatively easy and simple, scaling up is technically challenging. CHO cells are commonly used to stably express large quantities of recombinant proteins. The process involves transfection of DHFR-deficient CHO cells with the gene of interest and a DHFR selection cassette. The transfected cells are then screened in the presence of methotrexate to obtain stably transfected cell pools. The selection and expansion process takes 2-3 months
__Figure 5.__Mammalian cell expression systems – HEK293 and CHO
Transient or stable protein expression using mammalian cells involves the following steps:
- use of competent *E. coli* cells to take up DNA sequence of interest (Catalog Numbers [CMC0001](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0001), [CMC0004](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0004), [CMC0014](https://www.sigmaaldrich.com/US/en/product/sigma/CMC0014))
- integration of the DNA into bacterial genome or circularization of the DNA sequence to exist as a plasmid
- selection of transformed E. coli using a selection marker (antibiotic) (Catalog Numbers [L5667](https://www.sigmaaldrich.com/US/en/product/sigma/L5667), [L0168](https://www.sigmaaldrich.com/US/en/product/sigma/L0168), [L0543](https://www.sigmaaldrich.com/US/en/product/sigma/L0543), [L0418](https://www.sigmaaldrich.com/US/en/product/sigma/L0418), [L8795](https://www.sigmaaldrich.com/US/en/product/sigma/L8795))
- optional storage of clones in CloneStable™
- expansion of selected *E. coli* in appropriate culture media (Catalog Numbers [L2542](https://www.sigmaaldrich.com/US/en/product/sigma/L2542), [L3522](https://www.sigmaaldrich.com/US/en/product/sigma/L3522), [L3147](https://www.sigmaaldrich.com/US/en/product/sigma/L3147))
- isolation of DNA or plasmid
- transfection of the expression plasmid to mammalian cells using X-tremeGENE™ Transfection Reagents.
- selection of stable clones
- expansion of clones for transient batch expression OR expansion of clones for 2-3 months for stable expression
- isolation and purification of intracellular/secreted proteins
__Features__
- transient expression is easy and rapid
- provide all the post-translational modifications found in mammalian cells
- stable transfection results in higher yield, scalability and reproducible production
The following table compares the four protein expression systems and their respective advantages and disadvantages:
Protein expression systemAdvantages__Disadvantages__
*E. coli*Rapid expressionNo eukaryotic post-translational modifications
Less expensiveSome proteins are not properly folded
Simple process scale-upProteins are rarely secreted
Well characterized genetics
*S. cerevisiae*Moderately rapid expressionCharacteristic N-linked glycan structures of proteins are different when compared to the typical mammalian proteins
Works well for secreted and intracellular proteinsRequires use of yeast secretion signal peptides
Less expensiveEnhanced safety precautions are required
Most protein folding and post-translational modifications are possible
Baculovirus/Insect cellsModerately rapid expressionCharacteristic N-linked glycan structures of proteins are different when compared to the typical mammalian proteins
Works well for secreted, membrane and intracellular proteinsExpensive
Most protein folding and post-translational modifications are possibleDifficult to scale-up
Mammalian cells (transient expression)Moderately rapid expressionExpensive
Works well for secreted and membrane proteinsYields of intracellular proteins are low
All protein folding and most authentic post-translational modifications are possibleDifficult to scale-up
Mammalian cells
(stable expression)All protein folding and most authentic post-translational modifications are possibleExpensive
Works well for secreted and membrane proteinsYields of intracellular proteins are low
Difficult to scale-up
Lengthy expression time
Choose an expression system that expresses the protein with all the desirable post-translational modifications. Irrespective of the choice, we offer a wide range of competent cells, insect and mammalian cell lines and related products that suit any protein expression system.
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related articles__
- [Residual Peroxide Determination in Food Packaging](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/extracellular-microenvironment)
- [Hypoxia Detection Assays](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/hypoxia-detection-assays)
- [3xFLAG® System Expression Vectors for Ultra-Sens](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/3x-flag)
- [FLAG® Tag Bacterial and Mammalian Expression Vectors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/vector-maps)
- [Cloning & Expression Vectors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/vector-systems)
- [p53: DNA Damage Response and Cancer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/epigenetics/p-fiftythree-dna-damage-response-and-cancer)
- [MISSION® shRNA Lentiviral Transduction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/gene-expression-and-silencing/mission-shrna-lentiviral-transduction)
- [Duolink® Proximity Ligation Assay Instructional Video](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-and-nucleic-acid-interactions/duolink-assay)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related product categories__
- [Microbial Culture Media](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media)
- [Molecular cloning & protein expression](https://www.sigmaaldrich.com/US/en/products/molecular-biology-and-functional-genomics/cloning-and-expression/molecular-cloning-and-protein-expression)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)