Vasopressin and Oxytocin Receptors
The pituitary hormones arginine vasopressin (AVP) and oxytocin (OT) are structurally related cyclic nonapeptides that act as hormones and neurotransmitters. Both peptides are synthesized in neurons in the hypothalamus that project to the posterior pituitary from which they are released into the circulation. In addition, OT and AVP neurons innervate various regions of the brain and spinal cord. Their amino acid sequences differ only in two of the nine amino acids, but despite this similarity they exhibit distinct biological activities. AVP/OT receptor subtypes V1a, V1b, V2, OT have been cloned from rat and human tissues and characterized. They are members of the G protein-coupled superfamily of receptors and show significant structural homology to one another. The receptor selectivity of OT and AVP for their own receptors is not absolute and significant cross-talk can occur with OT at AVP receptors (and vice versa) at higher concentrations.
A primary physiological role of AVP involves regulation of cardiovascular function where its contractile actions on vascular smooth muscle, via V1a receptors, maintain peripheral resistance under certain adverse conditions such as hemorrhage, and its antidiuretic actions on kidney, via V2 receptors, correct fluctuations in blood osmolality. Binding of AVP to the V1a receptor subtype also stimulates glycogenolysis in the liver and promotes platelet aggregation. In addition, V1a receptors are present in the CNS where they may be involved in certain behaviors such as learning and memory. Activation of the recently discovered V1b (also known as V3) receptor by hypothalamic AVP causes adrenocorticotropic hormone release from the anterior pituitary and is believed to mediate various stress-related behavioral responses.
OT binds to receptors in the uterus and mammary gland to mediate important functions in parturition such as contraction of the uterine myometrium during labor and the mammary myoepithelium postpartum to elicit milk letdown. In addition, OT receptors are located in a variety of other peripheral tissues as well as the brain where OT may have physiological effects on cardiovascular, renal, endocrinological and behavioral functions. In particular, OT has been shown to promote maternal, sexual and affiliative-type behaviors in various animal species including nonhuman primates. At present, only one OT receptor subtype has been identified pharmacologically or structurally.
The diverse functions of OT and AVP give rise to a number of possible therapeutic opportunities based on the modulation of their receptor activity. Two examples of current therapeutics are OT itself, for the promotion of labor and delivery, and desmopressin (dDAVP), a V2 agonist for the treatment of diabetes insipidus in which there is a deficiency of circulating AVP. Nonpeptide V2 agonists such as OPC 51803 have recently been described. Nonpeptide OT agonists have also been disclosed and are being explored for possible utilities in sexual function and affiliative disorders such as social phobias and obsessive-compulsive behavior. The OT antagonist atosiban was approved and launched (Tractocile®) in Europe for the treatment of preterm labor. Blockade of vascular V1a receptors with OPC 21268, atosiban or YM471 may have utility in hypertension and dysmenorrhea, whereas an aquaretic response (water diuresis) caused by antagonism of renal V2 receptors with OPC 41061(tolvaptan) or SSR121463A could be useful in counteracting fluid retention and hypo-osmolality resulting from a variety of conditions. A V2 antagonist may also be useful in the treatment of congestive heart failure, particularly in combination with a V1a antagonist or, with simply, a compound exhibiting both properties such as YM471 and conivaptan (YM087), used for the treatment of hyponatremia. Selective V1b antagonists such as SSR149415 are being investigated for possible utility in various stress-related disorders such as depression and anxiety.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Currently Accepted Name V1a V1b V2 OT
Alternate Name V1 V3    
Structural Information 418 aa (human) 424 aa (human) 371 aa (human) 391 aa (human)
Preferred Endogenous Ligand Vasopressin (V9879, V0377) Vasopressin (V9879, V0377) Vasopressin (V9879, V0377) Oxytocin (O6379)
Selective Agonists [Phe2,Orn8]VT deamino-[D-3’-(pyridyl)-Ala2]AVP
dDAVP (V1005)
dVDAVP (V2013)
OPC 51803 (non-peptide)
Compound 39 (nonpeptide)
Selective Peptide Antagonists desGly9-d(CH2)5 AVP
d(CH2)5-[Tyr(Me)2] AVP
dP[Tyr(Me)2] AVP d(CH2)5-[D-Ile2,Ile4] AVP
desGly(NH2)9-d(CH2)5 [D-Ile2,Ile4] AVP
desGly(NH2)9d(CH2)5 [Tyr(Me)2Thr4]OVT
Atosiban (A3480)
Selective Non-Peptide Antagonists OPC 21268 (O1016)
SR 49059 (S5701)
SSR149415 OPC 31260 (O1266)
SR 121463
OPC 41061 (Tolvaptan) (T7455)
L-368,899 (L2540)
Signal Transduction Mechanisms Gq/11 (increase IP3/DAG) Gq/11 (increase IP3/DAG) Gs (increase cAMP) Gq/11 (increase IP3/DAG)
Radioligands of Choice [3H]-AVP
[3H]-d(CH2)5-[Tyr(Me)2] AVP
[3H]-SR 49059
[3H]-AVP [3H]-AVP
[3H]-desglycinamide-d(CH2) [D-Ile2,Ile4]AVP
[125I] d(CH2)5[Tyr(Me)2- Thr4-Tyr(NH2)9] OVT
Tissue Expression Vasculature Pituitary
Kidney Uterus
Mammary gland
Physiological Function Smooth muscle contraction ACTH release
Antidiuresis Smooth muscle contraction
Maternal, sexual behavior
Disease Relevance Hypertension
Congestive heart failure
Congestive heart failure
Preterm labor
Sexual dysfunction
The information in the Table refers largely to data obtained with rat receptors although some data exist for human receptors as well. It should be noted that substantial species differences in the affinities can exist between the pharmacology of rat and human receptors. For example, OPC 21268 is a high affinity antagonist ligand at rat, but not human receptors, and atosiban is a selective OT antagonist in rats, but has V1a selectivity at human receptors. Also, for several cases for the compounds in the Table, compounds that are selective for certain AVP receptor subtypes have not been evaluated fully for OT receptor affinity. It is recommended that the reader go to the literature of a given compound before making assumptions of affinity and degree of selectivity within and across species.
ANTAG III: β,β-(3-Thiapentamethylene)-β-mercaptopropionic acid1,D-Trp2,penicillamine6,Arg8-OT
AVP: Arginine vasopressin
dDAVP: [des-amino D-Arg4]Vasopressin
dVDAVP: [des-amino Val4,D-Arg8]Vasopressin
Compound 39: Pitt, et al., Bioorg Med Chem Lett., 14, 4385-4589 (2004).
L-362,662: 1-(1-{4-[1-(2-Methyl-1-oxidopyridin-3-ylmethyl)piperidin-4-yloxy]-2-methoxybenzoyl}piperidin-4-yl)-1,4-dihydrobenz[d][1,3]oxazin-2-one
L-368,899: 1S((-7,7-Dimethyl-2-endo-(2S-amino-4-(methylsulfonyl)butyramido)-bicyclo(2.2.1)-heptan-1-yl)methanesulfonyl)-4-(2-methylphenyl)piperazine
LVP: Lysine vasopressin
OPC 51803: (5R)-2-[1-(2-chloro-4-(1-pyrrolidinyl)benzoyl)-2,3,4,5-tetrahydro-1H-1-benzazepin-5-yl]isopropylacetamide
OPC 31260: (+)-5-Dimethylamino-1-(4-[2-methylbenzoylamino]benzoyl)-2,3,4,5-tetrahydro-1H-benzazepine
OPC 41061: 7-Chloro-5-hydroxy-1-[2-methyl-4-(2-methylbenzoylamino)benzoyl]-2,3,4,5-tetrahydro-1H-1-benzazepine
OPC 21268: 1-(1-{4-[3-Acetylaminopropoxy]benzoyl}-4-piperidyl)-3,4-dihydro-2(1H)-quinolinone
OT: Oxytocin
OVT: [Orn8]Vasotocin
Phaa: Phenylacetic acid
SR 121463A: (1-[4-(N-Tert-butylcarbamoyl)-2-methoxybenzene sulfonyl]-5-ethoxy-3-spiro-[4-(2-morpholinoethoxy)cyclohexane]indol-2-one
SSR149415: ((2S,4R)-1-[5-Chloro-1-[(2,4-dimethoxyphenyl)sulfonyl]-3-(2-methoxyphenyl)-2-oxo-2,3-dihydro-1H-indol-3-yl]-4-hydroxy-N,N-dimethyl-2-pyrrolidinecarboxamide
SR 49059: ((2S)1-[(2R,3S)-(5-Chloro-3-(2-chlorophenyl)-1-(3,4-dimethoxybenzene-sulfonyl)-3-hydroxy-2,3-dihydro-1H-indole-2-carbonyl]-pyrrolidine-2-carboxamide
SSR126768A: (4-Chloro-3-[(3R)-(+)-5-chloro-1-(2,4-dimethyoxybenzyl)-3-methyl-2-oxo-2,3-duhydro-1H-indol-3-yl]-N-ethyl-N-(3-pyridylmethyl)-benzamide
VDAVP: [Val4,D-Arg8]Vasopressin
VT: Vasotocin
YM087: 4’-[(2-Methyl-1,4,5,6-tetrahydroimidazo[4,5-d][1]benzazepin-6-yl)-carbonyl]-2-phenylbenzanilide
YM471: ((Z)-4’-{4,4-Difluoro-5-[2-(4-dimethyaminopiperidino)-2-oxoethylidene]-2,3,4,5-tetrahydro-1H-1-benzoazepine-1-carbonyl}-2-phenylbenzanilide
Similar Products