EnPresso® Y Defined Growth (yeast protein data)
Results of Yeast Culture (Proteins for Research)
The first tablet-based, chemically-defined growth system for yeast, EnPresso Y Defined, enables production of more protein in less time from cultures of Pichia pastoris using AOX promoters with methanol induction.
More active protein in less time
Data courtesy of Florian Krainer, University of Graz, Austria
Researchers obtained similar yields obtained in 3 days using EnPresso Y Defined in a baffled Ultra Yield Flask as compared to 7 days using YPD media in a baffled shake flask. Protein expressed was Candida Antarctica lipase B (CalB) in Pichia pastoris MutS phenotype.
Significant increase in protein yield/mL culture
Result courtesy of Els Van Damme, University of Ghent
Researcher compared EnPresso Y Defined  with a commercially-available growth media. Protein expressed is Rhizopus oryzae lipase in Pichia pastoris.
Up to 4-fold increase in yield of active protein
Data courtesy of A. Mursula and J. Panula-Perälä, Bioprocess Engineering Laboratory, Oulu
Researchers compared EnPresso Y Defined with conventional buffered minimal medium. Protein expressed was Rhizopus oryzae lipase in Pichia pastoris.
EnPresso is registered trademark of BioSilta Oy.
Related Links