Anti-HA Troubleshooting
Product No. ROAHA
Chemiluminescent or chromogenic signal weak or not visible
  • Poor isolation of tagged protein
— Use a different cell lysis procedure
  • Antibody too dilute
— Double the concentration of the Anti-HA and/or the secondary antibody
  • Too little protein on the gel
— Add more protein to gel.
  • Poor transfer of proteins from gel to membrane
— Increase the electrical current and/or the transfer time for the blot. Be sure there are no air bubbles between the membrane and gel during transfer.
  • Wrong type of membrane
— For maximum signal, use PVDF membranes for transfer.
  • Antibody incubation too short
— Incubate Anti-HA and/or the secondary antibody with the membrane blot for a longer time.
  • Signal development time too short
— Double the development time.
  • Wash time too long or too stringent
— Shorten the washing time. Omit Tween 20 from the Wash Buffer.
  • Enzyme on antibody conjugate inactivated by preservative
— Do not use sodium azide in any Western blot reagent if you use Peroxidase-conjugated antibodies.
  • Substrate inactive
— Make fresh dilution of substrate or start with a different stock of substrate.
  • Epitope tag sequence is not detectable due to Proteolytic cleavage
— Include protease inhibitors in lysis buffer.
  • Epitope tag sequence is not detectable due to low level of expression
— Use alternative expression system or optimize your expression system. Insert multiple tag sequences into target protein to increase avidity of antibody reaction.
  • Epitope tag sequence is not detectable due to premature translation termination resulting in loss of C-terminal tag
— Use alternative insertion site within the target gene for the epitope tag sequence.
High background additional bands on blot
  • Antibody too concentrated
— Reduce concentration of Anti-HA and/or secondary antibody by half.
  • Wash time too short
— Prolong wash time.Incubation of membrane with substrate too long — Leave blot membrane in substrate for a shorter time.
  • Wrong type of membrane
— For minimum background, use PVDF membranes for transfer.
  • Blocking Reagent too dilute
— Use nonfat dry milk (5% w/v) dissolved in Reagent Diluent as Blocking Reagent. Note: High concentrations of nonfat dry milk may reduce specific signal as well as background.
  • Contaminated reagents or equipment
— Use clean equipment, freshly prepared buffers, and new membranes. Always avoid touching membranes with bare hands; use gloves and forceps.
  • Secondary antibody binds untagged proteins.
— Use an F(ab′)2 fragment of a secondary antibody, rather than an intact IgG.
  • Heavy and light chains of primary antibody visible on blot membrane
— Use direct detection with peroxidase-conjugated monoclonal antibody to visualize tagged proteins.
  • Signal development time too long
— Reduce development time by half.
Related Links