cOmplete™ ULTRA Tablets, glass vials Troubleshooting
Product No. COUL-RO
Inhibition does not work
Increase of the concentration of the inhibitor cocktail and reduce the storage time.
Also, note that some ′′rare′′ proteases might not be inactivated. In general, it cannot be guaranteed that all proteases will be blocked. In some special cases, the addition of specific inhibitors may be indicated. In those cases, the customer must screen for the right inhibitor.
Protein not found in western blot
The loss of protein may be due to other factors:- Subunits that are separated (for example, as a result of using reduced conditions).- Inaccurate blotting conditions.- Detection problems.
Additional bands in western blots
Try running a blank on the gel containing only the inhibitor cocktail.If the inhibitor cocktail has been used at a very high concentration (other than as advised in the package insert), there may be slight bands in the low molecular weight region.
Interference with isoelectric focusing
This might be caused by one of the components that may be visible on the gel after staining. Apart from that, nothing else should disturb this application; try using a blank to verify this.
Inhibition of inhibition activity
Theoretically, there is no reason to believe that SDS or other detergents are not compatible with the inhibitor cocktail. However, no direct experiments have been carried out on this subject.
Note that Ca2+, Co2+ Ni2+, Mn2+, and Mg2+ can inhibit the inhibition activity.
Recommended solvent
One tablet can be dissolved in 5 ml double-distilled water. If the tablet will be dissolved in lysis buffer extra time may be required. Leaving the solution at +15 to +25 °C for 30 - 60 minutes or vortexing may help.
Interference with DTT
This has not been checked. From the components of the tablet, we would expect that reducing agents normally used in protein biochemistry (like mercaptoethanol, DTT up to 5 mM, or DTE) should not interfere with the function of the tablets.
cOmplete is a trademark of Roche.
Related Links