Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Quick Spin Columns for radiolabeled RNA purification Troubleshooting
Product No. 11273990001, 11274015001
Troubleshooting
Nucleotide contamination of filtered sample using mini Quick Spin Columns or Quick Spin Columns
Possible reasons are:
  1. Sample should not be applied to the side of the column, so that molecules to flow around the matrix; samples should be placed on top of the matrix for purification.
    Recommendation: Apply sample directly to center of the column bed.
  2. Column is overloaded, which causes nucleotides to flow through the matrix.
    Recommendation: Apply a sample containing 0.02 - 1.0 mg/ml nucleic acid. Apply no more than 50 μl sample to a G-25 column, and no more than 100 μl to a G-50 column.
  3. The incorrect g-force was used during centrifugation: At too high speeds the column matrix can collapse and unincorporated nucleotides may pass freely through the column.
    Recommendation: Use 1,100 x g for centrifugation spins. Make certain that the centrifuge is correctly calibrated.
  4. The inappropriate rotor was used during centrifugation.
    Recommendation: Use swinging-bucket rotor. A fixed-angle rotor may cause nucleotide contamination of the resulting sample.
  5. Column was vortexed too long or too vigorously during matrix resuspension:
    Recommendation:
    - Do not vortex column longer than 5 seconds.
    - Vortex the column at low speed only.
    - Do not use medium or high speed.
Poor sample recovery of mini Quick Spin Column or Quick Spin Column
Possible reasons for poor sample recovery:
  1. Centrifugation speed is too fast or the centrifugation time too short: Do not centrifuge the columns faster than the recommended speed.
  2. Matrix not evenly resuspended prior to packing step: To fully resuspend the matrix before packing step, do one of the following:
    - Invert column vigorously several times and flick the column sharply to help resuspending the matrix.
    - Vortex column gently (5 seconds or less, at low speed).
  3. The column was tipped to the side, which causes backflow of sample and reduced nucleic acids recovery: Keep the column upright during and after application of sample, especially after centrifugation.
  4. Sample volume too small (<20 μl):
    Recommendation: Do one of the following:
    - Add 1 x STE buffer to sample until total sample volume is 20 μl
    - After applying sample add 1x STE buffer to the matrix. Total volume applied (sample + STE buffer) must not be greater than the maximum sample volume recommended for the column.
    - At < 0.02 mg/ml DNA/RNA recovery may be low. To improve recovery of diluted samples, add carrier, such as glycogen, tRNA or sperm DNA, to sample.
  5. There is too much sample. At > 1.0 mg/ml, the sample is viscous and may not migrate through the columns easily, leading to poor recovery and / or contamination with smaller molecules.