Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Sera-Mag™ SpeedBeads Protein A/G Magnetic Particles
Protein enrichment
Sera-Mag SpeedBeads Protein A/G Magnetic Particles provide a fast and convenient method for manual and automated magnetic isolation of affinity bound antibodies or antigens to Protein A/G (Fig 1). These particles can be used for isolating antibodies from serum, cell culture supernatant or ascites, and for immunoprecipitation and co-immunoprecipitation of antigens from cell or tissue extracts.
  • Manual and automated magnetic isolation of IgA and IgG proteins is accomplished in a single isolation instead of running two separate isolations
  • Presence of Protein A/G binding sites allows use of the broadest range of antibody species and sub-classes
  • Lower non-specific binding for faster and cleaner Purification
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Cauliflower-like surface provides a much larger area for binding reactions than smooth surface particles
  • Uniform, nominal 1 μm diameter provides high surface area and excellent lot-to-lot reproducibility
SEM image showing the cauliflower-like surface of the Sera-Mag Magnetic SpeedBeads
Fig 1. SEM image showing the cauliflower-like surface of the Sera-Mag Magnetic SpeedBeads. The surface dramatically increases the overall surface area available for binding.
Sera-Mag SpeedBeads Protein A/G Magnetic Particles have a surface blocked and covalenty coupled to a recombinant Protein A/G. Protein A/G contains four Fc-binding domains from Protein A and two from Protein G. By taking advantage of the affinity provided by both proteins, these particles are a flexible alternative to either Protein A or Protein G alone. The particles can be used with the broadest range of antibody species and subclasses, which makes these particles well suited for antibody purification immunoprecipitations, and co-immunoprecipitation applications.
Sera-Mag SpeedBeads Protein A/G Magnetic Particles are uniform, nominal 1 μm diameter, colloidally stable, nonporous, superparamagnetic spheres made by a proprietary core-shell method.
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer (Fig 2). Two layers of magnetite are coated onto this core, resulting in faster magnetic response times, while the surface is modified with a proprietary method that minimizes non-specific binding of proteins. Protein A/G is then covalenty bound to the particle surface. To learn more about the Sera-Mag SpeedBeads Protein A/G Magnetic Particles, please review the application note “Sera-Mag SpeedBeads Magnetic Protein A/G Particles Applications Procedure” (code no. 29-1079-14) available at www.gelifesciences.com/sera-mag.
SpeedBeads are made by a modified Sera-Mag process
Fig 2. SpeedBeads are made by a modified Sera-Mag process in which a larger quantity of magnetite is added as a second layer within the particle, resulting in a 2×-faster increase in speed in response to magnetic field.
Specifications
Particle properties
Particle composition Double layer of magnetite
Surface Amine-modified with Protein A/G covalently bound to the surface
Density ~2.0 g/cm3
Magnetite content ~60%
Protein surface
Binding capacity 55-85 μg IgG bound per mg of particle
Number of binding sites Recombinant Protein A/G with 6 Fc binding sites
Isoelectric Point 4.65
Product attributes
Concentration 10 mg/ml
Magnetization Superparamagnetic (no magnetic memory)
Nominal diameter 1 μm
Additives 0.05% sodium azide
Package includes Certificate of Analysis/Packaging Insert
Storage conditions Unless otherwise stated, refrigerate (2 °C to 8 °C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with inversion by hand, roller or vortex mixer.
Materials
GE and GE monogram are trademarks of General Electric Company.
Sera-Mag is a trademark of General Electric Company or one of its subsidiaries.
Tween is a trademark of the Croda Group of Companies.
KingFisher is a trademark of Thermo Fisher Scientific LLC.
All other third party trademarks are the property of their respective owners.
© 2011–2014 General Electric Company - All rights reserved.
Related Links