[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Protein Expression](https://www.sigmaaldrich.com/US/en/applications/protein-biology/protein-expression)Simplicon™ *in vivo* Protein Expression System
# Simplicon™ *in vivo* Protein Expression System
## A non-genome integrating, tunable, and synthetic self-replicating RNA based expression system
Simplicon™ is a novel system that generates immediate, high-sustained protein expression of multiple genes into transfected human cells without the risk of genome integration. The technology employs a single, synthetic, polycistronic, self-replicating RNA based on the Venezuelan equine encephalitis (VEE) genome. The Simplicon™ RNA contains four genes (nsP1-4) encoding the VEE RNA replication machinery, whereas the structural proteins that are required to make an infectious particle have been removed and can be replaced with transgene(s) of interest (__Figure 1A__). The Simplicon™ RNA is a synthetic RNA generated from the Simplicon™ Cloning Vector (E3L) plasmid (Cat.#: [SCR724](https://www.sigmaaldrich.com/US/en/product/mm/scr724)). Up to a total of 8.3kb of transgene(s) has been successfully cloned in Simplicon™ Cloning Vector (E3L) for high sustained protein expression in BJ human foreskin fibroblasts. Simplicon™ RNA can be also introduced into LX2 human hepatic stellate cell line (Cat#: [SCC064](https://www.sigmaaldrich.com/US/en/product/mm/scc064)), human mesenchymal stem cells (MSCs, Cat#: [SCC034](https://www.sigmaaldrich.com/US/en/product/mm/scc034)), human iPSCs and human primary T cells (__Figure 4__).
Introduction and replication of the Simplicon™ RNA is expected to elicit a strong interferon (IFN) response in transfected cells. To suppress the IFN responses, a Vaccinia virus protein, B18R along with another Vaccinia virus protein, E3L, collaboratively suppresses the IFN responses in Simplicon™ RNA expression. B18R neutralizes type I interferons by direct binding, while E3L inhibits the cytoplasmic signaling pathways of IFN responses (__Figure 2__ and __3__, Cat #s: [SCR722](https://www.sigmaaldrich.com/US/en/product/mm/scr722) and [SCR727](https://www.sigmaaldrich.com/US/en/product/mm/scr727)).
One day after transfection of the Simplicon™ RNA, a spike in the levels of transgenes can be observed (__Figure 2__ and __3__). The expression levels are maintained by addition of [B18R-E3L](https://www.emdmillipore.com/US/en/product/B18R-E3L-Plasmid-human-codon-optimized-for-B18R-and-E3L,MM_NF-SCR727) and the selective agent, puromycin throughout the duration of the experiment. Over time, expression levels are expected to diminish and stabilize to 1/5 – 1/10 the levels initially observed and may be close to physiological levels after one week (__Figure 3__). Expression levels and duration may change depending upon the cell types, transgenes and media conditions used (__Figure 4__). Therefore, the fluorescence tag Simplicon RNAs (__Figure 2__ and __4__, Cat#: [SCR720](https://www.sigmaaldrich.com/US/en/product/mm/scr720), [SCR721](https://www.sigmaaldrich.com/US/en/product/mm/scr721), [SCR725](https://www.sigmaaldrich.com/US/en/product/mm/scr725) or [SCR726](https://www.sigmaaldrich.com/US/en/product/mm/scr726)) can be used to monitor and optimize the transfection of experiment Simplicon™ RNAs into the cell line of interest. For more sustained transgene(s) expression, recombinant B18R protein (Cat#: [GF197](https://www.sigmaaldrich.com/US/en/product/sigma/gf197) or [GF156](https://www.sigmaaldrich.com/US/en/product/mm/gf156)) or [B18R-E3L](https://www.emdmillipore.com/US/en/product/B18R-E3L-Plasmid-human-codon-optimized-for-B18R-and-E3L,MM_NF-SCR727) conditioned medium (B18R-CM) produced from produced from [B18R-E3L](https://www.emdmillipore.com/US/en/product/B18R-E3L-Plasmid-human-codon-optimized-for-B18R-and-E3L,MM_NF-SCR727) RNA can be used.
The Simplicon™ technology has been successfully utilized for efficient human iPSC generation through the sustained expression of critical reprogramming factors ( Simplicon™ technology utilized for efficient human iPSCs generation video) __Figure 5__ and __6__, Cat#s: [SCR549](https://www.sigmaaldrich.com/US/en/product/mm/scr549), [SCR550](https://www.sigmaaldrich.com/US/en/product/mm/scr550), [SCR703](https://www.sigmaaldrich.com/US/en/product/mm/scr703) and [SCR729](https://www.sigmaaldrich.com/US/en/product/mm/scr729)). Reprogramming using self-replicating RNA is more efficient and convenient than the synthetic mRNA-based method (Steinle, H et al. 2019). A key advantage of using self-replicating RNA is the cost about 24 times lower compared to daily transfection required for nucleoside-modified mRNA cocktail. Furthermore, using the self-replicating RNA, iPSCs were emerged and obtained earlier than after the mRNA transfection. The puromycin resistance in self-replicating RNA confers positive selection with puromycin during the early time point of reprogramming. Recently, Simplicon™ RNA Reprogramming Kit (OKSG) ([SCR550](https://www.sigmaaldrich.com/US/en/product/mm/scr550)) was used and cited in a Nature publication as a scientific report (Shiohira, C.M *et al*. 2018). In this publication, the self-replicating synthetic Simplicon™ RNA for Oct4, Klf4, Sox2 and Glis1 was transfected into mouse pancreatic tissue, resulting in efficient generation of induced tissue-specific stem cells from pancreas (iTS-P cells). These iTS-P cells carry genetic markers of endoderm and pancreatic pancreatic progenitors and differentiation into insulin-producing cells more efficiently than embryonic stem (ES) cells. It also has been applied to the creation of cell lines that express and retain the metabolic activities of five cytochrome P450 enzymes ([SCR724](https://www.sigmaaldrich.com/US/en/product/mm/scr724), [please see the poster](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/documents/simplicon-used-to-express-p450-enzymes-poster.pdf)). Please review the FAQs, Troubleshooting Tips and [Protocols](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/cloning-and-expression/simplicon-rna-transfection-electroporation-protocols) for more details.
__Figure 1. Structure of the Simplicon™ RNA.__The Simplicon™ RNA encodes four non-structural replication complex proteins (nsPs) as a single ORF at the 5´ end of the RNA. At the 3´ end, the viral structural proteins ORFs are replaced with the transgenes of interest. [Simplicon™ Cloning Vector (E3L)](https://www.sigmaaldrich.com/product/mm/scr724) (Cat #: [SCR724](https://www.sigmaaldrich.com/product/mm/scr724)) can accommodate transgene cassettes up to a total size of 8,300 bp by combination with 2A peptides and IRES. E3L is included to protect the resulting RNA from interferon responses caused by RNA replication. Locations of the 26S Subgenomic promoter, E3L, IRES and Puromycin (Puro)-resistance gene are indicated.
__Figure 2. RNA gels and TagGFP2 expression in HFFs.__(A) TagGFP2 and B18R-E3L RNA gels. Marker: 6, 4, 3, 2, 1.5, 1, 0.5, 0.2 kb. (B, C) TagGFP2 Simplicon™ RNA (E3L) (Cat. #: SCR720) and B18R-E3L RNA (Cat.#: SCR722) were co-transfected into HFFs with MessengerMax™ transfection Reagent. One day after transfection, GFP expression was analyzed by fluorescence microscopy and FACS.
__Figure 3. E3L increased Simplicon™ RNA expression levels.__(A) BJ human foreskin fibroblasts were co-transfected with Simplicon™ TagGFP2 (Cat. #: SCR713M) and B18R RNA (Cat. #: SCR723) or B18R-E3L RNA (Cat. #: SCR722). (B) E3L worked for continuous expression of Simplicon™ RNA. Simplicon™ TagRFP or TagRFP Simplicon (E3L) was co-transfected with B18R-E3L RNA and cultured with medium containing B18R protein and puromycin for 14 days. RFP expressing cells were imaged on Day 12, and analyzed by FACS on Day 14.
__Figure 4. Simplicon™ RNA can be transfected into a wide variety of cell types.__Simplicon™ TagGFP2 RNA (Cat. #: SCR713M) and B18R-RNA (Cat. #: SCR723) were co-transfected with Human iPSCs, LX2 human hepatic stellate cell line (Cat. #: SCC064), human mesenchymal stem cells (MSCs, Cat. #: SCC034) by MessengerMAX™ transfection reagent. For human primary T cells (PBMCs stimulated with CD3/CD28), electroporation method was used.
__Figure 5. Human iPS cells can be genereated with a single transfection of polycistronic RNA.__Once created, the reprogramming RNA can be selectively degraded by removing B18R from the culture media, creating transgene-free, replicon-free iPSCs.
__Figure 6. Time course of human iPSC colonies generated using Human Simplicon™ OKSG-cMyc TagRFP RNA__(Cat #: SCR714). BJ human foreskin fibroblasts were transfected with Simplicon™-OKSG-cMyc TagRFP and B18R RNAs One day after transfection (Day 2), approximately 37% cells are RFP positive (a, b) and TagRFP expression became strong on Day 5 (c). Cells were replated on Day 7. Colonies with TagRFP positive cells on Day 10 (d). At later time points, colonies may express variable levels of TagRFP, with some expressing partial TagRFP expression (e, h, i) or not at all (f, g, j). Distinct human iPSC colonies are observed by days 21 (h, i, j). Alkaline phosphatase staining of human iPSC colonies (k).
### Which Simplicon™ product is right for you?
Product NameCat. No.Application
__Simplicon™ Plasmids__
[Simplicon Cloning Vector (E3L)](https://www.sigmaaldrich.com/US/en/product/mm/scr724)[SCR724](https://www.sigmaaldrich.com/US/en/product/mm/scr724)To synthesize Simplicon™ RNA *in vitro* containing the transgene(s) of interest. At least a total of 8.3kb of transgene(s) can be synthesized into Simplicon™ RNA, followed by successful high sustained protein expression in transfected human cell lines, without the risk of genome integration.
[TagGFP2 Simplicon™ Plasmid (E3L)](https://www.sigmaaldrich.com/US/en/product/mm/scr725)[SCR725](https://www.sigmaaldrich.com/US/en/product/mm/scr725)To determine optimal transfection conditions to express the self-replicating RNA of your interest through using Simplicon Cloning Vector (E3L) (Part #: [SCR724](https://www.sigmaaldrich.com/US/en/product/mm/scr724)) in hard-to-transfect somatic or primary cells.
[TagRFP Simplicon™ Plasmid (E3L)](https://www.sigmaaldrich.com/US/en/product/mm/scr726)[SCR726](https://www.sigmaaldrich.com/US/en/product/mm/scr726)The TagRFP Simplicon™ Plasmid (E3L) was developed for the synthesis of TagRFP Simplicon™ RNA (E3L). The Simplicon™ TagRFP RNA may be used to determine optimal transfection conditions to express the self-replicating RNA in hard-to transfect somatic or primary cells.
[Human OKSG-cMyc TagRFP Simplicon™ Plasmid](https://www.sigmaaldrich.com/US/en/product/mm/scr729)[SCR729](https://www.sigmaaldrich.com/US/en/product/mm/scr729)To produce unlimited amount of Human OKSG-cMyc TagRFP Simplicon™ RNA for human iPSCs generation along with a red fluorescent protein (TagRFP).
__Simplicon™ RNAs__
Fluorescence Tag Simplicon™ RNAs
TagGFP2 Simplicon™ RNA (E3L) Kit[SCR720](https://www.sigmaaldrich.com/US/en/product/mm/scr720)To determine optimal transfection conditions to express the self-replicating RNA of your interest through using Simplicon™ Cloning Vector (E3L) (Part #:[SCR724](https://www.sigmaaldrich.com/US/en/product/mm/scr724)) in hard-to- transfect somatic or primary
TagRFP Simplicon™ RNA (E3L) Kit[SCR721](https://www.sigmaaldrich.com/US/en/product/mm/scr721)To monitor and optimize the transfection of experiment Simplicon™ RNAs into the human cell line of interest.
Human Simplicon™ Reprogramming RNAs
[Simplicon™ Reprogramming RNA (OKSG)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/stem-cell-culture/simplicon-rna-reprogramming)[SCR549](https://www.sigmaaldrich.com/US/en/product/mm/scr549)To generate integration free, virus-free human iPS cell using a single transfection step of self-replicating synthetic RNA (Simplicon™ RNA) containing Oct4, Klf4, Sox2, and Glis1.
[Simplicon™ RNA Reprogramming Kit (OKSG)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/stem-cell-culture/simplicon-rna-reprogramming)[SCR550](https://www.sigmaaldrich.com/US/en/product/mm/scr550)To generate integration free, virus-free human iPS cell using a single transfection step of self-replicating synthetic RNA (Simplicon™ RNA) containing Oct4, Klf4, Sox2, and Glis1. Kit includes Human Recombinant B18R Protein and Human iPS Reprogramming Boost Supplement II.
Human OKSG-cMyc Simplicon RNA[SCR703](https://www.sigmaaldrich.com/US/en/product/mm/scr703)To generate integration free, virus-free human iPS cell using a single transfection step of self-replicating synthetic RNA (Simplicon™ RNA) containing Oct4, Klf4, Sox2, Glis1 and cMyc. The 5-factor OKSG-cMyc Simplicon is especially useful for iPSCs generation from somatic cells that are more difficult to reprogram (i.e. slower proliferating cells or aged somatic cells).
Human OKSG-cMyc TagRFP Simplicon RNA[SCR714M](https://www.sigmaaldrich.com/US/en/product/mm/scr714m)The OKSG-cMyc transgenes are especially useful for iPSCs generation from somatic cells that are more difficult to reprogram (i.e. slower proliferating cells or aged somatic cells) while the TagRFP provides a rapid assessment of transfection efficiency. Presence of the TagRFP transgene also allows for optimization of the transfection conditions in hard- to- transfect somatic or primary cells.
__B18R Plasmids, RNAs and Proteins__
[B18R](https://www.emdmillipore.com/US/en/product/B18R-Plasmid-human-codon-optimized,MM_NF-SCR728) Plasmids
[B18R-E3L](https://www.sigmaaldrich.com/US/en/product/mm/scr727) Plasmid (human codon optimized for B18R and E3L)[SCR727](https://www.sigmaaldrich.com/US/en/product/mm/scr727)To strongly suppress more interferon responses than single B18R to enable more expression of a Simplicon™ RNA or regular mRNA(s).
[B18R Plasmid (human codon optimized)](https://www.sigmaaldrich.com/US/en/product/mm/scr728)[SCR728](https://www.sigmaaldrich.com/US/en/product/mm/scr728)To supply B18R RNA that suppresses IFN responses to support Simplicon™ RNA or normal mRNA expression.
B18R RNAs
B18R-E3L RNA (human codon optimized for B18R and E3L)[SCR722](https://www.sigmaaldrich.com/US/en/product/mm/scr722)To suppress the strong interferon (IFN) response elicited by the introduction and replication of the Simplicon™ RNA in the transfected human cells.
B18R RNA (human codon optimized)[SCR723](https://www.sigmaaldrich.com/US/en/product/mm/scr723)Required for high expression of Simplicon™ RNA at transfection and available for production of B18R-CM (conditioned medium).
B18R Proteins
Vaccinia Virus B18R protein, recombinant expressed in HEK 293 cells, Carrier-Free[GF197](https://www.sigmaaldrich.com/US/en/product/sigma/gf197)To suppress the strong interferon (IFN) response elicited by the introduction and replication of the Simplicon™ RNA in the transfected human cells and enable more sustained transgene(s) expression.
__Related Articles__
- [Residual Peroxide Determination in Food Packaging](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/extracellular-microenvironment)
- [Hypoxia Detection Assays](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/hypoxia-detection-assays)
- [3xFLAG® System Expression Vectors for Ultra-Sens](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/3x-flag)
- [FLAG® Tag Bacterial and Mammalian Expression Vectors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/vector-maps)
- [Cloning & Expression Vectors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/vector-systems)
- [p53: DNA Damage Response and Cancer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/epigenetics/p-fiftythree-dna-damage-response-and-cancer)
- [MISSION® shRNA Lentiviral Transduction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/gene-expression-and-silencing/mission-shrna-lentiviral-transduction)
- [Duolink® Proximity Ligation Assay Instructional Video](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-and-nucleic-acid-interactions/duolink-assay)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.