Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Reference Standards: Synthesis and Characterization of ±-threo-Ritalinic Acid-D10 Hydrochloride
By: Elizabeth B. Marek PhD, Huahua Jian PhD, Uma Sreenivasan PhD, Isil Dilek, PhD,
Abstract
As a major metabolite of methylphenidate (1, Ritalin), ±-threo-Ritalinic acid (2) is of clinical relevance. To this end ±-threo-ritalinic acid-D10 HCl (3) was synthesized in seven steps with a purity of 99% and an isotopic purity ratio of D0/D10 = 0% and a significant amount of the D9-D7 isomers. Because practical ion monitoring is based on the ratio of D0/D10, the standard was found to be suitable for use as an internal standard in LC-MS/MS analysis of ritalinic acid and related compounds. The presence of significant amounts of the D9 isomer prompted extensive structure elucidation work using 1D, 2D, and qNMR techniques.
metabolite of methylphenidate
Introduction
Methylphenidate, most commonly known by the Novartis trade name Ritalin®, is a pyschostimulant used to treat attention-deficit hyperactivity disorder, postural orthostatic tachycardia syndrome, and narcolepsy, by increasing alertness and attention and by counteracting fatigue. Methylphenidate was originally sold as a mixture of diastereomers, although it has been shown that the majority of the activity is attributed to the ±-threo isomer. More recent products such as Focalin® contain only the active ±-threo isomer. While analytical reference standards of the diastereomeric mixture of methylphenidate and its metabolites are available, standards containing only the active isomer are now desirable to reflect the current directive of using only the active isomer in drug products. Therefore it is also desirable to synthesize stable-labeled derivatives of the active isomers of methylphenidate and its metabolites, such as ritalinic acid, for use as internal standards.
Ritalinic acid is a major metabolite of and synthetic precursor to methylphenidate and may be monitored clinically and forensically. The synthesis of deuterated ±-threo-ritalinic acid was therefore undertaken to develop an analytical reference standard and as a precursor to deuterated ±-threo-methylphenidate.
Synthesis of ±-threo-Ritalinic acid-D10 HCl
Based on literature precedence 1-3, ±-threo-Ritalinic acid-D10 HCl was synthesized in seven steps from pyridine-D5. During the synthesis, the crucial reduction of the pyridine moiety to the fully deuterated piperidine proceeded in good yield but LC/MS SIM indicated that product contained a mixture of 55% D10, 34% D9 and 11% D8-D7. The presence of the D0 isomer was not detected. This deuterium ratio was carried through to the final product.
threo-Ritalinic acid-D10
Attempt to enrich the deuterium content of ±-threo-Ritalinic acid-D10 HCl
deuterium content
Further treatment of ±-threo-ritalinic acid-D10 HCl with Pt/C, D2, and AcOD did not result in an enrichment of the deuterium content. The isotopic distribution was fully characterized to determine suitability for use.
Characterization of ±-threo-Ritalinic acid-D10 HCl
Isotopic distribution by LC/MS-SIM  
Deuterium Content Percent
(xi)
D10 55.16%
D9 33.98%
D8 9.21%
D7 1.48%
D6 0.148%
D5 0.0128%
D4 0.0010%
D3 0.0%
D2 0.0%
D1 0.0%
D0 0.0%
The identity of ±-threo-Ritalinic acid-D10 HCl was established through NMR and mass spectrometry. The chemical purity was established through HPLC/UV, Karl GC/FID Headspace and ROI. LC- MS/MS studies were performed to evaluate isotopic purity, deuterium distribution, fragmentation patterns and suitability for use as an internal standard. HPLC analysis indicated a purity of 99% with isotopic purity ratio D0/D10 = 0% by LC/MS-SIM. Additionally, LC/MS-SIM confirmed the presence of 45% D9-D7 isomers (see isotopic distribution at left).
Finally, NMR techniques including 1H, 13C, COSY, HSQC, and qNMR were used to determine the placement of the deuterium on the molecule.
Determination of deuterium placement using NMR techniques
1H NMR
Analysis of LC/MS-SIM, 1H NMR, and 13C NMR
LC/MS-SIM clearly shows incomplete deuterium exchange with only 55% D10. Additionally, the data suggested that some proton to deuterium exchange had also occurred on the aromatic ring. It was therefore desirable to determine the placement of the deuterium labels. 1H NMR clearly shows the presence of a roughly even distribution of protons on the piperidine moiety. Unfortunately, the lack of a reference peak in the 1H NMR made it impossible to determine the extent of the distribution of deuterium on this ring or whether the exchange had also occurred on the aromatic ring. Therefore, Quantitative NMR was used to determine the percent deuterium on each carbon in the molecule.
Isotopic distribution by quantitative NMR
Using maleic acid as an internal standard, Quantitative NMR (qNMR) was used to determine the percentage of hydrogen and therefore deuterium on each carbon of ±-threo-Ritalinic acid-D10 HCl.
Ritalinic acid-D10
Isotopic
H% = % hydrogen
I = integer of signal
n = number of protons under the signal of interest
m = mass of analyte (a)
M = molecular weight
std = internal standard
A or a = component or analyte whose purity is to be calculated
P = purity
Product
Internal Standard
±-threo-Ritalinic acid-D10
Maleic acid
Mass balance Istd = 2000 Ma = 265.223
Purity Pa= 95.51% Mstd = 116.07
  mstd= 4.029 ma= 15.552
  nstd = 2 Pstd = 99.78%
Position nA IA H% D%
Ortho 2 3122 0571 96 540 3.460
Para&meta 3 4708.465 97.063 2.937
Aromatic 5 7830.522 96.854 3.146
2 1 61.743 3.818 96.182
3 1 130.420 8.066 91.934
4 2 158.628 4.905 95.095
5 2 264.270 8.172 91.828
6 2 185.778 5.745 94.255
7 2 208.357 6.443 93.557
The molar mass of the product is calculated based on isotopic purity results obtained by LC/MS-SIM.
maleic-acid
isotopic-distribution
qNMR confirmed the presence of a nearly even distribution of deuterium throughout the piperidine moiety and also revealed that a small percentage of the hydrogens on the aromatic moiety had exchanged for deuterium as well. These results suggest that the Pt/C catalyst used in step 5 of the synthesis also facilitated the exchange of some of the aromatic hydrogens deuterium.
Conclusions
±-threo-Ritalinic acid-D10 HCl was synthesized in good yield, sufficient ratio of D0/D10, and 99% purity making it an extremely useful internal standard for the quantitation of ritalinic acid. LC/MS-SIM indicated significant amounts of D9-D7 with no D0. 1H and 13C NMR spectra could not provide definitive information as to where the deuterium were placed. qNMR was, therefore, used to determine exactly what percentage of deuterium is present on each carbon in the molecule. This data indicated that the reduction in step 5 using Pt/C resulted in incomplete deuterium exchange and even caused the exchange of a small percentage of the aromatic protons to deuterium. This study highlights chemical interactions that factor into the design of stable labeled internal standards for LCMS applications.
Materials
References
  1. J. Heterocyclic Chem. 44; 2007; 1485.
  2. US Patent 5936091.
  3. J. Med. Chem. 39; 6; 1996; 1201.
Related Links