Ultrafree®-MC and Ultrafree®-CL Centrifugal Filters for Microfiltration
Spin filters for clarification and filtration
Ultrafree®-MC and Ultrafree®-CL Centrifugal Filters for MicrofiltrationUltrafree®-MC and Ultrafree®-CL centrifugal filters remove particles and precipitates from aqueous and some solvent-based samples. These fast filtration units, containing high-recovery Durapore® (PVDF) or hydrophilic PTFE membranes, provide fast, reproducible performance. Ultrafree® centrifugal filters are ideal for clarifying protein and nucleic acid solutions.
Ultrafree®-MC and CL centrifugal filter advantages:
  • High recovery Durapore® (PVDF) and hydrophilic PTFE membranes
  • Five different pore sizes from 0.1 to 5.0 μm
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 mL tubes       
Sterile Ultrafree®-MC and CL centrifugal filter units:
  • Easy, pre-sterilized, centrifugal sample clarification units for either 0.5 mL (MC) or 2 mL (CL) maximum volumes
  • High recovery Durapore® (PVDF) membrane
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 mL tubes (MC) or 15 mL tubes (CL)    
Choose the Ultrafree® Filter that suits your needs:
Filter Units with Microporous Durapore® PVDF Membrane
Catalog Number Description Pore Size Color Code Sterility Pack Size Add to Cart
UFC30DV00 Ultrafree-MC DV Centrifugal Filter 0.65 µm Purple Non-Sterile 100
UFC30DV0S Ultrafree-MC DV Centrifugal Filter 0.65 µm Purple Sterile 50 (5x10)
UFC30DV25 Ultrafree-MC DV Centrifugal Filter 0.65 µm Purple Non-Sterile 25
UFC30GV00 Ultrafree-MC GV Centrifugal Filter 0.22 µm Yellow Non-Sterile 100
UFC30GV0S Ultrafree-MC GV Centrifugal Filter 0.22 µm Yellow Sterile 50 (5x10)
UFC30GV25 Ultrafree-MC GV Centrifugal Filter 0.22 µm Yellow Non-Sterile 25
UFC30GVNB Ultrafree-MC GV Centrifugal Filter 0.22 µm Yellow Non-Sterile 250
UFC30HV00 Ultrafree-MC HV Centrifugal Filter 0.45 µm Red Non-Sterile 100
UFC30HV25 Ultrafree-MC HV Centrifugal Filter 0.45 µm Red Non-Sterile 25
UFC30HVNB Ultrafree-MC HV Centrifugal Filter 0.45 µm Red Non-Sterile 250
UFC30SV00 Ultrafree-MC SV Centrifugal Filter 5.0 µm Dark green Non-Sterile 100
UFC30VV00 Ultrafree-MC VV Centrifugal Filter 0.1 µm Orange Non-Sterile 100
UFC30VV25 Ultrafree-MC VV Centrifugal Filter 0.1 µm Orange Non-Sterile 25
UFC40DV25 Ultrafree-CL DV Centrifugal Filter 0.65 µm Purple Non-Sterile 25
UFC40GV00 Ultrafree-CL GV Centrifugal Filter 0.22 µm Yellow Non-Sterile 100
UFC40GV0S Ultrafree-CL GV Centrifugal Filter 0.22 µm Yellow Sterile 50 (5x10)
UFC40GV25 Ultrafree-CL GV Centrifugal Filter 0.22 µm Yellow Non-Sterile 25
UFC40HV00 Ultrafree-CL HV Centrifugal Filter 0.45 µm Red Non-Sterile 100
UFC40HV25 Ultrafree-CL HV Centrifugal Filter 0.45 µm Red Non-Sterile 25
UFC40SV25 Ultrafree-CL SV Centrifugal Filter 5.0 µm Dark green Non-Sterile 25
UFC40VV00 Ultrafree-CL VV Centrifugal Filter 0.1 µm Orange Non-Sterile 100
UFC40VV25 Ultrafree-CL VV Centrifugal Filter 0.1 µm Orange Non-Sterile 25
Filter Units with Microporous Hydrophilic PTFE Membrane
Catalog Number Description Pore Size Color Code Sterility Pack Size Add to Cart
UFC30LG25 Ultrafree-MC LG Centrifugal Filter 0.2 µm Yellow Non-Sterile 25
UFC30LH25 Ultrafree-MC LH Centrifugal Filter 0.45 µm Red Non-Sterile 25
UFC40LG25 Ultrafree-CL LG Centrifugal Filter 0.22 µm Yellow Non-Sterile 25
UFC40LH25 Ultrafree-CL LH Centrifugal Filter 0.45 µm Red Non-Sterile 25