Application Note: Ultrapure Water for Determination of Elemental Impurities per USP
By: Anastasia Domanova1, Juhani Virkanen2, Glenn Woods3, Stephane Mabic1, 1. MilliporeSigma, Lab Water, Saint-Quentin-en-Yvelines, France 2. University of Helsinki, Helsinki, Finland 3. Agilent Technologies Ltd, Stockport, UK
This paper demonstrates the suitability of fresh ultrapure water produced using Milli-Q® water purification systems
for analyses of elemental impurities according to General Chapters USP <231>; <232>; and <233>.
Introduction and Water Quality Requirements
In the pharmaceutical industry, it is crucial to monitor and control inorganic impurities in pharmaceuticals because some metals are used as reagents and catalysts during production and formulation processes. Moreover, metal impurities can be introduced into pharmaceutical products non-intentionally through contaminated reagents, or when products are in contact with pharmaceutical packaging or with metal surfaces during the development process. This is why the United States Food and Drug Administration (FDA) and similar international health agencies have had long-standing regulations in place for controlling harmful impurities in pharmaceutical products marketed for human consumption. Historically, four heavy metals, or the “Big Four”, were required to be tested according to the United States Pharmacopeial Convention (USP) General Chapter <231> “Heavy Metal Limit Test”. However, new mandatory guidelines to control potential toxic impurities in drugs were recently established in two USP General Chapters <232>1 and <233>.2
These new Elemental Impurities chapters in the USP subdivide the metals desired for analyses into several groups. In this application note, the first and the second groups comprising 15 metals (cadmium, lead, arsenic, mercury, iridium, osmium, palladium, platinum, rhodium, ruthenium, chromium, molybdenum, nickel, vanadium and copper) are addressed. Other elemental impurities listed in the FDA’s Guidance for Industry to Q3D Elemental Impurities3 and in the European Medicine Agency’s (EMA) Guideline on the Specification Limits for Residues of Metal Catalysts or Metal Reagents4 are addressed in an extended ICP-MS application note for pharma.
Different types of uses for ultrapure water in ICP-MS analysis
Figure 1. Different types of uses for ultrapure water in ICP-MS analysis.
Inductively coupled plasma–mass spectrometry (ICP-MS) is a sensitive technique for analyses of elemental impurities.2 Ultrapure water is a main reagent in ICP-MS and is used extensively from sample preparation to analytical instrument cleaning (Figure 1). In the pharmaceutical industry, the choice of water quality is dictated by its intended use. Milli-Q® ultrapure water purification systems are designed to be compliant* with water quality standards described in various pharmacopeias. However, water selected as a reagent must not only comply with specific pharmacopeial standards, but must also meet the requirements of modern instrumentation to ensure the best performance of ICP-MS instrumentation.5 Thus, reagent water of very high quality must be selected to avoid sample and analytical instrument contamination.6 The aim of this study was to evaluate the suitability of fresh ultrapure water produced using Milli-Q® ultrapure water purification systems for use in ICP-MS trace element analyses.
Results and Discussion
In the pharmaceutical industry, element analyses are performed in the range of mg/L (ppm) to sub-μg/L
(sub-ppb) levels. As sensitivity, accuracy, precision and recovery must be appropriately demonstrated during
the method validation process, achieving a low and stable background equivalent concentration (BEC) as well as
low and consistent detection limits is critical. Table 1 presents the BEC and limit of detection (LOD) in Milli-Q®
ultrapure water
for each element in ng/L (ppt) level.
Element BEC (ppt) LOD (ppt)
51V 0.72 0.39
52Cr 2.10 0.37
58Ni 0.76 0.14
63Cu 0.19 0.18
75As 3.10 0.11
95Mo 0.62 0.72
101Ru 0.69 0.36
103Rh 0.16 0.19
105Pd 0.07 0.26
111Cd 0.06 0.06
189Os 0.28 0.20
193Ir 0.08 0.12
195Pt 0.29 0.19
202Hg 0.49 0.25
208Pb 1.37 0.33
Table 1. The levels in ng/L (ppt) of elemental impurities in freshly produced ultrapure water from a Milli-Q® system measured under normal laboratory conditions (not in a clean room).
* Compliance reports are available on demand.
From Table 1, it can be observed that certain elements are present in slightly higher levels than sub-ppt. This can be explained by contamination coming from the laboratory environment, since the present analyses were performed under normal laboratory conditions.7 In case there is a need to achieve significantly lower levels of elements, it is reasonable to apply an additional polishing step, such as a Q-POD® Element unit which makes it possible to obtain BECs at sub-ppt and ppq levels in a clean room or metal-free laboratory environment.8,9
Ultrapure water samples from a MilliporeSigma Milli-Q® Advantage A10 water purification system, equipped with Q-Gard® and Quantum® TEX cartridges, Millipak® final filter, and fed by an Elix® Essential 5 water purificationsystem, were analyzed for the levels of V, Cr, Ni, Cu, As, Mo, Cd, Pd and Pb using an Agilent® 7700s ICP-MSinstrument, and for the levels of Ru, Rh, Os, Ir and Pt using an Agilent® 7500s ICP-MS instrument. Ultrapure watersamples from a MilliporeSigma Milli-Q® Direct water purification system, equipped with Q-Pak® TIX cartridge and Millipak® final filter, were analyzed for Hg levels using an Agilent® 7500s ICP-MS instrument. All experiments wereperformed under regular laboratory conditions (not in a clean room).
Agilent® 7700s instrumental details and parameters: PFA-50 nebulizer, PFA spray chamber, sapphire inerttorch, quartz 2.5 mm i.d. torch injector, platinum sample and skimmer cone, RF power 600/1600 W, samplingposition 12 / 8 mm, carrier gas flow 0.90 L/min, makeup gas flow 0.32 / 0.51 L/min, auto detector mode,calibration through 1, 5, 10, 50 ng/L.
Agilent® 7500s instrumental details and parameters: quartz nebulizer, quartz spray chamber, quartz i.d. torchinjector, nickel sample and skimmer cone, RF power 1300/1550 W, sampling position 8 mm, carrier gas flow0.96 L/min, makeup gas flow 0.23 L/min, auto detector mode, calibration through 1, 20, 50, 100 ng/L.
The calibration standards used in experiments with the Agilent® 7700s were a mixture of Agilent® and SPEXCertiPrep®, and with the Agilent® 7500s, ROMIL PrimAg®-xtra was used. Containers were all PFA pre-cleaned withultrapure water. All ultrapure water samples (resistivity of 18.2 MΩ·cm and TOC below 5 ppb) from Milli-Q® waterpurification systems were analyzed immediately after water collection.
Low levels of elemental impurities in ultrapure water produced by Milli-Q® ultrapure water purification systems were demonstrated. Laboratories in the pharmaceutical industry performing trace element analyses can rely on Milli-Q® water purification systems to meet their stringent requirements for the highest purity water for their experiments. Choosing ultrapure water from Milli-Q® water purification systems for element analyses will help to ensure the generation of high quality data.
  1. USP, Chapter 232, Elemental impurities – Limits.
  2. USP, Chapter 233, Elemental impurities – Procedures (2013).
  3. FDA. Q3D Elemental Impurities: Guidance for Industry (2015).
  4. EMA. Guideline on the specification limits for residues of metal catalysts or metal reagents (2008).
  5. Lewen N. The use of atomic spectroscopy in the pharmaceutical industry for the determination of trace elements in pharmaceuticals. J Pharm Biomed Anal 2011;55:653-61.
  6. USP, Chapter 1231, Water for Pharmaceutical Purposes.
  7. Rodushkin I, Engstrom E, Baxter DC. Sources of contamination and remedial strategies in the multi-elemental trace analysis laboratory. Anal Bioanal Chem 2010:396:365-77.
  8. Ishii N, Mabic S. Optimal water quality for trace elemental analysis. MilliporeSigma, Lab Water Application Note (2011).
  9. Khvataeva-Domanov A, Woods G, Mabic S. Poster: Choosing optimal high purity water source in accordance to ICP-MS application needs and
    laboratory environment. Winter Conference on Plasma Spectrochemistry, 2014 (Amelia Island, Florida, USA).