Milli-Q® HX 7000 Series of High-Throughput Water Purification Systems
Connected, sustainable central pure water solutions for up to
9000 L daily
At the heart of your total pure water solution, the Milli-Q® HX 7000 system serves as the nerve center for the entire water purification chain, and provides total control of all functions, operating parameters and standard accessory components.
Milli-Q® HX 7040/7080/7120/7150 systems comprise several water purification technologies, including complete pretreatment; advanced reverse osmosis (RO) and Evolutive Reject Adjustment (E.R.A.™) water usage and quality optimization; Elix® electrodeionization (EDI) technology; and treatment by bactericidal UV lamp.
The combination of these complementary purification technologies ensures the production of optimum constant-quality analytical-grade water.
Features & Benefits
  • Power – Milli-Q® HX 7000 systems are designed to deliver the highest pure water quality for your regular laboratory applications and instrument feed — up to 9000 L daily, with a range of systems providing flow rates of 40, 80, 120 or 150 L/h. 
  • Communication – In line with the evolution in today's labs, Milli-Q® HX 7000 systems offer users flexible monitoring with connectivity at the heart of their water purification system. Authorized users can access system data via computer, tablet or smartphone 24/7.
  • R&D Excellence – Cutting-edge, complementary water purification technologies make it all possible. Full Progard® pretreatment, advanced reverse osmosis, E.R.A.™ (Evolutive Reject Adjustment) technology, and patented Elix® module electrodeionization produce the best pure water — efficiently, economically and sustainably.
  • Ease of Use – With Milli-Q® HX 7000 systems, you can see all important information on the large color touch screen — water production, storage, dispensing, consumable use, alarms/alerts — and easy, step-by-step instructions from a wizard whenever you need to change a consumable.
Water Applications
Type 2 Water Applications
It is suitable for the following applications:
  • Microbiological media preparation
  • Buffer preparation
  • Hydroponics
  • Manufacturing chemical and biochemical reagents
  • Pharmaceutical laboratories
Feed to laboratory equipment: point-of-use ultrapure "polishing" systems (Milli-Q® IQ 7000 systems) washing machines, clinical analyzers, stability chambers, humidifiers, autoclaves, weathering test equipment, hydrogen gas generators, high-throughput ultrapure water purification systems (Super-Q® systems)
Milli-Q® HX 7000 Systems Performance
Parameter Value or Range
Resistivity > 5 MΩ.cm @ 25 °C (10-15 MΩ.cm @ 25 °C typically)
Conductivity < 0.2 µS/cm @ 25 °C
Total Organic Carbon (TOC) Typically < 30 ppb
Microorganisms < 10 CFU/mL
Dissolved silica < 3 ppb (rejection > 99.9 %)