Milli-Q® Integral for Water Purification
Type 1 and Type 2 Water Purification from Tap Water
Milli-Q® Integral is a water purification system providing labs with pure and ultrapure water, directly from tap water. The system has the capacity to produce up to 300 L/day of pure and/or ultrapure water, satisfying the needs of most labs. Water is conveniently delivered respectively through the E-POD® and Q-POD® remote dispensers.
Features and Benefits
  • Milli-Q® Integral: high quality type 2 and type 1 purified water in an optimized lab space.
  • Thanks to the combination of technologies inside, only one water purification unit can be connected to tap, and meet the scientists’ requirements in terms of water purity reliability and consistency, in terms of ergonomics thanks to the POD®s remote dispensers, validation and traceability
The system is delivered with calibration certificates for all monitors, and full validation support is available to satisfy the most stringent regulatory demands
The Milli-Q® Integral water purification system integrates patented technologies - Elix® Electrodeionization modules and TOC monitor meeting the demands of USP and EP regulations.
Water Applications
Ultrapure Water Applications
Milli-Q Integral Type 1 and Type 2 Water Purification System
  • Chromatography: HPLC, UPLC, LC-MS and GC
  • Elemental analysis: AA, GFAA, ICP-MS, ICP-OES and ILC
  • Cell Culture
  • Molecular biology and biochemistry PCR, DNA sequencing, DNA microarray, electrophoresis and blotting
Pure Water Applications
  • Analytical techniques with sensitivity at ppm level or above such as pH measure or Kjeldahl test
  • Chemical synthesis: reagents preparation
  • Staining solutions preparation for histology
  • Instrument feed: feed water for clinical analyzers, weatherometers, glassware washing machines, stainless steel autoclaves, stability test chambers, hydrogen generators
Water Specifications
  Milli-Q® Integral ultrapure water (Type 1) Milli-Q® Integral ultrapure water (Type 2)
Resistivity at 25 °C1
18.2 MΩ•cm
5-15 MΩ•cm
TOC2 ≤ 5 ppb < 30 ppb
Particulates (size > 0.22 μm)3 < 1 particulate/mL < 1 particulate/mL
Bacteria3,4 < 0.01 CFU/mL < 0.1 CFU/mL
Pyrogens (endotoxins)4 < 0.001 EU/mL < 0.001 EU/mL
RNases4 < 1 pg/mL < 0.01 ng/mL
DNases4 < 5 pg/mL < 4 pg/µL
Flow Rate Up to 2 L/min Up to 2 L/min
1 resistivity can also be displayed non-temperature-compensated as required by USP
2 in the appropriate test conditions
3 with Millipak® final filter
4 with Biopak® polisher
These values are typical and may vary depending on the nature and concentration of contaminants in the feed water.
If you have a question about the Milli-Q® Integral water purification system, or if you would like to request a quote, please click the button below.