Milli-Q® Ultrapure Water Solutions
Find the perfect Type 1 water purification system for your lab
Type 1 water purification system with Q-POD dispenser and touchscreenYou now have more choice of Milli-Q® ultrapure (Type 1) systems to perfectly satisfy your laboratory needs—and constraints.
  • Choose from our state-of-the-art Milli-Q® IQ 7000 system that offers a full Qualification program, advanced data traceability, and accurate TOC monitoring. Download the brochure
  • Or discover our newest Milli-Q® EQ 7000 system with its intuitive touchscreen, agile Q-POD® dispensing, and space-saving installation options. This system offers dedicated innovations for flexibility to adapt to your lab’s constraints. Download the brochure
Video: Milli-Q® IQ 7000 water purification system
See how the Milli-Q® IQ 7000 system can change the way you work in the lab.
Invest in Lab Efficiency
See more Milli-Q® IQ 7000 videos.  
Video: Milli-Q® EQ 7000 water purification system
The authentic Milli-Q® ultrapure experience, designed for modern-day heroes.
Flexibility that Adapts to Your Lab
Confidence in high-quality Milli-Q® ultrapure water
Type 1 water purification system with Q-POD dispenser and touchscreen interfaceWith a Milli-Q® ultrapure water system, you’re sure that every dispense is a high-quality dispense. Both Milli-Q® IQ and EQ 7000 systems contain the same powerful combination of purification media to provide ultrapure water quality consistently to the trace level. You can fully rely on either system's water purity for your critical experiments.
Two systems. One authentic Milli-Q® experience.
  • Easy and agile Q-POD® dispensing. Dispensing ultrapure water is easy, precise and effortless with the “thumb wheel”. Access a range of dispensing functions that are useful for day-to-day work in the laboratory.
  • Intuitive touchscreen control. Go anywhere in just a few clicks. The interactive screen simplifies system use and data access.
  • Space-saving design. Only interact with your Q-POD® dispenser and touchscreen — the compact system can easily be integrated in the lab environment.
  • Improved sustainability. These systems have a 25% smaller system footprint and contain 33% smaller purification cartridges vs. our previous generation systems. Plus, Lab Close mode saves energy when you are away from the lab for extended periods.
Compare Milli-Q® IQ and Milli-Q® EQ 7000 system features
Review the comparison table below to understand the similarities and differences between Milli-Q® IQ 7000 and Milli-Q® EQ 7000 ultrapure water systems.
Features Milli-Q® EQ 7000 Milli-Q® IQ 7000
Water quality Type 1 ultrapure water
Feed water quality Pretreated*
IPAK Meta® & IPAK Quanta® polishing packs Yes
Application POD-Paks 6
Touchscreen interface Yes
Lab Close mode Yes
Network connectivity and USB port Yes
System footprint ++++
Flexibility of installation +++ ++++
Flexibility of Q-POD® dispenser +++ ++++
Number of Q-POD® dispensers 1 Up to 4
Dispensing flow rates 3 8
Volumetric dispensing ++ ++++
Drop-by-drop dispensing No Yes
Assisted dispensing No Yes
Data management ++ ++++
Dispense reports & QR codes No Yes
Oxidative UV lamp Mercury Mercury-free
TOC accuracy In-line Indicator A10® Monitor
Compatible with Milli-Q® IQ Element unit No Yes
Qualification No Yes
TOC calibration/verification No Yes
USP suitability testing No Yes
*By electrodeionization (EDI), deionization (DI), reverse osmosis (RO), or distillation technologies.
†Only temperature and conductivity cell calibration.
Select the Type 1 water system that works for you
The selection of the proper ultrapure water system for your laboratory will depend on several parameters, such as:
  • Available feed water
  • Daily volume needs
  • Monitoring requirements
  • Expected certification levels
Contact our Application Specialists to discuss the ultrapure water system that’s best adapted to your needs.