SDS 500 Pure Water Storage Tank
Large volume storage, protection and distribution system
A key component of our total pure water solution, the SDS 500 storage, protection and distribution system is designed to store up to 460 liters of purified water, provide effective protection against airborne contamination, and to distribute the purified water under pressure to multiple locations.
Together with the Milli-Q® HX 7000 or Milli-Q® HR 7000 series of high-throughput water purification systems, SDS 500 storage tanks can be combined in compact and modular turnkey installations to meet the needs of one or several laboratories — or even an entire building.
Features & Benefits
  • Standard turnkey solution
    • All-in-one solution with main loop options and consumables available as part of the configuration
    • Flexible and adaptable: pump(s), inverter(s) and flow controllers adapted to a broad range of applications; distribution flow up to 40 L/min
    • Certified equipment backed by our company's stringent design and Quality standards
  • Constant pure water quality
    • State-of-the-art tank design: cylindrical shape with conical bottom for best drainage; optimal polyethylene tank and IR Polypropylene piping construction materials to ensure high-quality storage/distribution
    • Protection of pure water quality: vent filter protects against airborne contaminants; optional filters and bactericidal UV lamps available
    • Constant velocity maintained in the loop ensuring a turbulent flow
    • Programmable automatic flush through the Milli-Q® HX or HR 7000 system MMI, to maintain water quality during weekends or other long non-use periods.
  • Ease of use
    • Preventive maintenance and consumables’ lifetime management with alerts and alarms on the
      Milli-Q® HX or HR 7000 screen.
    • Ergonomic design to ensure easy access to consumables / accessories, and rapid replacement of consumables; sanitary sampling port(s).
  • Connectivity
    • Remote control provided by the Milli-Q® HX or HR 7000 system: easy access to SDS 500 key data (tank level measurements, preventive alerts, and consumables status) via computer, tablet, or smart phone
    • Real-time data with on-line measurements
    • Connectable to a Building Management System (BMS) or Laboratory Information Management System (LIMS) via Milli-Q® HX or HR 7000 series system
Water Applications
Type 2 Water Applications
It is suitable for the following applications:
  • Microbiological media preparation
  • Buffer preparation
  • Hydroponics
  • Manufacturing chemical and biochemical reagents
  • Pharmaceutical laboratories
Feed to laboratory equipment: point-of-use ultrapure "polishing" systems (Milli-Q® IQ 7000) washing machines, clinical analyzers, stability chambers, humidifiers, autoclaves, weathering test equipment, hydrogen gas generators, high-throughput ultrapure water purification systems (Super-Q® systems)