Whatman™ Filter papers for use in the Beer Industry
Quality control (QC) methods used by the brewing industry are published by a number of national and international bodies such as the European Brewery Convention (EBC), Mitteleuropäische Brautechnische Analysenkommission (MEBAK), and the American Society of Brewing Chemists (ASBC). Many of these QC tests involve a filtration step for sample preparation. In this study, three grades of Whatman filter paper (Grades 2V, 597½, 2555½) were evaluated in analyses representing different steps in the brewing process. These tests, from ASBC methods, were 1) removal of turbidity from wort, 2) removal of CO2 from finished beer, and 3) removal of yeast cells after fermentation. The data presented here demonstrate that all three filter grades are suitable for use in the filtration steps that these methods require.
Introduction
Growing competitiveness in the global market for beer has led brewers and standard-setting agencies for regional varietals to increase focus on product consistency for characteristics such as color, alcoholic strength, flavor, and clarity.
Several national and international bodies, notably the EBC, MEBAK, and ASBC, publish analytical methods to guide beer quality testing. Many of these require filtration to prepare the sample for testing. Some methods rely on the traditional concept of filtration, wherein the filter’s sole purpose is to remove particulate matter from a liquid sample prior to analysis of the clarified liquid (i.e., the filtrate). Other methods require testing of the retained solids (i.e., the retentate). See Table 1 for examples of these tests.
This application note shows how Whatman filters of three grades performed in supporting three ASBC methods of importance to the overall QC process. These tests were 1) removal of turbidity from wort (Malt-4), 2) removal of CO2 from finished beer for quality control purposes (Beer-1,D), and 3) removal of yeast cells after fermentation (Beer-8).
Table 1. MEBAK analytical methods that use filtration. Numbers refer to the designation within the MEBAK handbook1
1.4.3.1 Soluble extract in wet spent grains obtained by pressing (rapid method)
1.4.3.2 Soluble extract in wet and dry spent grains obtained by rinsing (EBC)
1.4.4.2 Available residual extract
1.4.5 Iodine value of brewery spent grains
1.6.1 Solids in wort (Labor veritas method)
1.6.2 Solids or trub material (Field method)
1.6.3 Cold trub
2.6.2 Coaguble nitrogen (thermal coagulation of protein)
2.6.3.1 Nitrogen fractionation (precipitation with magnesium sulfate)
2.6.3.2 Nitrogen fractionation (precipitation with phosphomolybdic acid)
2.8.1 Limit of attenuation in wort (fermentation tube method)
2.8.2 Limit of attenuation in wort (reference method – EBC)
2.8.3 Limit of attenuation in wort (rapid method – EBC)
2.9.1 Degassing a sample (EBC)
2.10.3.2.1 Total glucose – hydrolysis method
2.12.2 Spectrophotometric color (EBC)
2.14.2.2 Alcohol chill haze test, CHAPON (cold sensitivity)
2.16.3 Tannoids
2.17.3 Determination of hop bitter substances in wort and beer (EBC)
2.20.1 Membrane filterability test of beer
2.21.3.3 4-vinyl guaiacol and 4-vinyl phenol detection
2.21.8.3 Detection of SO2 with continuous flow rate
2.22.1 Chloride, sulfate, nitrate and phosphate in beer (EBC)
2.22.5 Sulfate ions
Wort filtration
The quality of a wort batch is dependent on the quality of the malt used to produce it. Measurement of wort filtration speed and turbidity allows a brewer to qualify whether the malt used to produce it is leading to a desirable end product and to quantify a tolerance range for quality control of end product consistency. A slower filtration speed reflects a lower wort solubility. Measuring the filtration speed also helps the brewer understand the malt’s contribution to fermentable extract, pH, color, viscosity, and nitrogen content2. A wort sample with low turbidity is required to support later photometric testing. Gravity filtration using an appropriate filter paper supports all of these objectives.
CO2 removal prior to further testing
During fermentation, CO2 is produced and dissolves into the solution. When a QC lab prepares a sample of this solution for analysis, this dissolved CO2 may lead to inaccurate results in tests such as total acid determination. Therefore, the CO2 content needs to be minimized prior to testing. One method for achieving this is to pass the beer sample through an appropriate paper filter using gravity filtration.
Removal of yeast cells after fermentation
Measuring the total acidity of the final brewed product requires that the remaining yeast cells are removed. This simple separation of particulate matter from liquid sample can be achieved through gravity filtration.
Methods
In this study Whatman Grades 2V, 597½, and 2555½ from Cytiva’s Life Sciences business were evaluated for their suitability in the three tests described below. All filters were 320 mm in diameter and prepleated with 16 pleats (Fig 1).
prepleated Whatman filter paper
Fig 1. A prepleated Whatman filter paper.
All tests were performed according to the ASBC methods presented in Table 2. All testing was performed by the Biotechnology School at Jiangnan University, No 1800 Lihu Avenue, Wuxi, Jiangsu, 214122, China.
Table 2. Study design
Whatman filter Product code Lot number  Tests (ASBC Method)
Grade 2V 1202-320 G8605164 Wort filtration (Malt-4) CO2 removal
(Beer-1,D) Yeast cell removal (Beer-8)
Grade 597½ 10311853 G7471143
Grade 2555½ 10313953 G5137158
Removal of turbidity from wort
Two batches of malt were prepared to support testing of the filtration step for method ASBC Malt-4. The malt was prefiltered prior to further testing in order to isolate the wort. Spent grain was removed by prefiltering the malt using a coarse filter bag. The wort in the filtrate was adjusted to a turbidity level of 10 prior to measuring the base turbidity.
A 150 ml sample of wort was filtered using gravity through Whatman filter paper Grade 2V, 597½, or 2555½. The turbidity pre- and post-filtration was measured, and the percent reduction calculated. This process was repeated for a total of three samples. The filtration times were also recorded.
CO2 removal from bottled beer
The amount of CO2 in beer is expressed as mg/ml and is derived from the following formula:
C x (V-V0) x 44 × V1 +1
10               V1
Where:
C = concentration (M) of HCl standard
V0 = volume of HCl (ml) required to adjust pH in a water blank to 3.9
V1 = sample volume (ml)
V = volume of HCl (ml) required to adjust pH of the sample to 3.9
Per ASBC method Beer-1,D a 4 ml sample of beer chilled to 4°C was filtered using gravity through Whatman Grade 2V, 597½, or 2555½ and transferred to a conical flask. NaOH (1 ml of a 10 M stock) was added to the beer and mixed thoroughly. A 10 ml sample of this mixture was transferred to a fresh beaker and 20 ml of distilled water added. The mixture was then titrated against 0.5 M HCl, with the volume required to reduce the pH to 3.9 recorded. The measurement was repeated with 50 ml of unfiltered beer and then with 50 ml of water (as a blank control).
The amount of CO2 in unfiltered and filtered beer samples was measured, and the amount and percentage of CO2 removed was calculated. This process was repeated for a total of three samples.
Removal of yeast cells after fermentation
A typical fermentation broth containing yeast cells suspended in solution was prepared. In order to test such a broth for acidity per ASBC method Beer-8 such a broth would first have yeast cell content removed. The initial number of suspended yeast cells was estimated using a cell counting chamber. Aliquots of 150 ml of this suspension were then filtered by gravity through one of the three filter paper grades. The filtrate was subjected to serial dilution.
The resulting samples were plated on YPD agar plates to determine the number of viable yeast cells not retained by the filter. The difference between the two counts was used to determine the efficiency of filtration. This process was repeated for a total of three samples.
Results and discussion
Removal of turbidity from wort
The results for the two prefiltered wort batches are provided in Table 3.
Table 3. Turbidity reduction for wort batches. Results are the average of triplicate measurements
Whatman filter grade Initial turbidity Turbidity after filtration Turbidity reduction (%)
  Batch 1 Batch 2 Batch 1 Batch 2 Batch 1 Batch 2
2V   10.6   30.1 6.54 6.39 38.3 78.8
597½ 8.02 7.25 24.3 75.9
2555½ 7.24 8.96 31.7 70.2
Although Grade 2V was superior for turbidity removal, filtration time was much longer than with the other two papers (up to 413 s for a 120 ml sample compared to 83 s and 52 s for Grades 597½ and 2555½, respectively). The 597½ grade paper showed better performance characteristics with higher turbidity starting material, and Grade 2555½ performed better when initial turbidity was low. Filter selection ultimately depends on the time allotted to perform the test and the required level of sample clarity. Table 3 and the filtration times should allow for an informed choice with respect to these parameters.
CO2 removal from bottled beer
Results for CO2 removal are given in Table 4. The Grade 2V paper showed the highest efficiency in removing CO2 from the beer, indicating its particular suitability for this analysis.
Table 4. CO2 removal from bottled beer by Whatman paper type. Results are the average of triplicate measurements.
Filter grade CO2 (mg/ml) CO2, removed (mg/ml) % removal
  Unfiltered Filtered    
2V   4.75 1.06 3.69 77.68
597½ 1.94 2.81 59.16
2555½ 2.07 2.68 56.42
Removal of yeast cells after fermentation
Table 5 provides results for yeast cell removal from fermentation broth. All three papers performed well in the test, removing greater than 99.5% of suspended yeast cells from the broth. Whatman Grade 2V filter paper achieved 99.99% removal.
Table 5. Yeast cell removal from fermented beer. Results are the average of triplicate measurements
  Filter grade   Yeast cells (number/ml)   % removal
  Before filtration After filtration  
2V   2 x 107 1.9 × 103 99.991
597½ 4.3 × 104 99.783
2555½ 2.5 × 104 99.874
Conclusions
Cytiva produces a wide range of Whatman filter papers suitable for use in the beer industry. A sample of this range, Whatman Grades 2V, 597½, and 2555½, were tested in support of three representative quality control procedures from ASBC. All three grades of paper performed well in removing turbidity, CO2, and yeast cells. Whatman Grade 2V in particular stands out as providing the highest rate of turbidity reduction and CO2 and yeast cell removal from samples. However, use of Grade 2V comes at a cost of a longer time to filter. The data presented in this application note will be useful to brewers deciding what filtration papers to use given their resource, time, and quality constraints.
Materials
References
  1. Jacob, F. ed. Wort, beer, beer-based beverages: collection of brewing analysis methods of the Mitteleuropäische Brautechnische Analysenkommission/MEBAK. Freising-Weienstephan: MEBAK (2013).
  2. Kühbeck et al. Influence of Lauter turbidity on wort composition, fermentation performance and beer quality – a review. J Inst. Brew. 112, 215–221 (2006).
Related Links