Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
2,4,6-Trimethylbenzoyl chloride
Aldrich ChemFiles 2007, 7.3, 6.
Aldrich ChemFiles 2007, 7.3, 6.
Whereas acid chlorides might typically be thought of as building blocks, they can also be used to protect alcohols as their corresponding esters. 2,4,6-Trimethylbenzoyl chloride is a particularly useful acid chloride in this regard, as it forms the mesitoate esters. These esters are very stable to base hydrolysis, yet can be easily cleaved reductively with LiAlH4. 1,2
back to top
Materials
References
  1. Corey, E. J. et al. J. Am. Chem. Soc. 1969, 91, 4318. (b) Bolton, I. J. et al. J. Chem. Soc. C 1971, 2944.
  2. Greene, T. W.; Wuts, P. G. M. In Protective Groups in Organic Synthesis, 3rd ed.; John Wiley & Sons: New York, 1999; p.178–179.