Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
2-Chlorotrityl Resins Preloaded with N-α-Fmoc-Protected Amino Acids
Chemfiles Volume 3 Article 4
The mild cleavage conditions for highly acid-labile 2-chlorotrityl resin enables the preparation of fully protected peptide fragments for convergent synthesis and selective side-chain derivatization. Because the amino acids attached to the 2-chlorotrityl resin are N-terminal-free, these resins are ready for the coupling reaction without any deprotection pre-treatments. The polymer matrix for our preloaded 2-chlorotrityl resins is polystyrene cross-linked with 1% DVB.