Asymmetric Aldol Reaction Using Bis-ProPhenol Ligands
By: William Sommer, Aldrich ChemFiles 2008, 8.6, 10.
Aldrich ChemFiles 2008, 8.6, 10.
The aldol reaction is an important tool for the synthesis of complex molecules. However, the chemo- and regioselectivity of this reaction is usually poor. In view of these challenges, Trost and co-workers developed a new ligand capable of performing aldol reactions enantioselectively.1 The approach taken by the researchers was to look at the different requirements for a good catalyst for asymmetric transformations. The final structure of the catalyst is a semi-crown complex which allows good chiral recognition as well as a path for easy exchange of product. The ligand structure is based on a p-cresol backbone. To probe the activity of the ligand, various aldehydes were reacted with various aryl ketones with overall good yields and excellent selectivity (Scheme 1).
Scheme 1.
back to top 
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2000, 122, 12003.