Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Asymmetric Epoxidation of Allylic Alcohols | Technical Article
By: William Sommer, Aldrich ChemFiles 2008, 8.6, 5.
Aldrich ChemFiles 2008, 8.6, 5.
Catalytic asymmetric epoxidation of olefins has become a reaction of choice to generate various chiral building blocks used in the synthesis of natural products and biogically active molecules. Yamamoto and co-workers developed bishydroxamic acid based ligands for the efficient asymmetric epoxidation of various allylic alcohols. Using only 1 mol% of catalyst (derived from vanadium and the bishydroxamic acid ligand), a variety of allylic alcohols were reacted to form enantiopure epoxides with high selectivity.
back to top 
Materials
References
  1. Zhang, W. et al. Angew, Chem., Int. Ed. 2005, 44, 4389.