Asymmetric Reduction of Ketones
By: William Sommer, ChemFiles 2008, 8.6, 5.
ChemFiles 2008, 8.6, 5.
The enantioselective reduction of ketones to alcohols gives access to a pool of chiral building blocks that can be used for the synthesis of natural products. Adolfsson and co-workers reported a novel class of ligands, based on a pseudo-dipeptide,1 for the mild and efficient reduction of ketones using 2-propanol as a hydrogen source. The ligand is used with [{RuCl2(p-cymene)}2] in presence of NaOH for the asymmetric reduction of a variety of acetophenone derivatives. Excellent yields and ee’s were reported (Scheme 1).
Scheme 1.
back to top 
Materials
References
  1. Bøgevig, A. et al. Chem. Eur. J. 2004, 10, 294.; Västilä, P. et al. Chem. Commun. 2005, 4039.