C–H Activation Reactions
By: Troy Ryba, ChemFiles Volume 11 Article 1
Troy Ryba, Ph.D.
Product Manager
Cationic palladium in combination with stoichiometric oxidant benzoquinone and silver nitrate successfully catalyzes ortho-functionalization of a variety of aryl acetamides in water at room temperature using this amphiphile (Scheme 1).
Scheme 1: Selected C–H activation reaction.
back to top 
  1. Lipshutz, B. H.; Ghorai, S. Aldrichimica Acta 2008, 41, 59.