Chiral Alcohols
ChemFiles 2005, 5.4, 11.
We offer a wide range of chiral alcohols to meet their ever-growing demand. These useful reagents may serve both as starting materials in the synthesis of single-stereoisomer drugs or intermediates, or as powerful resolving agents.
2-Hexanol
(R)-(–)-2-Hexanol and (S)-(+)-2-hexanol were used in the preparation of some key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1 (Scheme 22). By comparing the NMR chemical shifts of the synthesized model compounds with the ones of the isolated natural product, configurations of four previously unassigned stereocenters of cycloviracin B1 could be elucidated.1
Scheme 22
2-Heptanol
The (R)-enantiomer of 2-heptanol was used by Kondo et al. in resolving the diastereoisomeric mixture of a key intermediate in the synthesis of 1-(2-chloro-4-pyrrolidin-1-ylbenzoyl)-2,3,4,5-tetrahydro-1H-1-benzdiazepine, known to be a strong vasopressin V2 receptor agonist, which helps to maintain normal plasma osmolality, blood volume, and blood pressure (Scheme 23).2 A nonpeptidic V2 agonist may find use in the treatment of diabetes insipidus and nocturnal enuresis.
Scheme 23
back to top 
Materials
References
  1. Fürstner, A. et al. J. Am. Chem. Soc. 2003, 125, 13132.
  2. Kondo, K. et al. J. Med. Chem. 2002, 45, 3805.