Chiral Diols
Aldrich ChemFiles 2005, 5.4, 12.
Aldrich ChemFiles 2005, 5.4, 12.
2,5-Hexanediol and 3,6-Octanediol
Enantiomerically pure C2-symmetrical alkanediols such as (2S,5S)-, and (2R,5R)-hexanediol are valuable building blocks for the synthesis of chiral ligands such as (2S,5S)-dimethylpyrrolidine or (2S,5S)-dimethylthiolane (Scheme 24, R = Me). (3R,6R)-Octanediol was used as a convenient starting material for the preparation of enantiomerically pure bis(phospholano)ethane ligands (Scheme 24, R = Et). Their use in Rh-catalyzed highly enantioselective hydrogenation reactions was described by Burk et al.1
Scheme 24
Huang et al. reported a concise asymmetric total synthesis of (2S,3S,7S)-3,7-dimethylpentadecan-2-yl acetate and propionate, the sex pheromones of the pine sawflies.2 Starting from (S)-2-Methyl-1,4-butanediol, these pheromones are synthesized efficiently as depicted in Scheme 25.
Scheme 25
cis-4-Cyclopentene-1,3-diol 1-acetate
(1R,3S)-(+)-cis-4-Cyclopentene-1,3-diol 1-acetate has proven to be a valuable precursor for the synthesis of carbocyclic nucleosides and prostaglandins (Scheme 26).3
Scheme 26
back to top 
  1. Burk, M. J. et al. J. Am. Chem. Soc. 1993, 115, 10125.
  2. Huang, P.-Q. et al. J. Org. Chem. 2004, 69, 3964.
  3. (a) Theil, F. et al. J. Chem. Soc., Perkin Trans. I 1996, 255; (b) Myers, A. G. et al. Tetrahedron Lett. 1996, 37, 3083; (c) Paquette, L. A. et al. Org. Synth. 1995, 73, 36.