Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
ChiroSolvTM Kits
Chemfiles Volume 7 Article 3
ChiroSolvTM kits are 96-well format kits that enable rapid screening of resolving agents and solvents for chiral separation. Each of these ready-to-use disposable kits contains a combination of eight resolving agents and twelve solvents. Racemates that can be separated include: acids, bases, alcohols, amino acids, aldehydes, and ketones. Optimum resolution that typically requires over two months can be achieved within one day. ChiroSolvTM Kits
Available in three Acid Series kits (681431, 681423, 681415 ) and three Base Series kits ( 681407 , 681393 , 681377).