Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Cross-Couplings in Water
By: Troy Ryba, ChemFiles Volume 11 Article 1
Conducting transition metal-catalyzed cross-coupling chemistry in water instead of organic solvent has a number of potential benefi ts in terms of cost, environmental impact, safety, and impurity profiles. Increasing focus on the “green-ness” of chemical processes has further promoted recent developments in this field. The actual means of implementing reactions in water, however, especially at room temperature and for water-insoluble organic substrates, has not always been clear. One solution that has been applied to a broad range of transition metal-catalyzed processes is the use of small amounts of a nanomicelle-forming amphiphile in water, which provides a lipophilic medium in which cross-coupling reactions can take place.
back to top Back to Top
Related Links