By: William Sommer, Aldrich ChemFiles 2009, 9.5, 7.
Aldrich ChemFiles 2009, 9.5, 7.
Following their work developing chiral gold complexes for the enantioselective hydroamination, Toste and coworkers looked at the [2+2] cycloaddition of allenenes.1 Resulting from this work was the first catalyzed cycloisomerization of allenenes to alkylidenes-cyclobutanes. The cycloaddition reaction yields enantioenriched bicyclo[3.2.0]- structures. In a typical reaction, 3 mol % of gold catalyst is used with 6 mol % of AgBF4 in methylene chloride at room temperature. Good yields and selectivity are obtained for the various bicyclic products (Scheme 1).
Scheme 1.
More recently, the Toste group reported the [4+2] and [4+3] cycloaddition of allene dienes.2 Using a triarylphosphite-gold complex, a series of allene-dienes were reacted. The reaction showed good tolerance for substituents on the diene. Using 5 mol % of the gold catalyst with 5 mol % of AgSbF6 in methylene chloride at room temperature a variety of cycloadducts containing a quaternary carbon were obtained in good yields (Scheme 2). It is important to note that all reactions were diastereoselective affording exclusively the trans-fused cycloadducts in good yields.
Scheme 2.
back to top 
  1. Luzung, M. R. et al. J. Am. Chem. Soc. 2007, 129, 12402.
  2. Mauleón, P. et al. J. Am. Chem. Soc. 2009, 131, 6348.